|
Reagents and Equipment Procedure Troubleshooting Guide References
The electrophoretic transfer of proteins from sodium dodecyl sulphate polyacrylamide gels (SDS-PAGE) to sheets of nitrocellulose was initially referred to as "Western" blotting. In order to avoid this geographical jargon, the term immunoblotting is used.
Reagents and Equipment
- 0.3 M TRIZMA® base (Product No. T1503), 20% methanol.
- 0.025 M TRIZMA® base (Product No. M1770), 20% methanol.
- 0.02 M TRIZMA® base, 20% methanol, 5.25 mg/ml epsilon-amino caproic acid (Product No. A2504).
- Tris buffered saline (TBS) (Product No. T6664).
- Tris buffered saline with 0.05% Tween 20 (TBS/Tween) (Product No. T9039).
- Bovine serum albumin (BSA) (Product No. A9647).
- Nitrocellulose (NC) membrane 0.45 5m pore size. 0.2 5m pore size nitrocellulose membrane may be used for low molecular weight molecules.
- Blotting paper, extra thick.
- Primary antibody.
- Alkaline phosphatase-conjugated secondary antibody.
- Phosphate buffered saline with 0.05% Tween 20 (PBS/Tween) (Product No. P3563).
- SIGMA FAST™ BCIP/NBT Alkaline Phosphatase Substrate Tablets (Product No. B5655 or B1911 Liquid BCIP/NBT).
- Minigel system (Product No. B2157).
- 2000 volt power supply.
- Semi-dry blotter (Product No. B2529).
- Orbital shaker (Product No. Z367605).
- Troughs 2-3 mm depth, 7-10 mm wide.
Procedure
SDS-PAGE Protein Blotting Immuno-Detection
SDS-PAGE
- Carry out SDS-PAGE (cassette size approx. 80 x 80 mm). For analytical runs, load 5-20 micrograms of protein per well; for preparatory purposes, use 600-800 micrograms of total cell or tissue homogenate per gel. Apply constant voltage at 70-140 V or 1-2 hours until the tracking dye migrates 1.0 cm from the gel end. Many simple techniques for preparing gel slabs have been described in the literature and in the book: ''Disc electrophoresis and related techniques of polyacrylamide gel electrophoresis'' by H.R. Maurer (de Gruyter; Berlin, New York, 1971).
Protein Blotting
- Build the transfer "sandwich" onto the anode(+) plate as follows: 2 sheets blotting paper soaked in 0.3 M TRIZMA base, 20% methanol, 1 sheet blotting paper soaked in 0.025 M TRIZMA base, 20% methanol, NC membrane pre-wet with deionized water, Slab gel, 3 sheets blotting paper soaked with 0.02 M TRIZMA. base, 20% methanol, 5.25 mg/ml epsilon-aminocaproic acid.
- Carry out the transfer at 90 mA for 1.5 hours at room temperature.
- Remove the NC membrane from the apparatus and air dry the NC blot thoroughly.
Note: The dry membrane may be stored at 2-8 °C between two sheets of blotting paper in a plastic sleeve.
Immuno-Detection
The following incubation and washing steps are carried out at room temperature on an orbital shaker platform. Primary antibody is diluted in 1% normal serum from the secondary antibody host animal in TBS. Other non-interfering proteins (e.g., BSA, hemoglobin, ovalbumin) may be substituted.
- Cut the membrane into strips. Place the NC strips, with the side in contact with the gel during the transfer facing up, in the troughs.
- Block NC blot using 5% w/v BSA in TBS, overnight at 4 °C or 2 hrs. at room temperature. The choice of blocking reagent depends on the type of probe that will be subsequently used in the overlay procedure and should be chosen accordingly.
- Remove the blocking buffer.
- Overlay the blot with 5 ml primary antibody at an appropriate dilution (generally 1:50-1:500). Incubate for 1-3 hours at room temperature on shaker.
- Wash the NC strips four times for 5 minutes each, with sufficient TBS/Tween.
- Incubate the strips for 1 hour in alkaline phosphatase-secondary antibody conjugate at an appropriate dilution. (1:30,000 for upgraded alkaline phosphatase conjugates) in TBS/Tween.
- Wash the strips as in step 4. Rinse 3 times in TBS for 5 minutes each.
- Prepare SIGMA FASTTM BCIP/NBT Tablets according to package directions. Liquid BCIP/NBT substrate may be used (Product No. B1911).
- Incubate the NC strips in the substrate mixture for 10-30 minutes until color development.
- Stop the reaction by washing the strips in several changes of distilled water.
- Air dry the strips and store in the dark in a plastic sleeve.
Note: The dry membrane may be stored at 2-8 °C between two sheets of blotting paper in a plastic sleeve.
References
- Burnette, W.N., Anal. Biochem., 112, 195-203 (1981).
- Hames, B.D., and Rickwood, D. (eds.), "Gel Electrophoresis of Proteins: A Practical Approach", IRL Press Ltd., Oxford (1981).
- Gershoni, J.M., and Palade, G.E., Anal. Biochem., 131, 1-15 (1983).
back to Antibodies Protocols back to top
|