|
CF™ Dyes: Enhanced Fluorophores with Novel Dye Chemistry
CF dyes are a series of highly water-soluble fluorescent dyes spanning the visible and near-infrared (near-IR) spectrum (Figures 1A and 1B) for labeling antibodies, proteins, nucleic acids, and other biomolecules. Developed by scientists using new breakthrough chemistries, the brightness, photostability, and color selection of CF dyes rival or exceed the quality of other commercial dyes as a result of rational dye design.
Fluorescent Dye Comparison Chart
Figure 1A
 Figure 1B
Achieve greater brightness, photostability, and sensitivity with CF dyes
Sigma currently offers 19 CF dyes spanning the visible and near-IR wavelengths, with additional colors in development. The CF dye product line includes reactive CF dyes, labeling kits, CF-labeled secondary antibodies, and other bioconjugates. This collection further expands Sigma’s broad range of carefully selected secondary antibodies and conjugates, allowing scientists to achieve greater sensitivity, brighter results, and better photostability in immunoassays (Figures 2A, 2B, and 2C).
 |
| Figure 2A. Relative fluorescence of CF543 and Alexa Fluor 546 (AF546) goat anti-mouse conjugates as a function of the number of dye molecules per protein (degree of labeling). |
|
Figure 2B. Relative photostability of CF633 and Alexa Fluor 647 (AF647) goat anti-mouse conjugates. Jurkat cells were fixed, permeabilized and stained with rabbit anti-CD3 followed by CF633 or Alexa Fluor 647 goat anti-rabbit IgG conjugates. Cells were imaged using a mercury arc lamp microscope equipped with a Cy5 filter set and CCD camera. Sequential images were captured at 0, 1, and 5 minutes. |
|
Figure 2C. Near-IR CF dyes are highly water soluble without carrying excessive negative charge, which can increase non-specific binding of antibody conjugates. Near-IR western blots imaged using the Odyssey system (Li-COR Biosciences). Two dilutions of HeLa cell lysate were probed with mouse anti-tubulin antibody followed by goat anti-mouse conjugated to Alexa Fluor 790 (AF790) (2, 3) or CF790 (5, 6). |
To help researchers evaluate their secondary antibody products with complete peace of mind, Sigma is offering the Bioguarantee on all CF dye labeled antibody products.
Fluorescent Dye Comparison Chart
| CF™ dye |
Reactive forms |
λEx (nm) |
λEx (nm) |
MW |
ε |
Excitation source |
Direct replacement for: |
Reactive forms |
λEx (nm) |
λEm (nm) |
MW |
ε |
Advantages of CF™ dye |
| CF™ 350 |
amine, aminooxy, hydrazide, maleimide, SE |
347 |
448 |
496 |
18,000 |
UV |
AMCA |
NHS, sulfo-NHS, hydrazide, HPDP, SE |
353 |
442 |
330 |
19,000 |
- Yields the brightest blue fluorescent antibody conjugates when excited at ~350 nm - Highly water-soluble and insensitive to pH |
| Cascade Blue® |
Acetyl azide, hydrazid, ethylenediame |
400 |
420 |
596 |
28,000 |
| DyLight™ 350 |
NHS, maleimide |
353 |
432 |
899 |
15,000 |
| CF™ 405S |
amine, aminooxy, maleimide, SE |
404 |
431 |
1169 |
33,000 |
405 nm laser |
Alexa Fluor® 405 |
SE, cadaverine |
400 |
424 |
1028 |
35,000 |
- Much brighter than Alexa Fluor® 405 due to better compatibility with excitation and emission windows on common instruments |
| DyLight™ 405 |
NHS, maleimide |
400 |
420 |
793 |
30,000 |
- Highly water-soluble and insensitive to pH |
| CF™ 405M |
aminooxy, maleimide, SE |
408 |
452 |
503 |
41,000 |
405 nm laser |
BD Horizon™ V450 |
Conjugated |
404 |
448 |
|
|
- More photostable than Pacific Blue® dye - As bright as Pacific Blue® dye in the blue channel - Less spill-over fluorescence in the green channel - Highly water-soluble |
| eFluor® 450 |
Conjugated |
405 |
450 |
|
|
| Pacific Blue® |
SE |
405 |
455 |
406 |
46,000 |
| CF™ 488A |
amine, aminooxy, hydrazide, maleimide, SE |
490 |
515 |
914 |
70,000 |
488 nm laser |
Alexa Fluor® 488 |
5-SDPE, 5-TFPE, maleimide, alkyne, azide, cadaverine, hydrazide, hydroxylamine, SE |
494 |
517 |
643 |
73,000 |
- Minimal charge compared to Alexa Fluor® 488 reduces nonspecific antibody staining - Less spill-over fluorescence in the red channel than Alexa Fluor® 488 - Extremely photostable - Highly water-soluble and pH-insensitive |
| Cy 2 |
Bis NHS |
489 |
506 |
714 |
150,000 |
| DY-495 |
NHS, maleimide, carboxylic acid, amino derivative |
493 |
521 |
623 |
70,000 |
| DyLight™ 488 |
NHS, maleimide |
493 |
518 |
1011 |
70,000 |
| FAM |
SE |
495 |
520 |
|
83,000 |
| FITC |
|
494 |
518 |
289 |
77,000 |
| Fluorescein |
NHS, maleimide |
490 |
514 |
332 |
93,000 |
| CF™ 543 |
amine, hydrazide, maleimide, SE |
541 |
560 |
870 |
100,000 |
532 nm or 543, 546 laser |
Alexa Fluor® 546 |
N/A |
556 |
573 |
N/A |
112,000 |
- Significantly brigher than Alexa Fluor® 546 - Highly water-soluble and pH-insensitive |
| CF™ 555 |
amine, hydrazide, maleimide, SE |
555 |
565 |
810 |
150,000 |
532 nm or 568 nm laser |
Alexa Fluor® 555 |
maleimide, alkyne, azide, cadaverine, hydrazide, SE |
555 |
572 |
1250 |
155,000 |
- Brighter and more photostable than Cy 3 - Minimal charge compared to Alexa Fluor® 555 reduces nonspecific antibody staining |
| Cy 3 |
Bis NHS, hydrazide, maleimide, NHS |
552 |
566 |
767 |
150,000 |
| DY-547 |
NHS, maleimide, carboxylic acid, amino derivative |
557 |
574 |
739 |
150,000 |
| DyLight™ 550 |
NHS, maleimide |
562 |
576 |
1040 |
150,000 |
| TRITC |
|
557 |
576 |
479 |
100,000 |
| CF™ 568 |
amine, aminooxy, hydrazide, maleimide, SE |
562 |
583 |
714 |
100,000 |
532 nm or 568 nm laser |
Alexa Fluor® 568 |
maleimide, cadaverine, hydrazide, SE |
578 |
602 |
792 |
88,000 |
- Optimal for the 568 nm line of the Ar-Kr mixed-gas laser - Brighter and more photostable than Alexa Fluor® 568 |
| DY-560 |
NHS, maleimide, carboxylic acid, amino derivative |
559 |
578 |
769 |
120,000 |
| Rhodamine Red |
NHS |
570 |
590 |
654 |
129,000 |
| CF™ 594 |
amine, aminooxy, hydrazide, maleimide, SE |
593 |
614 |
729 |
115,000 |
532 nm or 594 nm laser |
Alexa Fluor® 594 |
maleimide, alkyne, azide, cadaverine, hydrazide, SE |
590 |
617 |
820 |
92,000 |
- Yields the brightest antibody conjugates among spectrally similar dyes - Extremely photostable |
| DY-594 |
NHS, maleimide |
589 |
614 |
|
92,000 |
| DyLight™ 594 |
NHS, maleimide |
593 |
618 |
1078 |
80,000 |
| Texas Red® |
SE |
595 |
615 |
702 |
116,000 |
| CF™ 620R |
maleimide, SE |
617 |
639 |
728 |
115,000 |
633 nm or 635 nm laser |
LightCycler® |
Red 640 |
NHS |
625 |
640 |
758 |
- Excellent energy acceptor for FRET - Highly fluorescent - Extremely photostable - Highly water-soluble |
| CF™ 633 |
amine, aminooxy, hydrazide, maleimide, SE |
630 |
650 |
821 |
100,000 |
633 nm or 635 nm laser |
Alexa Fluor® 633 |
maleimide, hydrazide, SE |
621 |
639 |
1200 |
159,000 |
- Yields the brightest antibody conjugates among spectrally similar dyes Optimal for 633 nm He-Ne laser or the 635 nm red diode laser - Far more photostable than Alexa Fluor® 647 - Highly water-soluble |
| DY-630 |
NHS, maleimide, carboxylic acid, amino derivative |
636 |
657 |
732 |
200,000 |
| DyLight™ 633 |
NHS, maleimide |
638 |
658 |
1066 |
170,000 |
| CF™ 640R |
amine, aminooxy, hydrazide, maleimide, SE |
642 |
662 |
832 |
105,000 |
633 nm, 635 nm, or 640 nm laser |
Alexa Fluor® 647 |
maleimide, alkyne, azide, cadaverine, hydrazide, SE |
651 |
672 |
1250 |
270,000 |
- Most photostable among spectrally similar dyes - Yields highly fluorescent protein conjugates - Very water-soluble and pH-insensitive |
| Cy 5 |
Bis NHS, hydrazide, maleimide, NHS |
649 |
666 |
792 |
250,000 |
| CF™ 647 |
hydrazide, maleimide, SE |
650 |
665 |
836 |
240,000 |
633 nm, 635 nm, or 640 nm laser |
Alexa Fluor® 647 |
maleimide, alkyne, azide, cadaverine, hydrazide, SE |
651 |
672 |
1250 |
270,000 |
- Antibody conjugates have the best signal-to-noise ratio compared to Alexa Fluor® 647, DyLight™ 649 and Cy 5 - Highly fluorescent - Highly water-soluble and pH-insensitive |
| Cy 5 |
Bis NHS, hydrazide, maleimide, NHS |
649 |
666 |
792 |
250,000 |
| DY-647 |
NHS, maleimide, carboxylic acid, amino derivative |
653 |
672 |
762 |
250,000 |
| DyLight™ 650 |
NHS, maleimide |
652 |
672 |
1066 |
250,000 |
| CF™ 660C |
maleimide, SE |
667 |
685 |
3112 |
200,000 |
633 nm, 635 nm, or 640 nm laser |
Alexa Fluor® 660 |
maleimide, SE |
668 |
698 |
1100 |
132,000 |
- Much brighter than Alexa Fluor® 660 - More photostable than Alexa Fluor® 660 - Highly water-soluble |
| CF™ 660R |
aminooxy, maleimide, SE |
663 |
682 |
888 |
100,000 |
633 nm, 635 nm, or 640 nm laser |
Alexa Fluor® 660 |
maleimide, SE |
668 |
698 |
1100 |
132,000 |
- Brighter than Alexa Fluor® 660 - The most photostable 660 nm dye, ideal for confocal microscopy - Highly water-soluble |
| CF™ 680 |
maleimide, SE |
681 |
698 |
3241 |
210,000 |
680 nm or 685 nm laser |
Alexa Fluor® 680 |
maleimide, SE |
684 |
707 |
1150 |
183,000 |
- The brightest among spectrally similar dyes - Superior signal-to-noise ratio in imunostaining - Highly water-soluble and pH-insensitive |
| Cy 5.5 |
Bis NHS, hydrazide, maleimide, NHS |
675 |
695 |
1128 |
250,000 |
| DyLight™ 680 |
NHS, maleimide |
692 |
712 |
950 |
140,000 |
| IRDye® 680 |
NHS, maleimide |
676 |
693 |
1402 |
250,000 |
| CF™ 680R |
aminooxy, maleimide, SE |
680 |
701 |
912 |
140,000 |
680 nm or 685 nm laser |
Alexa Fluor® 680 |
maleimide, SE |
684 |
707 |
1150 |
183,000 |
- The most photostable 680 nm dye - Suitable for labeling nucleic acids and small bio-molecules - Highly water-soluble and pH-insensitive - CF680/CF770 double labeling compatible with Li-COR Odyssey System |
| Cy 5.5 |
Bis NHS, hydrazide, maleimide, NHS |
675 |
695 |
1128 |
250,000 |
| DyLight™ 680 |
NHS, maleimide |
692 |
712 |
950 |
140,000 |
| IRDye® 680 |
NHS, maleimide |
676 |
693 |
1402 |
250,000 |
| CF™ 750 |
SE |
755 |
777 |
3009 |
250,000 |
680 nm, 685 nm, or 785 nm laser |
Alexa Fluor® 750 |
maleimide, SE |
753 |
782 |
1300 |
290,000 |
- Exceptionally bright and stable - Less immunogenic than competing dyes - Better signal-to-noise ratio compared to APC- Alexa Fluor® 750 tandem dye with 633 nm excitation |
| Cy 7 |
Bis NHS, NHS |
743 |
767 |
818 |
250,000 |
| DyLight™ 750 |
NHS, maleimide |
752 |
778 |
1034 |
210,000 |
| APC-Alexa Fluor® 750 |
Conjugated |
650 |
779 |
|
240,000 |
| CF™ 770 |
SE |
770 |
797 |
3138 |
220,000 |
785 nm laser |
DyLight™ 800 |
NHS, maleimide |
777 |
794 |
899 |
270,000 |
- Exceptionally bright and stable - Less immunogenic than competing dyes - CF680/CF770 double labeling compatible with Li-COR Odyssey System |
| IRDye® 800CW |
NHS, maleimide, carboxylate |
778 |
794 |
1166 |
300,000 |
| CF™ 790 |
SE |
784 |
806 |
3267 |
210,000 |
785 nm laser |
Alexa Fluor® 790 |
SE |
785 |
810 |
1750 |
260,000 |
- Exceptionally bright and stable - Less immunogenic than competing dyes |