Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Cell Biology > Antibodies > IsoQuick™ Kits for Mouse Monoclonal Isotyping
Antibodies
IsoQuick™ Kits for Mouse Monoclonal Isotyping
IsoQuick for Mouse Monoclonal Isotyping is fast, flexible and makes it easy to classify mouse monoclonal antibodies (MAbs)
Lateral Flow Device Strips Image
IsoQuick is based on immunoassay technology in the format of a lateral flow device (LFD). This technology is simple, robust and proven. Isoquick is available in three separate LFD strips for maximum test flexibility and cost effectiveness. IsoQuick develops results in five minutes or less with clear, easy-to-read results and control lines. The simple interpretation guide and documentation pad make IsoQuick results easy to interpret and document in lab notebooks or project files.
Strips are ready to use directly from the tube. No extra tubes, buffers or controls are required. They can be used in cell supernatant with no dilution step. When testing ascites or purified antibody, a simple dilution step is required.
IsoQuick strips are packaged in kits with each of the three strips or they are available separately in multiples of 5 and 25 strips.
  Red Square Bullet Flexibility—Offered in a 3-strip configuration so you only test what you need
  Red Square Bullet Speed—Results are displayed in 5 minutes
  Red Square Bullet Convenient and Simple—Use directly out of the tube. No dilutions required with cell supernatant or purified MAbs. Simple dilution for use with ascites
  Red Square Bullet Accurate—Specific for each isotype/subtype and has sensitivity equal to or better than other isotype LDFs on the market
  Red Square Bullet Cost Effective—Available individually or in kits so you're only using the tests that you need
Frequently Asked Questions
Ordering Information
Description Product No. Number of Applications Add to Cart
Kits
IsoQuick Kit for Mouse Monoconal Isotyping ISOQ5 Sufficient for 5 applications Shopping Cart
ISOQ25 Sufficient for 25 applications Shopping Cart
Standalones
IgG1, IgG2a, IgG2b, IgG3 I9285-5EA Sufficient for 5 applications Shopping Cart
I9285-25EA Sufficient for 25 applications Shopping Cart
IgA, IgM I9410-5EA Sufficient for 5 applications Shopping Cart
I9410-25EA Sufficient for 25 applications Shopping Cart
Kappa, Lambda I9535-5EA Sufficient for 5 applications Shopping Cart
I9535-25EA Sufficient for 25 applications Shopping Cart
IsoQuick is a trademark of EnviroLogix Inc.
back to top