Sigma-Aldrich
Life Science > Cell Biology > Antibodies > Learning Center > Antibody Explorer > Kits
Antibody Explorer
Kits
Antibody Explorer Home
  ELISA Kits
Immunohistology Kits
Western Blotting Kits
ExtrAvidin Staining Kits
Antibody Microarrays
Conjugation Kits
Antibody Purification Kit
Immunoprecipitation Kits
Antibody Isotyping Kits
Antibody Labeling
PKH and CellVue® Fluorescent Cell Linkers
Sigma offers a variety of unique kits to support immunochemistry and immunodetection applications including immunogenicity enhancement, antibody purification and isotyping and probe signal amplification.
The study of the proteosome is a rapidly evolving field that frequently requires dissection of structural proteins, enzymes and cell signaling molecules into functional or structural domains involving relatively short amino acid sequences. Very often these peptides (haptens) are not immunogenic unless they are conjugated to larger molecules such as bovine serum albumin (BSA) or keyhole limpet hemocyanin (KLH). Sigma offers antibody conjugations kits that contain maleimide activated BSA and KLH which support the induction of immunogenicity of small sulfhydryl containing haptens.
Sigma provides kits to facilitate purification of polyclonal and monoclonal antibodies and epitope-tagged proteins (FLAG tagged). These kits utilize the antibody binding features of protein A to facilitate antibody purification from serum, ascites or cell culture supernatants and protein G for purification of antibodies immunoprecipitated (IP) from complex cell lysates. FLAG tagged proteins are recovered using a high affinity ANTI-FLAG M2 antibody agarose matrix.
When choosing detection reagents or developing a purification scheme it is generally useful to know the target antibody’s isotype or subclass. We offer isotyping kits for determination of an antibody’s subclass with simple immunodiffusion (Ouchterlony) or ELISA assays.
The use of avidin biotin based probes for immunoassay detection systems is a well established method for enhancing (amplifying) probe signals. We offer avidin-biotin based alkaline phosphatase and peroxidase system that utilizes a unique form of avidin, ExtrAvidin® that combines high specificity and affinity for avidin with low non-specific binding at physiological pH.
Elisa Kits
Product Name Product No.
cAMP Enzyme Immunoassay Ki   CA201
cAMP Enzyme Immunoassay Kit, Direct   CA200
cGMP Enzyme Immunoassay Kit   CG201
cGMP Enzyme Immunoassay Kit, Direct   CG200
Protein Tyrosine Kinase (PTK) Assay Kit   PTK101
Immunohistology Kits
Product Name Host Product No.
Monoclonal Anti- Actin, a-Smooth Muscle, Immunohistology Kit mouse IMMH2
Monoclonal Anti-Desmin Immunohistology Kit mouse IMMH5
Anti-Laminin Immunohistology Kit rabbit IMMH7
Anti-S-100 Immunohistology Kit rabbit IMMH9
Monoclonal Anti-Vimentin Immunohistology Kit mouse IMMH10
Western Blotting Kits    
Product Name Product No.
ProteoQwestä Chemiluminescent Western Blotting Kit, CPS Substrate   PQ0201
ProteoQwestä Colorimetric Western Blotting Kit, TMB Substrate   PQ0101
ExtrAvidin Staining Kits    
Product Name Host Product No.
Goat ExtrAvidin® Peroxidase Staining Kit   mouse EXTRA1
Mouse ExtrAvidin® Peroxidase Staining Kit goat EXTRA2
Rabbit ExtrAvidin® Peroxidase Staining Kit goat EXTRA3
Rat ExtrAvidin® Peroxidase Staining Kit rabbit EXTRA6
Mouse ExtrAvidin® Alkaline Phosphatase Staining Kit goat EXTRA2A
Rabbit ExtrAvidin® Alkaline Phosphatase Staining Kit goat EXTRA3A
Rapid Staining Kits  
Product Name Host Product No.
Mouse Rapid Staining Kit goat QUIK1
Conjugation Kits    
Product Name Product No.
Maleimide Activated BSA, KLH Conjugation Kit   MBK1
Antibody Purification Kit  
Product NameHost Product No.
Protein A Antibody Purification Kit   PURE1A
Immunoprecipitation Kits    
Product Name Product No.
Protein G Immunoprecipitation Kit   IP50
FLAG® Immunoprecipitation Kit   FLAGIPT1
Antibody Isotyping Kits    
Product Name Product No.
Sigma ImmunoType™ Kit   ISO1
Mouse Monoclonal Antibody Isotyping Reagents   ISO2
Antibody Labeling    
Product Name Product No.
BiotinTag™ Micro Biotinylation Kit   BTAG
ImmunoProbe™ Biotinylation Kit   BK101
FluoroTag™ FITC Conjugation Kit   FITC1
PKH and CellVue® Fluorescent Cell Linkers*    
Product Name Product No.
CellVue® Claret Far Red Fluorescent Cell Linker MIDI Kit for general cell membrane labeling MIDCLARET
CellVue® Claret Far Red Fluorescent Cell Linker MINI Kit for general cell membrane labeling MINCLARET
PKH26 Red Fluorescent Cell Linker Kit for general cell membrane labeling   PKH26GL
PKH26 Red Fluorescent Cell Linker Mini Kit for general cell membrane labeling   MINI26
PKH26 Red Fluorescent Cell Linker Kit for phagocytic cell labeling   PKH26PCL
PKH67 Green Fluorescent Cell Linker Kit for general cell membrane labeling   PKH67GL
PKH67 Green Fluorescent Cell Linker Mini Kit for general cell membrane labeling   MINI67
PKH2 Green Fluorescent Cell Linker Kit for general cell membrane labeling   PKH2GL
PKH2 Green Fluorescent Cell Linker Kit for phagocytic cell labeling   PKH2PCL
* More information on PKH and CellVue® Fluorescent Cell Linker Dyes
U.S. Patent 5,665,328 Distributed for Phanos Technologies, Inc. CellVue is a registered trademark of PTI Research, Inc. used under license.