Sigma-Aldrich
Life Science > Cell Biology > Antibodies > Learning Center > Antibody Explorer > Spotlights > Anti-Nitrotyrosine
Antibody Explorer
Anti-Nitrotyrosine
Peroxynitrite has been implicated in the pathogenesis of several inflammatory, infectious and degenerative human diseases, such as Alzheimer’s disease, amyotrophic lateral sclerosis (ALS), asthma, atherosclerosis and a variety of conditions precipitated by endothelial injury. In light of its highly reactive nature, evidence for a role of peroxynitrite in vivo is largely based on the detection of 3-nitrotyrosine, the major product of the peroxynitrite reaction, in injured tissues.
Anti-Nitrotyrosine recognizes nitrated proteins and 3-nitro-L-tyrosine. It does not cross-react with L-tyrosine, p-nitro-L-phenylalanine, 3-amino-L-tyrosine, 3-chloro-L-tyrosine and phospho-L-tyrosine BSA conjugates.
Applications: immunoblotting, ELISA, immunohistochemistry and immunofluorescence.
Click Image to View Figure in its Entirety Figure 1. Anti-Nitrotyrosine
Product Name Host Product No.
Anti-Nitrotyrosine Rabbit N0409
back to Spotlights
back to Antibody Explorer