Sigma-Aldrich
Life Science > Cell Biology > Antibodies > Learning Center > Antibody Explorer > Spotlights > Antibodies to Epitope Tags
Antibody Explorer
Antibodies to Epitope Tags
Recombinant DNA technology enables the insertion of specific foreign sequences to genes of interest. When these sequences encode short peptides they create an antigenic determinant (epitope) that can be recognized by antibodies. Thus, when the DNA sequence of interest is fused with the DNA sequence of the short peptide and introduced into cells, the resulting expressed protein is now a "tagged protein." Since antibodies to the peptide tag are commercially available, there is no need to generate specific antibodies to identify, immunoprecipitate or immunoaffinity purify the protein. Moreover, in most cases the stable fusion protein has the same bioactivity and biodistribution as the native protein.
Many different epitope tags have been engineered into recombinant proteins. These include FLAG®, HA, HIS, c-Myc, VSV-G, V5 and HSV.
Tag Sequence
HIS HHHHHH
c-MYC EQKLISEEDL
HA YPYDVPDYA
VSV-G YTDIEMNRLGK
HSV QPELAPEDPED
V5 GKPIPNPLLGLDST
FLAG DYKDDDDK
Due to the importance of this technology for protein expression, Sigma® offers a wide range of products for the detection, isolation and purification of tagged proteins. These include monoclonal and polyclonals antibodies specific for the most commonly used epitope tags, as well as antibody-enzyme conjugates, fluorescent antibodies, antibody-affinity resins and the specific peptide tag sequence. Our products are carefully tested in many applications to ensure the most highly efficient performance.
AntibodyExp_SS_EpiTag_F1sm Figure 1. Anti VSV-G Polyclonal Antibody (Product No. V4888).
AntibodyExp_SS_EpiTag_F2sm Figure 2. Anti c-Myc Agarose Affinity gel (Product No. A7470).
AntibodyExp_SS_EpiTag_F3sm Figure 3. Anti c-Myc-HRP (Product No. A5598).
AntibodyExp_SS_EpiTag_F4sm Figure 4. Anti c-Myc (Product No. C3956).
Product Table
TAG mAb Poly Ab Conjugates Resin Peptide
ALPH HRP FITC Biotin Cy3
FLAG (M1) F3040 F7425           A4596
A1080
F3290
F4799
FLAG (M2) F3165 F7425 A9469 A8592 F4049 F9291 A9594 A2220
F2426
F3290
F4799
FLAG (M5) F4042 F7425       F2922     F3290
F4799
HIS H1029   A5588 A7058       A5713  
c-MYC M5546
M4439
C3956 A5963 A5598 F2047 B7554 C6594 A7470 M2435
HA H9658 H6908 A5477 H6533 H7411 B9183   A2095 I2149
VSV-G V5507 V4888   A5977     C7706 A1970 V7887
V5 V8012 V8137   V2260       A7345 A7754
HSV   H6030             H4640
back to Spotlights
back to Antibody Explorer