Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Cell Biology > Antibodies > Learning Center > Antibody Explorer > Spotlights > Antibodies to MAGI-1, MAGI-2, MAGI-3/SLIPR
Antibody Explorer
Antibodies to MAGI-1, MAGI-2, MAGI-3/SLIPR
The MAGUK (Membrane Associated Guanylate kinase) family of proteins, whose prototypic member is PS95, is characterized by the presence of multi-PDZ and SH3 domains, and a single region of homology to Saccharomyces cerevisiae guanylate kinase (Guk) domain.1
All MAGUK studied to date localize to regions of cell-cell contact, such as tight junctions in epithelial cells and synaptic junctions in neurons, and are believed to be involved in the assembly of multiprotein complexes via their protein-protein interaction domains.2 Using different screening systems, three novel closely related MAGUK protein were isolated: MAGI-1/BAP1, MAGI-2/rSCAM and MAGI-3/SLIPR. Similar to other MAGUK members, these three proteins have the characteristic multi-PDZ domains, and a guanylate kinase domain. However, WW domain replaces the characteristic SH3 domain present in other MAGUK proteins. Different components interacting with these proteins in a complex were identified. MAGI-1 was first identified in mouse, as a protein interacting with K-RasB.3 Three isoforms were identified and named MAGI-1a, MAGI-1b and MAGI-1c, of 1139, 1171 and 1374 amino acids, respectively. MAGI-2 was initially identified in rat, as a protein interacting with N-methyl-D-aspartate receptors (NMDA-R) and neuronal cell adhesion proteins, and was named S-SCAM.4 Three isoforms of S-SCAM were identified, of 1277, 1113 and 1053 amino acids length, respectively.5 MAGI-3 (also termed SLIPR) was identified in a two hybrid screening as a protein interacting with the tumor suppressor PTEN,6 and Receptor Tyrosine Phosphatase β (RPTPβ).7 The overall homology among the MAGI proteins, especially regarding the structural domains, is high.
Sigma® has developed anti peptide antibodies that specifically recognize MAGI-1, MAGI-2 and SLIPR/MAGI-3. They are carefully tested by immunoblotting, and are able to distinguish between the three proteins.
Figure 1. Immunoblotting with anti MAGI-1, MAGI-1/S-SCAM and MAGI3/SLIPR
MAGI_page_2_smaller Figure 2. Specific recognition of MAGI isoforms by anti MAGI antibodies
MAGI_image_3_smaller Figure 3. Anti ALIPR/MAGI-3
Product Table
Description Host Product No.
Anti MAGI-1 Rabbit M5691
Anti MAGI-2 Rabbit M2441
Anti SLIPR/MAGI-3 Rabbit S1190
Anti SLIPR/MAGI-3 Mouse S4191
References
  1. Anderson, J.M., Curr. Biol., 6, 382-384 (1996).
  2. Fanning, A..S., and Anderson, J.M., Curr. Biol., 6, 1385-1388 (1996).
  3. Dobrosostakaya, I., et al, J. Biol. Chem., 272, 31589-31597 (1997).
  4. Hirao, K. et al., J. Biol. Chem., 273, 21105-21110 (1998).
  5. Hirao, K. et al., J. Biol. Chem., 275, 2966-2972 (2000).
  6. Wu, Y., et al., J. Biol. Chem., 275, 21477-21485 (2000).
  7. Adamsky, K. et al., J. Cell Sci., In press (2003).
back to Spotlights
back to Antibody Explorer
back to top