Sigma-Aldrich
Life Science > Cell Biology > Antibodies > Learning Center > Antibody Explorer > Spotlights > Antibodies to Tubulins, including Centrosome-Associated ε-Tubulin
Antibody Explorer
Antibodies to Tubulins, including Centrosome-Associated
ε-Tubulin
Microtubules of the eukaryotic cytoskeleton are composed of a heterodimer of α- and β-tubulin. In addition to α-and β-tubulin, several other tubulins have been identified, bringing the number of distinct tubulin classes to seven. Most of these tubulins have distinct subcellular localization and an emerging, diverse set of functions.1 Out of the seven different tubulins four new members of the tubulin family were identified recently, which consist of δ, χ, η, and ε-tubulin. η and ε-tubulins were discovered by database searches.2
ε-Tubulin is located in the pericentriolar area, and its localization is modulated by cell cycle progression. During the early phase of the cell cycle, the protein associates predominantly with the old centrosome, and only later does it become associated with both the old and the new centrosomes.2,3 Centrosomes can initiate microtubule assembly irrespective of ε-tubulin content, indicating that this protein is not involved in microtubule nucleation.3
A unique Monoclonal Antibody to ε-Tubulin, clone TUB-11 (Product No. T1323) is directed against the C-terminal region of ε-tubulin (amino acids 352-366 of human ε-tubulin) and may be used in ELISA and immunoblotting. The antibodies provide specific and useful tools for studying the intracellular distribution and cell cycle-dependent expression of ε-tubulin.
The innovative Monoclonal Anti-ε-Tubulin antibody complements our wide selection of antibodies to different tubulins, which include antibodies to α, β, and γ-tubulins, as well as antibodies to modified tubulins, such as acetylated-tubulin, tyrosine-tubulin and polyglutamylated tubulin.
AntibodyExp_SS_Tubulin_F1sm Figure 1. Immunoblotting with Monoclonal Anti-e-Tubulin, clone Tub-11
Product Table
Description Host Clone Product No.
Monoclonal Anti-α-Tubulin Mouse B-5-1-2 T5168
Monoclonal Anti-α-Tubulin Mouse DM1A T9026
Monoclonal Anti-α-Tubulin, FITC Conjugate Mouse DM1A F2168
Monoclonal Anti-β-Tubulin I Mouse SAP.4G5 T7816
Monoclonal Anti-β-Tubulin I+II Mouse JDR.3B8 T8535
Monoclonal Anti-β-Tubulin III Mouse SDL.3D10 T8660
Monoclonal Antiβ-Tubulin IV Mouse ONS.1A6 T7941
Monoclonal Anti-β-Tubulin Mouse 2-28-33 T5293
Monoclonal Anti-β-Tubulin Mouse D66 T0198
Monoclonal Anti-β-Tubulin Mouse TUB2.1 T4026
Monoclonal Anti-β-Tubulin, Cy3 Conjugate Mouse TUB2.1 C4585
Monoclonal Anti-β-Tubulin, FITC conjugate Mouse TUB2.1 F2043
Monoclonal Anti-γ-Tubulin Mouse GTU-88 T6557
Anti-γ-Tubulin (AK-15) Rabbit   T3320
Anti-γ-Tubulin (DQ-19) Rabbit   T3195
Anti-γ-Tubulin AIA Rabbit   T5192
Anti-γ-Tubulin, Cy3 Conjugate Rabbit   C7604
Anti-γ-Tubulin, IgG fraction Rabbit   T3559
Anti-ε-Tubulin Mouse TUB-11 T1323
Anti-Tubulin Rabbit   T3526
Monoclonal Anti-Tubulin, Polyglutamylated Mouse B3 T9822
Monoclonal Anti-Tubulin, Acetylated Mouse 6-11B-1 T6793
Monoclonal Anti-Tubulin, Tyrosine Mouse TUB-1A2 T9028
References
  1. Dutcher, S.K., et al. Curr. Opin. Cell Biol., 13, 49-54 (2001).
  2. McKean, P.G., et al. J. Cell Sci., 114, 2723-2733 (2001).
  3. Chang, P., Stearns, T. Nature Cell Biol., 2, 30-35 (2000).
back to Spotlights
back to Antibody Explorer