Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Cell Biology > Antibodies > Learning Center > Antibody Explorer > Spotlights > Anti-mTOR (mammalian target of Rapamycin): Marker for translation initiation activation
Antibody Explorer
Anti-mTOR (mammalian target of Rapamycin): Marker for translation initiation activation
mTOR (mammalian target of Rapamycin), also named FKBP12 rapamycin-associated protein (FRAP/RAFT/RAPT/SEP), is a serine/threonine protein kinase that is an evolutionary conserved member of the phosphoinositol kinase-related kinase (PIKK) family that also includes DNA-PK, ATM, ATR and several other proteins1-5. mTOR is involved in the regulation of cell growth through initiation of gene translation in response to nutrients such as amino acids (mainly leucine), growth factors, insulin and mitogens. mTOR initiates translation by activating the ribosomal p70S6k protein kinase (S6K1) and by inhibiting the eIF4E inhibitor 4E-BP1. mTOR is thought to be involved in numerous additional cellular functions including actin organization, membrane trafficking, secretion, protein degradation, protein kinase C signaling, ribosome biogenesis and tRNA synthesis1.
Rapamycin (Product No. R0395) complexes with the immunophilin FK-506 binding protein FKBP12 peptide prolyl cis/trans isomerase. Interaction of this FKBP12-rapamycin complex with mTOR inhibits its function. It has been suggested that mTOR may sense cellular ATP levels and suppress protein synthesis when ATP levels decrease6. mTOR is phosphorylated at serine 2448 via the phosphatidylinositol 3-kinase (PI3K)/Akt pathway and is autophosphorylated at serine 24817,8.
Anti-mTOR (FRAP) (Product No.T2949) is produced using a peptide corresponding to amino acids 2433-2450 of human mTOR. The corresponding sequence is identical in mouse and differs by one amino acid in rat. The antibody specifically recognizes human, mouse and rat mTOR (289 kDa) and is suitable for immunoblotting, immunoprecipitation and immunofluorescence.
T2949-iepssmall Figure 1. Immunoprecipitation - Anti-mTOR (T2949)
T2949blotsmall Figure 2. Immunoblot - Anti-mTOR (T2949)
References:
  1. Schmelzle, T. and Hall, M.N. Cell, 103, 253-262 (2000).
  2. Brown, E.J., et al. Nature, 369, 756-758 (1994).
  3. Sabatini, D.M, et al. Cell, 78, 35-43 (1994).
  4. Chiu, M.I and Berlin, V. Proc. Natl. Acad. Sci. USA, 91, 12574-12578 (1994).
  5. Sabers, C.J., et al. J. Biol. Chem., 270, 815-822 (1995).
  6. Dennis, P.B., et al. Science, 294, 1102-1105 (2001).
  7. Nave, B.T., et al. Biochem. J., 344, 427-431 (1999).
  8. Peterson, R.T., et al. J. Biol. Chem., 275, 7416-7423 (2000).
back to Spotlights
back to Antibody Explorer
back to top