Sigma-Aldrich
Life Science > Cell Biology > Antibodies > Learning Center > Antibody Explorer > Spotlights > Anti-Survivin: Marker for blocking apoptosis
Antibody Explorer
Anti-Survivin: Marker for blocking apoptosis
Progression of the cell cycle and control of apoptosis are crucial and intimately linked processes that occur in all multicellular organisms during development, normal cellular differentiation and tissue homeostasis. Survivin (TIAP, BIRC5), a 16 kDa protein, is a member of the inhibitors of apoptosis (IAP)/BIRP gene family, which includes XIAP, c-IAP-1, c-IAP-2, ILP-2, NAIP, Livin and Apollon1-4. IAP proteins are structurally characterized by the presence of one to three copies of a baculovirus IAP repeat (BIR), consisting of an approximately 70 amino acid zinc finger fold, and a RING finger domain. Several members of the IAP family block apoptosis by directly interacting with initiator and effector caspases and preventing their proteolytic processing and enzymatic activity. Unlike other IAPs, survivin contains only one BIR copy and a carboxyl-terminal RING finger and is unique for its dimeric structure.
Survivin is required for cell viability maintenance in mitosis, potentially coupling apoptosis to control of cell division. It is selectively expressed during mitosis during G2/M phase and is localized to mitotic spindle microtubules and midbodies of dividing cells. Survivin is over-expressed in a variety of human tumors, including adenocarcinomas of lung, pancreas, breast, colon, stomach and prostate, as well as in squamous lung cell carcinoma, acute myelogenous leukemia, large cell non-Hodgkin lymphoma, neuroblastomas and melanomas1,5-9.
Anti-Survivin (Product No. S8191) is produced using a peptide corresponding to amino acids 122-142 at the carboxyl-terminus of human survivin. This sequence is highly conserved (>70%) in rat and mouse survivin. Anti-Survivin recognizes human, mouse and rat survivin. The antibody is suitable for immunoblotting.
S8191blotsmall   Figure 1. Immunoblot - Anti-Survivin (S8191)
Related Antibodies
Description Host Product No.
Anti-ILP2 Rabbit I4782
Anti-NAIP Rabbit N6648
Anti-XIAP Rabbit X2503
Related Proteins
Description Product No.
Livin β/ML-IAP, human recombinant L1040
XIAP, human recombinant X3378
References:
  1. Ambrosini, G., et al. Nature Med., 3, 917-921 (1997).
  2. Ambrosini, G., et al. J. Biol. Chem., 273, 11177-11182 (1998).
  3. Li, F., et al. Nature, 396, 580-584 (1998).
  4. Salvesen, G.S. and Duckett, C.S. Nature Rev. Mol. Cell Biol., 3, 401-410 (2002).
  5. Tanaka, K., et al. Clin. Cancer Res., 6, 127-134 (2000).
  6. Grossman, D., et al. J. Invest. Dermatol., 113, 1076-1081 (1999).
  7. Kawasaki, H., et al. Cancer Res., 58, 5071-5074 (1998).
  8. Monz, M., et al. J. Clin. Oncol., 17, 2100 (1999).
  9. Moriai, R., et al. Anticancer Res., 21, 595-600 (2001).
back to Spotlights
back to Antibody Explorer