Sigma-Aldrich
Life Science > Cell Biology > Antibodies > Learning Center > Antibody Explorer > Spotlights > Monoclonal Anti-Thymine Dimer
Antibody Explorer
Monoclonal Anti-Thymine Dimer
Monoclonal Anti-Thymine Dimer (mouse IgG1 isotype) was produced using a tetra nucleotide containing cyclobutane thymine dimer (GTTG) as immunogen. The antibody reacts specifically with the (5’-6’) cyclobutane type of homothymine or thymine-cytosine heterodimers. It detects thymine dimers in single-stranded DNA, and has a lower affinity for the dimer in short oligonucleotides (a tail of minimum 10-20 thymine residues is required for efficient labeling of oligonucleotide probes). The product enables a sensitive and non-radioactive method for labeling, detection, and quantification of DNA fragments using ELISA, competitive ELISA, immunocytochemistry (laser-scan microscopy) and Southern immunoblotting.
A working concentration for Monoclonal Anti-Thymine Dimer of 0.5-1 5 µg/ml is determined by Southern immunoblotting using UV-induced thymine dimers of λ phage DNA fragments.
Distributed under license.
Patents WO87/01134, EP 0233 177 B1
Product Table
Description Host Clone Product No.
Monoclonal Anti-Thymine Dimer Mouse H3 T1192
back to Spotlights
back to Antibody Explorer