Sigma-Aldrich
Life Science > Cell Biology > Antibodies > Learning Center > Antibody Explorer > Spotlights > Protein-G Immunoprecipitation Kit
Antibody Explorer
Protein-G Immunoprecipitation Kit
Immunoprecipitation (IP) is one of the most widely used immunochemical techniques. IP followed by SDS-PAGE and immunoblotting is routinely used to: measure the molecular weights of protein antigens, study protein/protein interactions, determine specific enzymatic activity, monitor protein post-translational modifications and determine the presence and quantity of proteins. IP also enables the detection of rare proteins that otherwise would be difficult to detect, since immunoprecipitation can concentrate them as much as 10,000 fold.
The kit has several advantages:
  • Allows for maximum recovery of immunoprecipitates
  • Uses mini-spin columns, instead of microcentrifuge tubes
  • Easy and convenient washing of the antigen-antibodybound beads
  • Minimal loss of antigen/antibody bound beads during washing
  • Minimal or no non-specific signals by increasing the stringency of the washing steps
Sufficient for performing 50 IP assays.
Click Image to View Figure in its Entirety Figure 1. Procedure Diagram
Components
  • 10X IP buffer, 40 ml
  • 5 M NaCl, 15 ml
  • 10% SDS, 1 ml
  • Protein-G-Agarose, suspension, 2.0 ml
  • Spin columns and caps, 50 units
  • 2 ml Microcentrifuge tubes, 50 units
Product Table
Description Product No.
Protein G Immunoprecipitation Kit IP50
back to Spotlights
back to Antibody Explorer