|
||||||
|
Cell Biology
Mix-n-Stain™ Antibody Labeling TechnologyDirect labeling of primary antibodies is useful for flow cytometry applications and when primary antibodies from different species are not available for multi-color immunofluorescence experiments. Typically, labeling antibodies with reactive dyes requires antibody buffer exchange, long incubation times, and time-consuming purification steps to remove unconjugated dye. Mix-n-Stain antibody labeling kits are a breakthrough technology for simple and rapid small scale antibody labeling from 5 to 100 µg.
Simple Antibody Labeling Protocol – 3 Easy Steps
Advantages for using Mix-n-Stain Labeling Kits
Complete list of Mix-n-Stain Kits |
|||||
|
|
||||||