Antibodies
Immuno Procedure
|
|
|
Deparaffinization
Paraffin sections of 4 µm thickness are baked overnight at 50 °C. Prior to immunostaining, deparaffinization and hydration is done in xylene and graded ethanol to distilled water. During hydration, block endogenous peroxidase using a solution of 0.03% H2O2 in 95% ethanol for 5 minutes.
Standard Antigen Retrieval Method
The standard antigen retrieval method is Heat Induced Epitope Retrieval (HIER) in retrieval buffer pH 6, using a pressure boiler (Decloaking chamber, Biocare Medical, Walnut Creek, CA, USA) as heat source.
HIER is performed by heating the TMA-slides immersed in retrieval buffer for 4 minutes at 125 °C in the pressure boiler. After completed boiling, slides remain in the pressure boiler and are allowed to cool to 90 °C. The total processing time is approximately 45 minutes.
The standard primary antibody dilution is based on the primary antibody stock concentration:
dilution 1:25 for primary antibody stock concentration of <0.06 mg/ml
dilution 1:75 for primary antibody stock concentration of >0.06 mg/ml
dilution 1:150 for primary antibody stock concentration of >0.1 mg/ml
NOTE: The specified working dilutions of the primary antibodies are to be considered as a guideline only. Optimal dilutions must be determined by the user.
Immunohistochemical staining program, Autostainer 480®
(Lab Vision, Freemont, CA, USA)
All incubations are done at room temperature.
- Rinse in wash buffer
- Incubation with primary antibody for 30 minutes
- Rinse in wash buffer (x2)
- Incubation with labeled polymer for 30 minutes
- Rinse in wash buffer (x2)
- Developing in DAB solution for 10 minutes
- Rinse in distilled water (x2)
- Counterstaining in hematoxylin for 5 minutes**
- Rinse in tap water for 5 minutes**
- Rinse in lithium carbonate water, diluted 1:5 from saturated solution for 1 minute**
- Rinse in tap water for 5 minutes**
- Dehydration in graded ethanol and xylene**
- Coversliping**
All reagents are applied at a volume of 300 µl per slide.
** Steps 8-13 are done in a histostaining instrument (Leica Autostainer XL).
Reagents
For immunohistochemistry, the following reagents are commercially available from Lab Vision, Freemont, CA, USA:
- Wash buffer (10x concentrate). Working solution originally contains 0.05% (v/v) Tween 20. Extra Tween 20 is added to a final concentration of 0.20%
- Retrieval Solution: Citrate Buffer®, pH 6
- Antibody diluent
- UltraVision LP HRP Polymer® and DAB Plus Substrate System®
In addition, Mayer's hematoxylin (Sigma-Aldrich®, St. Louis, MO, USA) is required.
Alternative Antigen Retrieval Method
For selected Prestige Antibodies, alternative retrieval buffers and/or enzymatic antigen retrieval may have been used as stated on the Certificate of Analysis and on the Antigen/Antibody information page on the Human Protein Atlas website.
Enzymatic Antigen retrieval
Enzymatic retrieval is performed in the immunostaining instrument and refers to incubation of TMA-slides in Proteinase K (Lab Vision, Freemont, CA, USA) for 10 minutes at room temperature.
Heat Induced Epitope Retrieval (HIER) in retrieval buffer pH 9
HIER in retrieval buffer pH 9 is performed as the standard HIER except that retrieval buffer with pH 9 (Lab Vision, Freemont, CA, USA) is used instead of retrieval buffer with pH 6.
back to top