|
Prestige Antibodies®
Prestige Antibody Immunofluorescence Procedure
| Immunofluorescence Procedure (80 Kb PDF) |
 |
|
A large number of the Prestige Antibodies have been used in subcellular localization studies by immunofluorescence (IF) staining of three cell lines; A-431, U-2 OS and U-251MG. Each cell line is stained with a Prestige Antibody, two organelle probes specific for the endoplasmatic reticulum and micro-tubules, as well as counterstained with the nuclear probe DAPI. The IF images can be viewed on the Human Protein Atlas web portal (www.proteinatlas.org).
Reagents: Primary antibodies:
- Prestige Antibodies at a working concentration of 1-4 μg/ml.
Note 1: The dilution of the primary antibody is to be considered as a guideline only. Optimal dilution must be determined by the user.
- Chicken anti-Calreticulin polyclonal antibody
- Mouse anti-alpha Tubulin monoclonal antibody
Secondary antibodies:
- anti-mouse IgG -Alexa Fluor® 555 produced in goat
- anti-chicken IgG -Alexa Fluor 647 produced in goat
- anti-rabbit IgG -Alexa Fluor 488 produced in goat
Protocol: Cell Seeding All washes are performed at room temperature (RT).
- A glass-bottomed, multiwell plate is coated with fibronectin (conc. 12.5 μg/ml) for 1 h at room temperature.
- Cells are seeded (10,000-15,000 cells per well) and incubated at 37°C in humidified air with 5.2 % CO2, for 4 hours.
Immunostaining
- Growth medium is removed and the cells are washed in 1x PBS (8.1 mM Na2HPO4, 1.5mM KH2PO4, 137 mM NaCl, and 2.7 mM KCl, pH 7.2)
- The cells are fixed for 15 minutes in ice cold 4 % paraformaldehyde pH 7.2-7.3 in growth medium supplemented with 10 % fetal bovine serum (FBS).
- The cells are permeabilized 3 times for 5 minutes each with 0.1 % TRITON® X-100 in PBS.
- The cells are washed with 1x PBS and incubated overnight at 4 °C with the primary antibodies in 1x PBS supplemented with 4 % FBS.
- The following day the cells are washed 4 times for 10 minutes each with 1x PBS and incubated for 1.5 hours at room temperature with the secondary antibodies in 1x PBS supplemented with 4% FBS.
Note 2: The secondary antibodies are fluorescently labeled and thus light sensitive. The sample should be kept in dim light in this as well as in the following steps.
- The cells are counterstained for 4 minutes with the nuclear stain DAPI (0.6 μM in 1x PBS).
- The cells are washed 4 times for 10 minutes with 1x PBS and then mounted in glycerol + 10 % 10x PBS.
Back to Prestige Antibodies | Back to Antibodies
|
Alexa Fluor is a registered trademark of Molecular Probes. TRITON is a registered trademark of Union Carbide Corp.
|