Sigma-Aldrich
Life Science > Cell Biology > Antibodies > Prestige Antibodies® > Prestige Antibodies Images
Antibodies
Prestige Antibodies® Images
Prestige_antibodies_logo_v-small_-w200
Click Images to enlarge and go to HPA detail page and Sigma product detail page!
  HPA000288
Immunohistochemistry
Anti-ACE2:
Cat. No. HPA000288: Immunoperoxidase staining of formalin-fixed, paraffin-embedded human kidney tissue showing cytoplasmic and/or membranous staining of cells in tubuli.
  HPA001083
Immunohistochemistry
Anti-ARHGAP4:
Cat. No. HPA001083: Immunoperoxidase staining of formalin-fixed, paraffin-embedded human appendix tissue showing positive cytoplasmic and/or membranous staining of cells in lymphoid tissue.
  HPA002027
Immunohistochemistry
Anti-MBL2:
Cat. No. HPA002027: Immunoperoxidase staining of formalin-fixed, paraffin-embedded human liver tissue showing cytoplasmic and/or membranous staining of hepatocytes.
  HPA000609
Immunohistochemistry
Anti-EMD:
Cat. No. HPA000609: Immunoperoxidase staining of formalin-fixed, paraffin-embedded human oral mucosa tissue showing nuclear membranous staining of surface epithelial cells.
  HPA003353
Immunohistochemistry
Anti-SLC16A2:
Cat. No. HPA003353: Immunoperoxidase staining of formalin-fixed, paraffin-embedded human adrenal gland tissue showing positive membranous and cytoplasmic staining of cortical cells.
  HPA003396
Immunohistochemistry
Anti-ABCD4:
Cat. No. HPA003396: Immunoperoxidase staining of formalin-fixed, paraffin-embedded human lung tissue showing cytoplasmic and/or membranous staining of macrophages.
Over 700 IHC images available for each antibody.
Click here to search for 11,000 Prestige Antibodies.
back to Prestige Antibodies
back to Antibodies