Sigma-Aldrich
Life Science > Cell Biology > Antibodies > Prestige Antibodies® > Technical Aids
Prestige Antibodies
Technical Aids
Prestige_antibodies_logo_v-small_-250
  • White Papers
    Prestige Antibodies as Organelle Markers Prestige Antibodies in IHC Applications Prestige Antibodies as Tools in Detection Studies and Biomarker Discovery BioTechniques Article (449 Kb PDF) Prestige Antibodies in Western Blot Applications (438 Kb PDF) Mapping the Human Proteome Using Prestige Antibodies (2.5 Mb PDF) Prestige Antibodies in Immunofluorescence Applications (493 Kb PDF) Explore Proteins with Unknown Function by the Use of Prestige Antibodies A New Cancer Biomarker Discovered (1.38 Mb PDF) Prestige Antibodies powered by Atlas Antibodies in Cell and Cell Line Studies (657 Kb PDF) The Unique Features of Prestige Antibodies (863 Kb PDF) Exploring Colon Cancer
  • Publications
  • Protocols
    Immunohistochemistry Western Blot Protein Array Immunofluorescence
  • Posters
  • Webinars
    Creation of a Human Protein Atlas Antibody-based Biomarker Discovery
    in Cancer Research
Human Protein Atlas (HPA) Project Workflow
The HPA is currently exploring the human proteome through antibody-based proteomics, which combines the high-throughput generation of monospecific antibodies with protein profiling in standard sets of tissue and cell microarrays. Each antibody produced begins with the selection of a protein epitope signature tag (PrEST). The PrEST fragment is typically 50-150 amino acids in length and is as dissimilar as possible to other proteins. The recombinant PrEST protein is utilized as the antigen in generating affinity purified monospecific antibodies.
The antibodies are then thoroughly tested through a series of quality assurance steps. Plasmid inserts are sequenced to ensure that the correct PrEST sequence is cloned. The size of the resulting recombinant protein is analyzed using mass spectrometry to ensure that the correct antigen has been produced and purified. To ensure antigen specificity, the antibodies are tested on protein arrays spotted with PrEST fragments. All 700 IHC staining images in normal and disease tissues, cell lines and cell types are annotated and curated by certified pathologists or by specially educated research scientists. In addition, the antibodies are tested by Western blot analysis and immunofluorescent microscopy. The antibodies are then assigned a validation score that assesses all validation tests as well as bioinformatic predictions and literature data. All of the images and data are available publicly on the Human Protein Atlas website www.proteinatlas.org.
Human Protein Atlas (HPA) Project Workflow
back to Prestige Antibodies
back to Antibodies