Cancer Research
Pituitary Gland
Harderian Gland
Thyroid Gland
Ear/Zymbal's Gland
Liver
Mammary Gland
Testes
Clitoral/Preputial Gland
Pancreas
Ovary
Adrenal Gland
Pituitary Gland
Sigma Logo Carcinogen Selector Header
The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs across 9 biological systems.
Step 1:     Select a biological system.     Step 2:     Use the mouse model to select an organ.     Step 3:     Review the organ specific carcinogen table to identify products.
Digestive System
Endocrine System
Hematopoietic System
Integumentary System
Reproductive System
Respiratory System
Skeletal System
Urinary System
Vascular System
Mouse
Endocrine System: Pituitary Gland
Product # Description Product # Description
D40752 1,2-Dibromoethane I504 2-Imidazolidinethione
M3302 2-Mercaptobenzothiazole A8056 3-Amino-1,2,4-triazole
P3755 6-Propyl-2-thiouracil D4628 Diethylstilbestrol
E7023 Ethanol 464953 Isoprene
M3761 Metronidazole Z2125 Zearalenone