Cancer Research
Skin
Skin
Subcutaneous Tissue
Sigma Logo Carcinogen Selector Header
The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs across 9 biological systems.
Step 1:     Select a biological system.     Step 2:     Use the mouse model to select an organ.     Step 3:     Review the organ specific carcinogen table to identify products.
Digestive System
Endocrine System
Hematopoietic System
Integumentary System
Reproductive System
Respiratory System
Skeletal System
Urinary System
Vascular System
Mouse
Integumentary System: Skin
Product # Description Product # Description
D4393 1,6-Dibromo-1,6-dideoxy-D-mannitol 123358 1-Chloro-2-methylpropene
301930 2,2-Bis(bromomethyl)-1,3-propanediol 101397 2,4-Dinitrotoluene
437158 2,6-Dimethyl-1,3-dioxan-4-ol acetate, mixture of cis and trans 161195 2-Methoxy-5-nitroaniline
438804 2-Nitrotoluene 440027 3,3'-Dimethoxy-4,4'-biphenylene diisocyanate
191248 3,3'-Dimethoxybenzidine dihydrochloride A2385 5-Azacytidine
210315 Acid Red 114 A1010 Arsenic(III) oxide
401765 Benzene D2535 C.I. direct blue 15
G5809 Glycidol A7015 N-(2-Fluorenyl)acetamide
T6269 o-Tolidine dihydrochloride O0380 Oxymetholone
P3750 Pararosaniline hydrochloride 79346 Phorbol 12-myristate 13-acetate
T6069 Thio-TEPA T8656 Thiourea
387622 Vinyl chloride