Atto Dyes Overview
  λabs ε max λem ηem τem Catalog Number
Atto Dye (and Quencher) [nm] [m-1  cm-1] [nm] [%] [ns] Free Acid NHS Ester Maleimide Azide Amine Iodoacetamide Biotin
Atto 390 390 24'000 479 90 3.8 89313 89204 89740 68321      
Atto 425 436 45'000 484 90 3.5 56749 16805 49349 78948     28616
Atto 430LS 433 32'000 547 65 4.0 06714 56387 41882        
Atto 465 453 75'000 508 55 2.2 50712 53404 55607        
Atto 488 501 90'000 523 80 3.2 41051 41698 28562 72709 74417 74402 30574
Atto 490LS 496 40'000 661 30 2.6 06715 78362 78363 93688      
Atto 495 495 80'000 527 45 2.4 16951 00379 41022        
Atto 514 511 115'000 533 85 3.0 52595 67455 81062 44311 93728 80027 00713
Atto 520 516 110'000 538 90 3.8 70706 77810 16590 61911     01632
Atto 532 532 115'000 553 90 3.8 06699 88793 68499        
Atto Rho6G 535 115'000 560 90 4.1 88821 93516 87785       94169
Atto 540Q 542 105'000       40592 61683 62453        
Atto 550 554 120'000 576 80 3.2 42424 92835 30730 41986 39271   28923
Atto 565 563 120'000 592 90 3.4 75784 72464 18507 43069     92637
Atto Rho3B 565 120'000 592 50 1.5 93685 28743 76531       07876
Atto Rho11 571 120'000 595 80 4.0 56611 89101 51431       19096
Atto Rho12 576 120'000 601 80 4.0 44432 68761 72763       91254
Atto Thio12 579 110'000 609 15 2.0 56963   94079       87784
Atto Rho101 586 120'000 610 80 4.2 77085 50492 73522       94379
Atto 580Q 586 110'000       03722 44756 68152        
Atto 590 594 120'000 624 80 3.7 70425 79636 39887       43208
Atto 594 601 120'000 627 85 3.5 08637 08741 08717 72998 61583   03927
Atto Rho13 600 120'000 625 80 3.9 16973 68538 44535       76141
Atto 610 615 150'000 634 70 3.3 78493 93259 41061       43292
Atto 612Q 615 115'000       04327 53988 50332        
Atto 620 619 120'000 643 50 2.9 92716 67351 49728       72978
Atto Rho14 625 140'000 646 80 3.7 59746 61909 14397       76140
Atto 633 629 130'000 657 64 3.2 18620 01464 n/a 68517 73245 68652  
Atto 647 645 120'000 669 20 2.3 97875 07376 41784        
Atto 647N 644 150'000 669 65 3.4 04507 18373 05316 91000 95349 73353 93606
Atto 655 663 125'000 684 30 1.9 93711 76245 80661 11774 14918 73948 06966
Atto Oxa12 663 125'000 684 30 1.8 92606 55785 01971       61938
Atto 665 663 160'000 684 60 2.9 16851 04022 01407       01376
Atto 680 680 125'000 700 30 1.8 94875 75999 04971       55819
Atto 700 700 120'000 719 25 1.5 30674 16986 50611 59825 19571   73911
Atto 725 729 120'000 752 10 0.5 47156 93725 90979       69616
Atto 740 740 120'000 764 10 0.6 91394 59808 77071       50842
Atto MB2 658 100'000       75118 73499 51797       02934
λabs - longest-wavelength absorption maximum εmax - molar extinction coefficient at the longest-wavelength absorption maximum
λem - fluorescence maximum ηem - fluorescence quantum yield τem - fluorescence decay time