Sigma-Aldrich
Life Science > Cell Biology > Hematology & Histology > Certified Stains & Dyes
Hematology & Histology
Certified Stains & Dyes
55 stains and dyes certified by the Biological Stain Commission, in addition to 300 other popular stains and dyes.
BSC Certified Stains & Dyes back to top
Product Name Product # 
Acid Fuchsin calcium salt certified by the BSC for VanGieson connective tissue stain and Mallory aniline blue connective stain A3908
Alcian Blue 8GX certified A3157
Alizarin Red S certified by the BSC for (for gross staining of skeletons in small vertebrates and for differentiating bone from cartilage in mammalian embryos) A5533
Aniline Blue diammonium salt certified 415049
Auramine O certified by the BSC for staining acid-fast bacteria in sputum or in paraffin sections of infected tissue. A9655
Azure A certified by the BSC for (for staining cell granules, microorganisms and other tissue. A constituent of Tetrachrome Stain (Macneal) for differentiating leucocytes.) A2918
Azure B certified by the BSC for differentiating cellular RNA and DNA in plant tissues. A4043
Basic Fuchsin certified 857343
Basic Fuchsin certified by the BSC for staining bacterial flagella. B0904
Bismarck Brown B 861111
Brilliant Cresyl Blue ALD certified by the BSC for staining the reticulum of immature red blood cells and platelets. B5388
Brilliant Green certified by the BSC for determining the presence of the colon organisms in drinking water and as a bacteriostatic agent. B4014
Carmine certified by the BSC for Orth's lithium carmine technique. C6152
Congo Red certified by the BSC for (for use with Mayer's hemalum as a cytoplasmic stain on tissues fixed in Bouin's fluid, and as a stain for amyloid in Highman's technique.) C6277
Cresyl Violet acetate certified by the BSC for the Powers and Clark method on formalin or Bouin's fixed spinal cord or brain. C5042
Crystal Violet certified by the BSC for the Gram stain; the Flemming triple stain with iodine; and Newton's crystal violet-iodine technique. C0775
Darrow Red certified 211885
Eosin B Dye content min. 85 % E8761
Eosin Y disodium salt Dye content min. 80 % E4382
Erythrosin B sodium salt certified by the BSC for (for use with methylene blue as a plasma stain for nerve cells; for staining bacteria in soil; and as a phosphorescent triple probe for studying diffusion of membrane proteins.) E8886
Erythrosin extra bluish certified 198269
Eosin alcohol soluble certified 199540
Fast Green FCF certified by the BSC for use as a cytological counterstain and in Lillie's modification of the Masson's trichrome stain. F7258
Giemsa stain certified by the BSC for (for use in staining leukocytes in blood films and for staining malarial parasites in thin and thick films of blood.) G5637
Hematoxylin certified H3136
Indigo Carmine certified by the BSC for (for use with acid fuchsin as a stain for Negri bodies and with eosin Y as a counterstain to hematoxylins for staining vaginal smears.) I2387
Jenner's stain certified by the BSC for the Romanowsky-type stain for peripheral blood smears. J1875
Light Green SF Yellowish Dye content min. 65 % L1886
Malachite Green oxalate salt certified by the BSC for (for use as a counterstain on botanical material; as a spore stain for bacteria in the Schaeffer and Fulton method; and in Conklin's modification of the method.) M9015
Methyl Blue certified M5528
Methylene Blue certified by the BSC for histological and bacteriologic staining and as a constituent of Wright's blood stain. M9140
Methyl Orange certified 234109
Methylene Violet (Bernthsen) certified 216313
Methyl Green M5015
Methyl violet 2B certified 198099
Neutral Red certified by the BSC for supravital staining of living blood cells. N6634
Nigrosin certified 198285
Nile Blue A certified by the BSC for stain for fats and for the differentiation of melanins and lipofuscins. N0766
Oil Red O certified 198196
Orange G certified by the BSC for Mallory's connective tissue stain and as a counterstain in histology and cytology. O7252
Orange II sodium salt certified by the BSC for (as a general tissue stain with eosin and azure C; with fast green FCF, safranin O and crystal violet in quadruple staining of tissues; as a stain for keratin in skin sections.) O4505
Orcein synthetic certified by the BSC for 8020 O7505
Pararosaniline hydrochloride Dye content min. 88 % P1528
Phloxine B certified by the BSC for Mallory's phloxine-methylene blue stain. P4030
Pyronin B certified 30 % (dye content) 201308
Pyronin Y certified by the BSC for Kurnick's methyl green-pyronin stain on liver cells. P6653
Resazurin sodium salt certified 199303
Rose Bengal sodium salt certified by the BSC for (as a fluorochrome for UV study of fats; for plate counts of soil fungi (bacteriostatic, but allows growth of fungi).) R4507
Safranin O certified by the BSC for (for use as a chromosome stain in the Flemming triple stain in combination with light green SF yellowish or fast green FCF; and in Gram stain.) S8884
Sudan III certified by the BSC for use as a fat stain. S4136
Sudan IV certified by the BSC for use as a fat stain. S4261
Sudan Black B certified 199664
Thionin acetate Dye content min. 85 % T7029
Toluidine Blue O T0394
Wright stain certified by the BSC for staining blood and bone marrow films, and for staining malarial parasites in blood films. W3000
Other Popular Stains & Dyes   back to top
Product Name Grade Product # 
1-(2-Pyridylazo)-2-naphthol indicator grade Indicator 101036
2-(5-Bromo-2-pyridylazo)-5-(diethylamino)phenol 97 % Technical 180017
2,3,5-Triphenyltetrazolium chloride Technical T8877
2,6-Dichloroindophenol sodium salt hydrate Technical D1878
2',7'-Dichlorofluorescein ACS reagent ACS 410217
Chromotropic acid disodium salt dihydrate approx. 98 % Technical D5144
4-Amino-3-hydroxy-1-naphthalenesulfonic acid ACS reagent 98+ % ³98.0 % ACS 398969
Acid Fuchsin Technical F8129
Acridine orange hydrochloride hydrate 99 % (HPLC) HPLC/TLC/
Electrophoresis
318337
Acridine orange solution 10 mg/ml in water Technical A8097
Acridine orange hemi(zinc chloride) salt 90 % (dye content) Technical 158550
Adrenochrome Technical A5752
Alcian Blue 8GX Technical 236551
Alcian Blue 8GX powder Technical A5268
Alcian Blue, pyridine variant Technical 335630
Alcian Blue-tetrakis(methylpyridinium) chloride Technical A4045
Alizarin Technical 122777
Alizarin yellow R Technical 195073
Allura Red AC Technical 458848
Aurintricarboxylic acid ammonium salt ACS reagent ACS A36883
Amaranth Technical A1016
Anthrone Technical A1631
Anthrone ACS reagent 97 % ACS 319899
Arsenazo III Technical A92775
Astra Blue Technical 363405
Auramine O 80 % (dye content) Technical 856533
Azobenzene 99 % Technical 424633
Azure B Technical 227935
Azure II A mixture of Azure B and Methylene Blue in equal amounts. Technical 861065
Basic Fuchsin Technical 215597
Benzophenone imine 97 % Technical 293733
Biotin-4-Fluorescein Technical B9431
Bisbenzimide H 33258 min. 98 % (TLC) Technical B2883
Bismarck Brown Y (G) Technical B2759
Brilliant Black BN Technical 211842
Ponceau BS Technical B6008
Brilliant Blue G Dye content approx. 60 % by elemental analysis Technical B1131
Brilliant Blue G min. 90 % (TLC) HPLC/TLC/
Electrophoresis
B5133
Brilliant Blue G solution Concentrate Technical B8522
Brilliant Blue G Dye content approx. 90 % by elemental analysis Technical B0770
Brilliant Blue R Dye content approx. 50 % Technical B0149
Brilliant Blue R Dye content approx. 90 % Technical B7920
Brilliant Blue R Concentrate Technical B8647
Brilliant Blue R Staining Solution Ethanol based Technical B6529
PhastGel Blue R Technical B4921
Brilliant Cresyl Blue ALD Technical 860867
Brilliant Green Technical B6756
Bromocresol Green Sultone Form Technical B0381
Bromocresol Green sodium salt crystalline Technical B1256
Bromocresol Green sodium salt ACS reagent ACS 114367
Bromocresol Purple Technical B5880
Bromobimane min. 97 % Technical B4380
Bromocresol Green sodium salt solution 0.04 wt. % in water Technical 318701
Bromocresol Green Sultone Form ACS reagent ACS 114359
Bromocresol Purple indicator grade Indicator 114375
Bromocresol Purple solution 0.04 wt. % in water Technical 318736
Bromocresol Purple sodium salt indicator grade Indicator 860891
Bromophenol Blue solution 0.04 wt. % in water Technical 318744
Bromphenol Blue sodium salt ACS reagent ACS 114405
Bromophenol Red sodium salt Technical B7010
Bromosulfalein Technical S0252
Bromphenol Blue sodium salt Technical B6131
Bromphenol Blue sodium salt for molecular biology, for electrophoresis HPLC/TLC/
Electrophoresis
B8026
Bromphenol Blue sodium salt for electrophoresis HPLC/TLC/
Electrophoresis
B5525
Bromophenol Blue Technical B0126
Bromothymol blue sodium salt Technical B8630
Bromothymol blue sodium salt ACS reagent ACS 114421
Calcein Technical C0875
Calcein AM solution min. 4 mM in DMSO min. 90 % (TLC) liquid Technical C1359
Calcon® Technical H1878
Calmagite indicator grade Indicator C204
Canada balsam Technical C1795
Carbazole approx. 95 % Technical C5132
Carbol Fuchsin Technical C4165
Carbostyril 124 99 % Technical 363308
Cardiogreen Technical I2633
Carmine powder Technical C1022
Carminic acid Technical 229253
Celestine blue Technical 206342
Chicago Sky Blue 6B powder Technical C8679
Chlorophenol Red indicator grade Indicator 199524
Chlorophenol Red sodium salt Technical 235482
Chlorophyll a from Anacystis nidulans algae Technical C6144
Chlorophyll b from spinach Technical C5878
Chromeazurol B Technical M1782
Chromeazurol S Technical 199532
Chromotrope 2R suitable for modified Gomori Trichrome stain Technical C3143
Chromotrope 2R Technical 202088
Chromotropic acid disodium salt dihydrate ACS reagent ACS 213276
Chrysin Technical C80105
Cibacron Blue 3G-A Technical C9534
Congo Red Technical C6767
Coumarin 6 98 % Technical 442631
Coumarin 314 Technical 392995
Coumarin 343 Technical 393029
Practice of Thin Layer Chromatography, 3rd ed. Technical P5691
m-Cresol Purple sodium salt Technical 211761
Cresol red indicator grade Indicator 114472
Cresol Red sodium salt indicator grade Indicator 114480
Cresyl Violet acetate Technical C1791
Crocein Scarlet 7B Technical C8822
Crystal Violet Dye content approx. 90 % Technical C3886
Crystal Violet ACS reagent ³90.0 % as anhydrous ACS C6158
Curcumin from Curcuma longa (Turmeric) 65-70 % (typically) powder Technical C1386
Phloxine B crystalline Technical P2759
Demecolcine min. 98 % (TLC) Technical D7385
Diamidino yellow dihydrochloride Technical D0281
o-Dianisidine dihydrochloride tech. Technical 191248
o-Dianisidine dihydrochloride crystalline Technical D3252
o-Dianisidine reagent Technical D2679
Dihydroethidium Technical D7008
Dimidium bromide 95 % Technical 234281
Diphenylamine Technical D2385
Dithizone Technical D5130
Dithizone ³85.0 % ACS reagent ACS 194832
Direct Blue 71 Technical 212407
Direct Red 80 Technical 365548
Dithizone Technical D5130
Dithizone ³85.0 % ACS reagent ACS 194832
Eosin Y Dye content approx. 99 % Technical E4009
Eosin Y disodium salt Dye content approx. 85 % Technical E6003
Eosin Y solution 5 wt. % in water Technical 318906
Eriochrome Black T ACS reagent, indicator grade ACS 858390
Eriochrome Cyanine R practical grade Technical E2502
Erythrosin B 95+ % Technical 200964
Ethidium bromide approx. 95 % (HPLC) Technical E8751
Ethidium homodimer solution Technical E1903
Ethyl Orange sodium salt indicator grade Indicator 199567
Evans Blue Technical E2129
Fast Black K Salt hemi(zinc chloride) salt practical grade Technical F7253
Fast Blue BB Salt hemi(zinc chloride) salt Technical F3378
Fast Blue B Salt Technical 340774
Fast Corinth V zinc chloride double salt Technical 227366
Fast Garnet GBC sulfate salt Technical F8761
Fast Green FCF Dye content approx. 90 % Technical F7252
Fast Red Violet LB Salt Technical F3381
Fast Violet B Salt crystalline Technical F1631
Ferroin solution 0.1 wt. % in water Technical 318922
Fluo 3-AM approx. 70 % Technical F6142
Fluorescamine 98 % Technical 201650
Fluorescein sodium salt Technical F6377
Fluoresceinamine isomer I Technical 201626
Fluorescein diacetate Technical F7378
Fluorescein 5(6)-isothiocyanate approx. 90 % (HPLC) Technical F3651
Fluorescein isothiocyanate isomer I suitable for protein labeling min. 90 % (HPLC) powder Technical F7250
Fluorescein isothiocyanate isomer I approx. 98 % (HPLC) Technical F4274
Fluorescein isothiocyanate isomer I 90 % Technical F2502
Fluorescent Brightener 28 Technical F3543
Fura 2-AM powder Technical F0888
Fura PE 3-AM approx. 90 % Technical F0918
Fusidic acid Technical F0756
Gentian Violet USP USP G2039
Hematoxylin Technical H9627
Hydroxynaphthol Blue disodium salt ACS reagent ACS 219916
Indigo Carmine Technical I8130
Iodonitrotetrazolium chloride Technical I8377
Janus Green B Technical J1000
Jenner's stain suitable for blood stain Technical J1625
Keratin azure Technical K8500
Leishman's stain Technical L6254
Light Green SF Yellowish crystalline Technical L5382
Lucifer Yellow CH dilithium salt Technical L0259
Lucifer Yellow CH dipotassium salt 90 % (dye content) Technical 328197
Lumichrome Technical 103217
Malachite Green oxalate salt Technical M6880
Malachite Green Carbinol base Technical 229105
Melanin from Sepia officinalis Technical M2649
Metanil Yellow Technical 202029
Methyl 3-amino-5,6-dichloro-2-pyrazinecarboxylate 97 % Technical 277878
Methyl Blue Technical M6900
Methyl Green approx. 85 % Technical M8884
Methyl Orange ACS reagent ACS 114510
Methyl Purple in solution in water Technical 277339
Methyl Red Technical M7267
Methyl Red hydrochloride ACS reagent ACS 861235
Methyl Red sodium salt Technical M8256
Methyl Red crystals ACS reagent ACS 250198
Methyl Red sodium salt ACS reagent ACS 114502
Methylene Blue Technical M44907
Methylene Blue solution Technical 1808
Methylene Blue Technical MB1
Methylene Blue Cell/Molecular M4159
Methylene Violet 3RAX Technical 307505
Methylthymol Blue sodium salt Technical B42004
MTT Formazan powder Technical M2003
Murexide ACS reagent ACS 222461
Naphthol Blue Black Technical 195243
Naphthol Green B Technical N7257
Naphthol Yellow S Technical 199621
a-Naphtholphthalein practical grade Technical N8257
N,N-Diethyl-p-phenylenediamine hemioxalate salt Technical D5143
Neutral Red powder cell culture tested Cell/Molecular N4638
Neutral Red practical grade Technical N2880
Neutral Red Technical N7005
New Fuchsin Technical N0638
Nigrosin water soluble Technical N4754
Nile Red Technical N3013
Ninhydrin 97 % Technical 454044
Nitro Blue Tetrazolium tablet Cell/Molecular N5514
Nitrotetrazolium blue chloride approx. 98 % (TLC) HPLC/TLC/
Electrophoresis
N6876
Nitrotetrazolium blue chloride powder Cell/Molecular N6639
Nitrotetrazolium Blue chloride monohydrate Technical N15405
Nuclear Fast Red suitable for determination of calcium Technical N8002
Oil Red O powder Technical O0625
Orange G Technical O1625
Orange G Cell/Molecular O3756
Orcein synthetic Technical O7380
Orcinol monohydrate from lichens Technical O1875
Osmium tetroxide Technical O5500
Oxacillin sodium salt hydrate Technical O1002
Pararosaniline Base crystalline Technical P7632
Pararosaniline hydrochloride powder Technical P3750
Perylene Purified by sublimation 99.5+ % Technical 394475
Perylene 99+ % Technical P11204
PhastGel Blue R Technical B4921
Phenazine ethosulfate approx. 95 % Technical P4544
Phenol red solution liquid 0.5% cell culture tested Cell/Molecular P0290
Phenol red solution for electrophoresis HPLC/TLC/
Electrophoresis
P8421
Phenol Red Technical P4633
Phenol Red powder cell culture tested Cell/Molecular P3532
Phenol Red sodium salt Technical P4758
Phenol Red sodium salt powder cell culture tested insect cell culture tested Cell/Molecular P5530
Phenol Red ACS reagent ACS 114529
Phenol Red sodium salt solution 0.04 wt. % in water Technical 319244
Phenol Red sodium salt ACS reagent ACS 114537
Phenolphthalein Technical P9750
Phenolphthalein USP USP P9471
Phenolphthalein solution 0.5 wt. % in 50 wt. % ethyl alcohol Technical 319236
Phenolphthalein ACS reagent ACS 105945
Phenosafranin Technical 199648
Phenylhydrazine hydrochloride Technical P6926
Phloxine B crystalline Technical P2759
Ponceau BS Technical B6008
Ponceau S Technical 141194
Ponceau S practical grade Technical P3504
Ponceau S solution Technical P7170
Ponceau Xylidine Technical P2395
Potassium phthalate monobasic min. 99 % Technical P6758
Propidium iodide min. 95 % (HPLC) Technical P4170
Propidium iodide solution solution (1.0 mg/ml in water) Technical P4864
Pyrogallol Red Technical P8759
Pyronin B Technical P6892
Pyronin Y Cell/Molecular P9172
Quinaldine Red Technical 201316
Reactive blue 4 Technical 244813
Reichardt's dye Technical 272442
Reinecke salt Technical R4126
Remazol Brilliant Blue R Technical R8001
Resorufin sodium salt Technical 230154
Rhodamine B Technical R6626
Rhodamine B 90 % (dye content) Specialty-material science 252425
Rhodamine 6G 99 % (dye content) Technical 252433
Rhodamine 6G Technical R4127
Rhodamine 6G perchlorate 99 % (dye content) Technical 252441
Rhodamine 110 chloride 75 % (dye content) Technical 432202
Rhodamine 123 Technical R8004
Rhodamine B base Technical 234141
Rhodamine B isothiocyanate Technical 283924
Rose Bengal sodium salt 95 % (dye content) Technical 330000
Rose Bengal sodium salt Technical R3877
p-Rosolic acid Technical 861324
Ruthenium Red Technical R2751
Saffron Technical S8381
Safranin O Dye content approx. 85 % Technical S2255
Schiff's Fuchsin-sulfite reagent Technical S5133
Scopoletin Technical S2500
Silver nitrate >99 % (titration) Technical S6506
Silver nitrate SigmaUltra >99 % (titration) Ultra S8157
Silver nitrate >99 % (titration) plant cell culture tested Cell/Molecular S7276
Silver nitrate USP USP S1179
Solvent Blue 38 practical grade Technical S3382
Solvent Green 3 Technical 211982
Sudan Black B practical grade Technical S2380
Sudan III Technical S4131
SYBR® Green II RNA gel stain 10,000 × in DMSO Cell/Molecular S9305
SYBR® Green I nucleic acid gel stain 10,000 × in DMSO Cell/Molecular S9430
Tannic acid powder Technical T0125
Tetrazolium Blue Chloride Technical T4375
Thiazolyl Blue Tetrazolium Bromide approx. 98 % (TLC) Technical M2128
Thioflavin T Technical T3516
Thionin acetate, dye content approx. 90 % Technical T3387
Thymol Blue ACS reagent ACS 114545
Thymol Blue sodium salt ACS reagent ACS 861367
Thymol min. 99.5 % Technical T0501
Thymolphthalein ACS reagent ACS 114553
o-Tolidine dihydrochloride min. 97 % Technical T6269
Toluidine Blue O Technical T3260
Toluidine Red Technical 199753
True Blue aceturate salt Technical T5891
Trypan Blue Technical 302643
Trypan Blue powder cell culture tested Cell/Molecular T6146
Trypan Blue solution liquid 0.4% cell culture tested Cell/Molecular T8154
Uniblue A sodium salt Technical 298409
Victoria Blue B Technical V0753
Wright stain suitable for blood stain technical W0625
Xylene Cyanol FF Technical 335940
Xylene Cyanole FF for molecular biology Cell/Molecular X4126
Xylenol Orange tetrasodium salt ACS reagent ACS 398187
Xylenol Orange tetrasodium salt 90 % (dye content) Technical 227854
Xylidyl blue I Technical 248266
Zincon sodium salt Technical 201332