Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Hematology & Histology
Products by Topic
Adhesives
Product Name Product # 
Poly-Prep Slides. Glass slides coated with poly-l-lysine. Not recommended for microarray use. P0425
Posi-Prep™ Slides, Tested RNase and DNase free P8611
Poly-l-lysine solution (0.1% w/v with 0.1% thimerosal added as a preservative. Intended to attach tissue sections to glass slides. Not recommended for microarray use. P8920
Silane-Prep Slides Glass slides coated with silane (aminoalkylsilane). Not recommended for microarray use. S4651
back to top
Consumables
Product Name Product # 
Accumateä Tissue Embedding Medium (Paraplast Regular) A6330
Cell Maturation Lithograph (Human blood cells stained with Wright Stain) C6919
Hanks' balanced salt solution, modified H4385
Humid Chamber H6644
Coplin staining jar with screw cap high-density polyethylene S5641
back to top
Mounting Media
Product Name Product # 
Situ Mount TM Mounting Media. Specifically optimized for use in in-situ hybridization applications following the use of BC-50 Red Counterstain S0566
Crystal Mount TM Aqueous Mounting Media. For use with AEC, 4-chloro-1-Napthol, BCIP/INT, and Fast Red TR/Napthol AS-MX C0612
Gel Mount TM Aqueous Mounting Media. Suitable for use with conjugated tracers including fluorescein, rhodamine, Texas Red, phycoerythrin and phycocyanin G0918
Clarion TM Mounting Medium. Environmentally safe mounting medium for general use in Histology and Cytology C0487
back to top
Fixatives
Product Name Product # 
Formalin solution (approximately 4% formaldehyde and 1 to 1.5% methanol) HT50-1-1
Formalin solution Neutral buffered solution, 10% Formalin (approximately 4% formaldehyde and 1 to 1.5% methanol)general purpose histological fixative (4 Liters) HT50-1-128
Formalin solution (approximately 4% formaldehyde and 1 to 1.5% methanol), general purpose histological fixative 1 case (24X60 mL) HT50-1-2
Formalin solution (approximately 4% formaldehyde and 1 to 1.5% methanol), general purpose histological fixative (9.5Liters) HT50-1-320
Formalin solution (approximately 4% formaldehyde and 1 to 1.5% methanol), general purpose histological fixative 1 case (24X120 mL) HT50-1-4
Formalin solution Neutral buffered solution, 10% Formalin (approximately 4% formaldehyde and 1 to 1.5% methanol), general purpose histological fixative (19 Liters) HT50-1-640
back to top
Controls
Product Name Product # 
Acid Fast TISSUE-TROLä A2299
Amyloid TISSUE-TROLä A2424
Elastic TISSUE-TROLä E6143
Fungi TISSUE-TROLä F0766
Helicobacter TISSUE-TROLä H1898
Iron TISSUE-TROLä I3387
Mucin TISSUE-TROLä M9543
Pneumocystis TISSUE-TROLä P2418
PAS TISSUE-TROLä P8814
Reticulum TISSUE-TROLä R4768
Spirochetes TISSUE-TROLä S2645
Trichrome TISSUE-TROLä T8800
back to top
Instruments
Product Name Product # 
Accumateä H2100 Microwave Oven 110 Volt A9084
Accumateä H2100 Microwave Oven 220 Volt A9209
back to top
Certified Stains & Dyes
Product Name Product # 
Azure A (for staining cell granules, microorganisms and other tissue. A constituent of Tetrachrome Stain (MacNeal) for differentiating leucocytes A2918
Alcian Blue 8GX (used in Scott's pH 5.7 method, pH 2.5 method and Alcian Blue-PAS stain) A3157
Acid Fuchsin calcium salt VanGieson connective tissue stain and Mallory aniline blue connective tissue stain A3908
Azure B differentiating cellular RNA and DNA in plant tissues. Component of Wright Stain A4043
Alizarin Red S (used to demonstrate calcium deposits) A5533
Auramine O staining acid-fast bacteria in sputum or in paraffin sections of infected tissue. A9655
Basic Fuchsin staining bacterial flagella.(see product P1528 for standard Basic Fuchsin) B0904
Brilliant Green determining the presence of the colon organisms in drinking water and as a bacteriostatic agent. B4014
Brilliant Cresyl Blue ALD (used for demonstrating and counting reticulocyes and platelets). B5388
Crystal Violet used in Gram stain; the Flemming triple stain with iodine; and Newton's crystal violet-iodine technique. C0775
Cresyl Violet acetate (used in the Powers and Clark method for spinal cord or brain. Recommended substitute for Cresyl Echt violet or Cresyl Fast Violet) C5042
Carmine used in Orth's lithium Carmine technique. C6152
Congo Red (for use with Mayer's hemalum as a cytoplasmic stain on tissues fixed in Bouin's fluid, and as a stain for amyloid in Highman's technique). C6277
Eosin Y disodium salt (Cytoplasmic Counterstain) E4382
Eosin B (Cytoplasmic Counterstain) E8761
Erythrosin B sodium salt (for use as a contrast stain with hematoxylin and certain blue and violet nuclear stains) E8886
Fast Green FCF (used as a cytological counterstain and in Lillie's modification of the Masson's trichrome stain) F7258
Giemsa stain, modified (for use in staining leukocytes in blood films and for staining malarial parasites in thin and thick films of blood.) G5637
Hematoxylin (for use as a nuclear stain in various techniques) H3136
Indigo Carmine (for use with acid fuchsin as a stain for Negri bodies and with eosin Y as a counterstain to hematoxylins for staining vaginal smears) I2387
Jenner's stain used as a Romanowsky-type stain for peripheral blood smears. J1875
Light Green SF Yellowish (for use as a cytological counterstain) L1886
Methyl Green (used in methyl Green Pyronin stain for DNA and RNA) M5015
Methyl Blue (for use as Aniline blue in Mallory's connective tissue stain) M5528
Malachite Green oxalate salt (for use as a counterstain on botanical material; as a spore stain for bacteria in the Schaeffer and Fulton method; and in Conklin's modification of the method.) M9015
Methylene Blue hydrate (used as a histological and bacteriologic staining and as a constituent of Wright's blood stain.) M9140
Nile Blue A (stain for fats and for the differentiation of lipofuscins.) N0766
Neutral Red (supravital staining of living blood cells and as a nuclear stain.) N6634
Orange II sodium salt (used to demonstrate chromosomes and can be substituted for Orange G in certain histological procedures.) O4505
Orange G (used in Mallory's connective tissue stain and as a counterstain in histology and cytology.) O7252
Orcein synthetic, as a stain for elastic tissue and for hepatitis-B surface antigen. O7505
Pararosaniline hydrochloride (certified as Basic Fuchsin by the Biological Stain Commission.) P1528
Phloxine B. (used as a counterstain, with or without Eosin Y.) P4030
Pyronin Y (used in Kurnick's methyl green-pyronin stain on liver cells.) P6653
Rose Bengal sodium salt (as a fluorochrome for UV study of fats; for plate counts of soil fungi (bacteriostatic, but allows growth of fungi).) R4507
Sudan Black B (fat stain for animal tissue, bacteria and other microorganisms; as a stain for chromosomes, Golgi apparatus and leucocyte granules.) S0395
Sudan III (used as a fat stain.) S4136
Sudan IV (used as a fat stain.) S4261
Safranin O (for use as a chromosome stain in the Flemming triple stain in combination with light green SF yellowish or fast green FCF; and in various Gram stain procedures.) S8884
Toluidine Blue O (used for staining frozen sections and as a stain for connective tissue mucins, mast cells, and nissl granules.) T0394
Thionin acetate (used as a general nuclear stain for chromatin and mucin, for frozen section analysis and for staining Nissl granules.) T7029
back to top
Cytochemistry
Product Name Product # 
alpha-Naphthyl Butyrate solution 180-1
Naphthol AS-BI Phosphoric acid solution 180-2
Pararosaniline solution 180-4
Phosphate buffer 180-5
Methylene Blue solution 180-8
Acid Phosphatase, Lymphocyte 181-A
alpha-Naphthyl Butyrate Esterase, Lymphocyte 181-B
beta-Glucuronidase, Lymphocyte 181-C
Glutaraldehyde solution 380-2
Sudan Black B Staining System 380-B
Citrate Concentrate 386-1
Acetate solution 386-3
Naphthol AS-BI Phosphoric Acid 386-4
Acid Phosphatase, Leukocyte 386-A
Naphthol AS-BI Phosphoric acid solution 387-1
Fast Garnet GBC Base solution 387-2
Tartrate solution 387-3
Acid Phosphatase, Leukocyte 387-A
Peroxidase Indicator Reagent 390-1
Peroxidase (Myeloperoxidase), Leukocyte 390-A
Fast Violet B Salt 85-1
Sodium citrate solution 85-4
Sodium citrate solution 85-4C
Naphthol AS-MX phosphate 85-5
Alkaline Phosphatase (AP), Leukocyte 85L-1
Alkaline Phosphatase (AP), Leukocyte 85L-2
Alkaline Phosphatase (AP), Leukocyte 85L-3R
Naphthol AS-BI Alkaline solution 86-1
FRV-Alkaline solution 86-2
FBB-Alkaline solution 86-3
Alkaline Phosphatase (AP), Leukocyte 86-C
Alkaline Phosphatase (AP), Leukocyte 86-R
Trizmalä 6.3 Buffer Concentrate 90-3C
NAPHTHOL AS-D CHLOROACETATE 905
alpha-Naphthyl acetate 90-6
alpha-Naphthyl Acetate (Non-Specific Esterase) 90-A1
Naphthol AS-D Chloroacetate (Specific Esterase) 90-C2
Naphthol AS-D Chloroacetate solution 91-1
Fast Red Violet LB Base solution 91-2
Trizmalä 6.3 Concentrate 1 mol/l 91-3
Sodium nitrite solution 91-4
Citrate solution 91-5
alpha-Naphthyl acetate solution 91-6
Fast Blue BB Base solution 91-7
Sodium fluoride solution 91-9
alpha-Naphthyl Acetate (Non-Specific Esterase) 91-A
Naphthol AS-D Chloroacetate (Specific Esterase) 91-C
Fast Blue RR Salt Preweighted Capsules FBS-25
Methanol M1775
Neutral Red Solution. Counterstain used as a red nuclear counterstain for fixed blood smears. Neutral red 0.5% in pH 5.2 buffer, 50 ml bottle N6264
back to top
Hematology Stains
Product Name Product # 
Citrate Phosphate Buffer Concentrate 285-1
Acid hematoxylin solution pH 3.3 (25 °C) 285-2
Eosin B Cytoplasmic Counterstain solution 285-3
Ethylenediaminetetraacetic acid disodium salt (EDTA) solution 2%, Solution 285-4
Ethanol Fixative 80% v/v 285-8
Fetal Hemoglobin Kit 285-C
Fetal Hemoglobin Kit 285-D
NBT Vials 840-10
Stimulant 840-15
Nitroblue Tetrazolium (NBT) Reduction 840-W
Giemsa Stain, Modified Solution 0.4 % (w/v) in buffered methanol solution, pH 6.9, with stabilizers; 4 Liters GS-128
Giemsa Stain, Modified Solution 0.4 % (w/v) in buffered methanol solution, pH 6.9, with stabilizers; 1 Liter GS-1L
Giemsa Stain, Modified Solution 0.4 % (w/v) in buffered methanol solution, pH 6.9, with stabilizers; 500 mls GS-500
Giemsa Stain, Modified Solution 0.4 % (w/v) in buffered methanol solution, pH 6.9, with stabilizers; 2.5 Liters GS-80
May-Grunwald Stain, 0.25% in methanol; 4 Liters MG-128
May-Grunwald Stain, 0.25% in methanol; 1 Liter MG-1L
May-Grunwald Stain, 0.25% in methanol; 500 mls MG-500
May-Grunwald Stain, 0.25% in methanol; 2.5 Liters MG-80
Reticulocyte Stain R4132
Rinse solution no. 2 RS-2
Sigmacote® SL-2
Wright-Giemsa Stain, Modified WG-128
Wright-Giemsa Stain, Modified Solution 0.4%, pH 6.8 in methanol; 4 Liters WG-16
Wright-Giemsa Stain, Modified Solution 0.4%, pH 6.8 in methanol; 500 mls WG-32
Wright-Giemsa Stain, Modified Solution 0.4%, pH 6.8 in methanol; 1 Liter WG-80
Wright-Giemsa Stain, Modified Solution 0.4%, pH 6.8 in methanol; 2.5 Liters Hema-Tek® 1, 1000 and 2000 WG-HT
Wright Stain, Modified Solution 0.3% buffered at pH 6.9 in methanol; 4 Liters WS-128
Wright Stain, Modified Solution 0.3% buffered at pH 6.9 in methanol; 500 mls WS-16
Wright Stain, Modified Solution 0.3% buffered at pH 6.9 in methanol; 1 Liter WS-32
Wright Stain, Modified Solution 0.3% buffered at pH 6.9 in methanol; 2.5 Liters WS-80
Wright Stain for Geometric Data WSGD-128
Wright Stain Set for Hema-Tek® I, 1000 and 2000 WS-HT
Wright Stain sets for Hema-Tek I WSHT-S
Wright stain staining blood and bone marrow films, and for staining malarial parasites in blood films. W3000
back to top
Histology Routine Stains
Product Name Product # 
Gill Modified EA solution 391-5-128
Accumateä Differentiating solution 0.25% hydrochloric acid in 60% alcohol (ethanol) and 39.75% water; 1 Liter A3179
Accumateä Differentiating solution 0.25% hydrochloric acid in 60% alcohol (ethanol) and 39.75% water; 4 Liters A3429
Xylene Substitute A5597
GLYCEROL GELATIN GG1
Hematoxylin Solution, Gill No. 1, 2 g/L certified hematoxylin; 4 Liters GHS-1-128
Hematoxylin Solution, Gill No. 1, 2 g/L certified hematoxylin; 500 mls GHS-1-16
Hematoxylin Solution, Gill No. 1, 2 g/L certified hematoxylin; 1 Liter GHS-1-32
Hematoxylin Solution, Gill No. 2, 4 g/L certified hematoxylin; 4 Liters GHS-2-128
Hematoxylin Solution, Gill No. 2, 4 g/L certified hematoxylin; 500 mls GHS-2-16
Hematoxylin Solution, Gill No. 2, 4 g/L certified hematoxylin; 1 Liter GHS-2-32
Hematoxylin Solution, Gill No. 2, 4 g/L certified hematoxylin; 2.5 Liters GHS-2-80
Hematoxylin Solution, Gill No. 3, 6 g/L certified hematoxylin; 4 Liters GHS-3-128
Hematoxylin Solution, Gill No. 3, 6 g/L certified hematoxylin; 500 mls GHS-3-16
Hematoxylin Solution, Gill No. 3, 6 g/L certified hematoxylin; 1 Liters GHS-3-32
Hematoxylin Solution, Gill No. 3, 6 g/L certified hematoxylin; 2.5 Liters GHS-3-80
Hematoxylin Solution, Harris Modified 7 g/L Hematoxylin mercury free; 4 Liters HHS-128
Hematoxylin Solution, Harris Modified 7 g/L Hematoxylin mercury free; 500 mls HHS-16
Hematoxylin Solution, Harris Modified 7 g/L Hematoxylin mercury free; 1 Liter HHS-32
Hematoxylin Solution, Harris Modified 7 g/L Hematoxylin mercury free; 2.5 Liters HHS-80
Bouin's solution; 4 Liters HT10-1-128
Bouin's solution; 1 Liters HT10-1-32
Weigert”s Iron Hematoxylin Solution Part A HT10-7
Weigert”s Iron Hematoxylin Solution Set HT10-79
Weigert”s Iron Hematoxylin Solution Part B HT10-9
Eosin Y Cytoplasmic Counterstain Solution, Alcoholic 0.5% in acidified ethanol; 4 Liters HT110-1-128
Eosin Y Cytoplasmic Counterstain Solution, Alcoholic 0.5% in acidified ethanol; 500 mls HT110-1-16
Eosin Y Cytoplasmic Counterstain Solution, Alcoholic 0.5% in acidified ethanol; 1 Liter HT110-1-32
Eosin Y Cytoplasmic Counterstain Solution, Alcoholic 0.5% in acidified ethanol; 2.5 Liters HT110-1-80
Eosin Y Cytoplasmic Counterstain Solution, Aqueous, 0.5% 4 Liters HT110-2-128
Eosin Y Cytoplasmic Counterstain Solution, Aqueous, 0.5% 500 mls HT110-2-16
Eosin Y Cytoplasmic Counterstain Solution, Aqueous, 0.5% 1 Liter HT110-2-32
Eosin Y Solution, Aqueous, 0.5% 2.5 Liters HT110-2-80
Eosin Y Solution, Alcoholic with Phloxine, 0.5% Eosin Y and 0.1% Phloxine B; 4 Liters HT110-3-128
Eosin Y Solution, Alcoholic with Phloxine, 0.5% Eosin Y and 0.1% Phloxine B; 500 mls HT110-3-16
Eosin Y Solution, Alcoholic with Phloxine, 0.5% Eosin Y and 0.1% Phloxine B; 1 Liter HT110-3-32
Eosin Y Solution, Alcoholic with Phloxine, 0.5% Eosin Y and 0.1% Phloxine B; 2.5 Liters HT110-3-80
Papanicolaou Stain, OG-6; 4 Liters HT40-1-128
Papanicolaou Stain, OG-6; 500 mls HT40-1-16
Papanicolaou Stain, OG-6; 1 Liter HT40-1-32
Papanicolaou Stain, OG-6; 2.5 Liters HT40-1-80
Papanicolaou Stain, Modified EA; 4 Liters HT40-2-128
Papanicolaou Stain, Modified EA; 500 mls HT40-2-16
Papanicolaou Stain, Modified EA; 1 Liter HT40-2-32
Papanicolaou Stain, EA 50; 4 Liters HT40-3-128
Papanicolaou Stain, EA 50; 500 mls HT40-3-16
Papanicolaou Stain, EA 50; 1 Liters HT40-3-32
Papanicolaou Stain, EA 50; 2.5 Liters HT40-3-80
Papanicolaou Stain, EA 65; 4 Liters HT40-4-128
Papanicolaou Stain, EA 65; 1 Liters HT40-4-32
Hematoxylin Solution, Mayer's 1 g/L certified hematoxylin 100 mls MHS-1
Hematoxylin Solution, Mayer's 1 g/L certified hematoxylin 4 Liters MHS-128
Hematoxylin Solution, Mayer's 1 g/L certified hematoxylin 500 mls MHS-16
Hematoxylin Solution, Mayer's 1 g/L certified hematoxylin 1 Liter MHS-32
Hematoxylin Solution, Mayer”s MHS-80
Phosphate buffer pH 7.2 at 25° C P3288
Phosphate Buffer pH 6.6 at 25°C P8165
Reagent alcohol (95% ethanol, 5% methanol, 5% isopropanol) R8382
Scott¢s Tap Water Substitute Concentrate 10 Liter S5134
back to top
Histology Special Stains
Product Name Product # 
Peroxidase (Myeloperoxidase), Leukocyte 391-A
Periodic acid solution, 1%, 100 mL 395-1
Periodic acid solution, 1%, 1 Liter 395-1-32
Schiff's reagent, 395-2-016
Periodic Acid-Schiff (PAS) 395-B
Borax Solution 5 g/dL HT100-2
Silver methenamine HT100-3
Gold chloride solution HT100-4
Sodium thiosulfate anhydrous 2 g/dL HT100-5
Silver Stain (Modified GMS) HT100-A
Silver stain, modified Steiner-Steiner HT101-A
Reticulum Stain HT102-A
Trichrome stain LG solution - Gomoris one step trichrome procedure, 500 mls HT10-3-16
Trichrome stain AB solution - Gomoris one step trichrome procedure, 500 mls HT10-5-16
Trichrome Stain (Masson) - 1 kit, each component 250 mls HT15
Biebrich scarlet-acid fuchsin solution HT15-1
Phosphotungstic Acid Solution 10 % (w/v) HT15-2
Phosphomolybdic acid solution HT15-3
Iron stain HT20
Elastic stain HT25-A
Mucicarmine stock solution, 500 mls HT30-1-16
Mucicarmine stock solution, 250 mls HT30-1-8
Tartrazine solution, 120 ml cytoplasmic counter stain HT30-2-4
Tartrazine solution, 240 ml cytoplasmic counter stain HT30-2-8
Amyloid Stain, Congo Red Kit HT60
Methyl Green-Pyronin (MGP) solution HT70-1-16
Carbol-fuchsin solution, 500 mls HT80-1-16
Carbol-fuchsin solution, 250 mls HT80-1-8
Malachite green oxalate, 500 mls HT80-2-16
Malachite green oxalate, 250 mls HT80-2-8
Crystal Violet solution, 8 ounces HT90-1
Crystal Violet solution, 1 Liter HT90-1-32
Gram's iodine solution, 8 ounces HT90-2
Gram's iodine solution, 1 Liter HT90-2-32
Decolorizer solution HT90-3-32
Safranin O solution HT90-4-32
Gram Stain For Films HT90-A
Gram stain for tissue HT90-T
Nuclear Fast Red Solution. 0.1% nuclear fast red in 5% aluminum sulfate. Also referred to as Kernechtrot solution. Provides a red nuclear counterstain and as a substitute for hematoxylin, 100 ml N3020
back to top
Cell Separation
Product Name Product # 
HISTOPAQUE®-1077 Aseptically filtered solution, Density 1.077 g/mL, for Human Mononuclear Cell Separation 1077-1
HISTOPAQUE®-1083 Aseptically filtered solution, Density 1.083 g/mL, for small animal mononuclear cell separation 1083-1
HISTOPAQUE®-1119 Aseptically filtered solution, Density 1.119 g/mL, for Human Granulocytic series 1119-1
Accuspinä System-HISTOPAQUE®-1077 A0561
Accuspinä Tubes Sterile, 12 mL Capacity A1805
Accuspinä Tubes Nonsterile, 50 mL Capacity A1930
Accuspinä Tubes Sterile, 50 mL Capacity A2055
Accuspinä System-HISTOPAQUE®-1077, 40 each (3 ml fill of Histopaque in 12 ml tubes) A6929
Accuspinä System-HISTOPAQUE®-1077, 12 each (15 ml fill Histopaque in 50 ml tubes) A7054
back to top