Sigma-Aldrich
Life Science > Cell Biology > Ion Channels > Voltage Gated
Ion Channels
Voltage Gated
The majority of ion channels fall into two broad categories: voltage-gated ion channels (VGIC) and ligand-gated ion channels (LGIC). Members of the VGIC superfamily are usually closed at the resting potential of the cell. A change in the membrane potential causes conformational changes that result in the opening of the pore (voltage-dependent activation), which may be followed by a transitional conformational change (inactivation) to an inactivated state.1 The VGIC superfamily includes calcium channels, chloride channels, potassium channels and sodium channels. Other, smaller categories exist, such as the vanilloid (TRP) receptors, the ATP-gated channels, the cyclic nucleotide-gated channels (CNG) and aquaporins (water channels).
Voltage-gated-image Download a Powerpoint file of this image for personal or education use.
Request your free copies of our Ion Channels Technical brochure
and our Ion Channel Solutions brochure
.
Aquaporin (Water)
     Antibodies to Water Channels
Calcium
     Antibodies to Calcium Channels
     Calcium Channel Modulators
Chloride
     Antibodies to Chloride Channels
     Chloride Channel Modulators
Potassium
     Antibodies to Potassium Channels
     Potassium Channel Modulators
Sodium
     Antibodies to Sodium Channels
     Sodium Channel Modulators
Other Ion Channels
     Antibodies to Other Ion Channels
Ion Exchangers and Co-Transporters
     Ion Probes
     Ionophores
     Antibodies to Ion Pumps
     Ion Pump Inhibitors
Reference
  1. Ashcroft, F. M., Ed. Ion Channels and Disease; Academic Press: San Diego, CA, 2000.
Ion Channel Solutions page