|
Cell Biology
β Secretase (BACE1) Activity Assay Kit: A FRET Based Assay Designed for BACE1 Inhibitor Screening
Lisa Prostak, Efrat Barnea, Pnina Yaish and Dorit Zharhary Sigma-Aldrich Corporation
Introduction In recent years, the biological processes involved in the pathogenesis of Alzheimer’s disease (AD) have become increasingly better understood. It is now recognized that a key role is played by BACE1 proteases that cleave amyloid precursor protein (APP) to produce the amyloid β42 peptide fragment, ultimately resulting in the formation of amyloid plaques. Development of specific inhibitors of BACE1 proteases is thus one of the most active areas in the development of drugs for AD. This article describes a FRET-based screening system for the identification and assay of such inhibitors.
Background Due to the aging population in many countries, the prevalence of neurodegenerative diseases has increased, and finding a treatment or cure has become a focus of many researchers worldwide. AD is the most common cause of dementia, with age contributing to its onset. United Nations (UN) population projections estimate that the number of people older than 80 years will approach 370 million by the year 2050. Currently, it is estimated that 50% of people over age 85 are afflicted with AD, and that greater than 100 million people worldwide will suffer from dementia in 50 years. The vast number of people requiring constant care and related services will severely strain medical, monetary and human resources.
The onset of AD is gradual, with the appearance of clinical symptoms occurring between 60-70 years of age, and is characterized by the progressive loss of cognitive function. One of the primary causes for the progression of AD is believed to be the accumulation of amyloid beta peptide (Aβ) in the brain. Aβ is formed by the sequential cleaving of the amyloid precursor protein by the proteases BACE1 (β-secretase or beta-site APP-cleaving enzyme) and γ-secretase (Figure 1). The Aβ is secreted into the extracellular spaces and accumulates to form aggregates, fibrils and eventually amyloid deposits called senile plaques. The two types of Aβ formed from secretase cleavage are Aβ40 and Aβ42, with the latter being the prevalent form found in the senile plaques. Blocking the activity of BACE1 or γ-secretase can inhibit the production of Aβ peptides. Therefore, the identification of proteins or compounds that block secretase activity has been and remains a major goal of AD research.
This intense interest in studying the inhibition of γ- and β-secretases for therapeutic intervention in AD patients [1-3] is heightened by the fact that there is no effective drug for the treatment of the disease. Knockout studies show that BACE1 is critical for Aβ generation. Transgenic mice lacking BACE1 do not produce Aβ, but show an otherwise normal phenotype with no detrimental effects on viability or morphology [4]. This fact raises the possibility that BACE1 inhibition could be achieved without developing toxic side effects. Consequently, BACE1 has become a target for drug discovery research.
Figure 1. Cleavage of Amyloid Precursor Protein. Amyloid precursor protein (APP) is sequentially cleaved first by β-secretase (BACE1) and then by γ-secretase to form soluble amyloid precursor protein β (sAppβ) and the amyloid β42 peptide fragment (Aβ42). The Aβ42 fragments then aggregate and form the extracellular senile plaques common to Alzheimer’s disease.
back to top
Features and Benefits The Sigma-Aldrich BACE1 Activity Assay Kit (Product No. CS0010) is designed for BACE1 inhibitor screening and is based on a convenient method of fluorescence resonance energy transfer (FRET) in which the fluorescence signal is observed after the substrate is cleaved by BACE1 (Figure 2). Results are obtained in 2 hours, but data can be collected up to 24 hours after the addition of stop solution.
Figure 2. Principle of the Assay. The substrate is linked to a fluorescent dye on one end and to a quenching group on its other end. The fluorescence of the substrate is significantly reduced due to intramolecular resonance energy transfer to the quenching group. Upon substrate cleavage by the enzyme, there is a disturbance of the energy transfer resulting in the enhancement of the fluorescent signal.
The kit provides all the reagents required for the detection of BACE1 activity in a quantity sufficient for 250 inhibition reactions of 100 µl in 96 well plates. The reagents include fluorescence assay buffer, stop solution, substrate, standard and BACE1 enzyme (β-secretase).
back to top
Assay Procedure The BACE1 substrate is prepared in the buffer to a concentration of 50 µM. BACE1 enzyme is prepared in the same buffer to a concentration of approximately 0.3 units/µl. The standard is prepared to a concentration of 100 µM and added to the wells to give final values of 100 pmol, 200 pmol, 300 pmol and 500 pmol. Stock solutions of test samples are prepared in DMSO or other appropriate solvent, and then serially diluted in the buffer to give the desired working concentrations.
BACE1 enzyme, substrate, standard, test samples and buffer are then added to the wells for a total volume of 100 µl, with the BACE1 enzyme being added last, just prior to reading (Table 1).
Baseline fluorescence is recorded immediately after the addition of the BACE1 enzyme on a fluorometer set at excitation 320 nm, emission 405 nm. Plates are then covered and incubated at 37°C for 2 hours. Following which fluorescence is recorded again. Stop solution may then be added to stabilize the signal for up to 24 hours.
Table 1. Components of each assay run
| Sample |
Substrate |
Fluorescence Assay Buffer |
Test Sample |
Standard |
BACE1 enzyme |
| Negative Control |
20 µl |
80 µl |
none |
none |
none |
| Positive Control |
20 µl |
78 µl |
none |
none |
2 µl |
| Test Samples |
20 µl |
78-X µl |
X µl |
none |
2 µl |
| Standard (100 pmol) |
20 µl |
79 µl |
none |
1 µl |
none |
| Standard (200 pmol) |
20 µl |
78 µl |
none |
2 µl |
none |
| Standard (300 pmol) |
20 µl |
77 µl |
none |
3 µl |
none |
| Standard (500 pmol) |
20 µl |
75 µl |
none |
5 µl |
none |
back to top
Calculations and Results The BACE1 enzyme converts 5-20% of the substrate to a fluorescent product during 1-2 hours at 37°C. The standard curve is prepared by subtracting the fluorescence units (FU) of the negative control from all signal readings and plotting the FU against the concentrations of the standards (Figure 3). The FU of the test samples are extrapulated from the curve to give a pmol value, which is then used in the following equation to calculate the percent BACE1 cleaved by the sample:
% cleavage =
S x 50 500
Where S is the sample value in pmol and 500 is the value of the highest standard in pmol, which is calibrated to represent 50% BACE1 cleavage.
 Figure 3. Standard Curve
back to top
An inhibition curve can then be created by plotting the percent cleavage of each test sample dilution against the initial concentration on a logarithmic scale. An example curve is given in Figure 4. The curve was created using serial dilutions of the known statine-derived BACE1 inhibitor, [Asn670, Sta671, Val672]-Amyloid β/A4 Precursor Protein 770 Fragment [662-675], ammonium salt (Product No. A1847) in the standard kit assay protocol.
 Figure 4. Inhibition of BACE activity by a statine-derived inhibitor (Product No. A1847). BACE1 was incubated with various concentrations of the inhibitor for 2 hours at 37°C. BACE1 activity was determined using the BACE1 Activity Assay Kit (Product No. CS0010).
back to top
Additional Uses for BACE1 Assay Kit The BACE1 Activity Assay Kit can also be used to measure BACE1 activity in tissue homogenates. Figure 5 illustrates the measurement of BACE1 activity in rat brain and liver tissues using 10 µg of tissue homogenate per reaction. The activity was measured in the presence or absence of the inhibitor ([Asn670,Sta671,Val672]-Amyloid β/A4 Precursor Protein 770 Fragment [662-675] ammonium salt) at a concentration of 10 µM. The difference between the fluorescence obtained by the samples with and without the inhibitor (ΔFU) represents the actual BACE1 activity. As shown in Figure 5, BACE1 activity was detected in rat brain while there was no significant activity in rat liver.
 Figure 5. BACE1 activity in rat liver and brain homogenates. BACE1 activity was assayed in rat brain and liver homogenates according to the BACE1 Activity Assay Kit (Product No. CS0010). The results are presented as the Δ Fluorescence Units (ΔFU), i.e. the fluorescence detected in the sample with no inhibitor minus the fluorescence in the presence of the inhibitor ([Asn670, Sta671, Val672]-Amyloid β/A4 Precursor Protein 770 Fragment [662-675] ammonium salt, Product No. A1847). The fluorescence was followed for 4 hours. This Δ represents the net fluorescence obtained specifically from BACE1 activity.
In summary, the Sigma-Aldrich BACE1 Activity Assay Kit should serve as a useful tool to assay BACE1 enzyme activity. More importantly, the kit should assist researchers in identifying inhibitors of BACE1 proteases, a key player in the pathogenesis of AD.
References
- Citron, M., Beta-secretase as a target for the treatment of Alzheimer's disease., J. Neurosci. Res., 70, 373-379 (2002).
- Hong, L., et al., Memapsin 2 (beta secretase) as a therapeutic target., Biochem. Soc. Trans., 30, 530-534 (2002).
- Rochette, M.J., and Murphy, M.P., Gamma secretase: substrates and inhibitors., Mol. Neurobiol., 26, 81-95 (2002).
- Roberds, S.L., et al., BACE knockout mice are healthy despite lacking the primary beta-secretase activity in brain: implications for Alzheimer's disease therapeutics., Hum. Mol. Genet., 10, 1317- 1324 (2001).
back to top back to Cell Biology
|