| Product Code |
NAME |
Area of Interest |
Platform |
Size |
Sensitivity |
Species |
Description |
| CS0740 |
Acid Phosphatase Assay Kit |
Nitric Oxide/ Cell Stress |
Enzymatic |
1,000 in microwell plate |
0.2 mUnit |
mammalian |
Quantitative Acid Phosphatase activity measurement in tissues, whole cell extracts, column fractions and purified enzyme. Acid phosphatase can serve as a lysosomal marker |
| CS0120 |
Akt/PKB [pSer473] Phosphospecific ELISA |
Protein Phosphorylation |
ELISA Phosphospecific |
96 wells |
<0.8 units/mL |
h/m/r |
Quantitative ELISA for measurement of Akt/PKB phosphorylated at [pSer473] in cell lysates |
| CS0110 |
Akt/PKB [pThr308] Phosphospecific ELISA |
Protein Phosphorylation |
ELISA Phosphospecific |
96 wells |
<0.8 units/mL |
h/m/r |
Quantitative ELISA for measurement of Akt/PKB phosphorylated at [pThr308] in cell lysates |
| CS0160 |
Akt/PKB ELISA |
Protein Phosphorylation |
ELISA |
96 wells |
<0.4 ng/mL |
h/m/r |
Quantitative ELISA for measurement of Akt/PKB in cell lysates |
| CS0500 |
Amyloid Precursor Protein (APP) ELISA, Human |
Apoptosis/ Cell Cycle |
ELISA |
96 wells |
<0.4 ng/mL |
h |
Quantitative ELISA for measurement of APP in human cell lysates |
| APO-AC |
Annexin V-Cy3 Apoptosis Detection Kit |
Apoptosis/ Cell Cycle |
Fluorescence |
200 tests |
NA |
mammalian |
Fluorescent detection of Annexin V binding as an early apoptosis marker |
| APO-AF |
Annexin V-FITC Apoptosis Detection Kit |
Apoptosis/ Cell Cycle |
Flow Cytometry |
20 or 50 tests |
NA |
mammalian |
Assay of Annexin V binding as an early apoptosis marker by flow cytometry |
| APO-BRDU |
Apoptosis Flow Cytometry Kit |
Apoptosis/ Cell Cycle |
Flow Cytometry |
60 cell susp. |
|
mammalian |
Immunohistochemical labeling of Br-dUTP in apoptotic cells and flow cytometry detection |
| CS0550 |
ATF2 [pThr69/pThr71] Phosphospecific ELISA |
Gene Regulation |
ELISA Phosphospecific |
96 wells |
<0.8 units/mL |
h/m/r |
Quantitative ELISA for measurement of ATF2 phosphorylated at [pThr69/pThr71] in cell lysates |
| CS0540 |
ATF2 ELISA |
Gene Regulation |
ELISA |
96 wells |
<1 ng/mL |
h |
Quantitative ELISA for measurement of ATF2 in cell lysates |
| CS0010 |
BACE1 Activity Assay Kit (FRET) |
Neurobiology |
Enzymatic |
250 reactions in microwells |
10 ng purified enzyme/test |
h/r (tested) |
Quantitative b-secretase activity measurement and inhibitor screening by a fluorescence based assay |
| BE0100 |
b-Amyloid 1-40 ELISA, Human |
Neurobiology |
ELISA |
96 wells |
<10 pg/mL |
h |
Quantitative colorimetric ELISA for measurement of Ab40 in biological fluids |
| BE0200 |
b-Amyloid 1-42 ELISA, Human |
Neurobiology |
ELISA |
96 wells |
<10 pg/mL |
h |
Quantitative colorimetric ELISA for measurement of Ab42 in biological fluids |
| CS0530 |
Bcl-2 [pSer70] Phosphospecific ELISA |
Apoptosis/ Cell Cycle |
ELISA Phosphospecific |
96 wells |
<0.5 units/mL |
h |
Quantitative ELISA for measurement of Bcl-2 phosphorylated at [pSer70] in cell lysates |
| CS0520 |
Bcl-2 ELISA, Human |
Apoptosis/ Cell Cycle |
ELISA |
96 wells |
<0.1 ng/mL |
h |
Quantitative ELISA for measurement of Bcl-2 in human cell lysates |
| CA-201 |
cAMP Enzyme Immunoassay Kit |
G Proteins |
ELISA |
96 wells |
0.037 pmol/mL |
h |
Quantitative colorimetric immunoassay of cAMP in biological fluids or samples with low cAMP concentration |
| CA-200 |
cAMP Enzyme Immunoassay Kit, Direct |
G Proteins |
ELISA |
96 wells |
0.039 pmol/mL |
h |
Quantitative colorimetric immunoassay of total cAMP in tissue and cell cultures |
| CS0400 |
Casein Kinase Assay Kit - General |
Protein Phosphorylation |
Enzymatic |
100 tests |
10 ng CKI and 0.06 units CKII |
mammalian |
Enzymatic assay of different casein kinases by an in vitro phosphorylation of a-casein and radioactive detection in cell lysates |
| CS0600 |
Casein Kinase I Assay Kit |
Protein Phosphorylation |
Enzymatic |
50 tests |
10 ng purified enzyme |
mammalian |
Enzymatic activity of casein kinase I (CK-1) assayed by radioactive detection of the phosphorylated peptide substrate |
| CS0610 |
Casein Kinase II Assay Kit |
Protein Phosphorylation |
Enzymatic |
70 tests |
0.02 units |
mammalian |
Enzymatic activity of casein kinase II (CK-2) assayed by radioactive detection of the phosphorylated peptide substrate |
| CS0280 |
Caspase 1 FAM-FLICA Assay |
Apoptosis/ Cell Cycle |
Histochemistry |
100 tests |
NA |
mammalian |
Fluorometric or flow cytometry detection of active cellular caspase 1 using Fluorochrome Inhibitor of Caspase 1 activity (FLICA) |
| CS0310 |
Caspase 10 FAM-FLICA Assay |
Apoptosis/ Cell Cycle |
Histochemistry |
100 tests |
NA |
mammalian |
Fluorometric or flow cytometry detection of active cellular caspase 10 using Fluorochrome Inhibitor of Caspase 10 activity (FLICA) |
| CS0320 |
Caspase 13 FAM-FLICA Assay |
Apoptosis/ Cell Cycle |
Histochemistry |
100 tests |
NA |
mammalian |
Fluorometric or flow cytometry detection of active cellular caspase 13 using Fluorochrome Inhibitor of Caspase 13 activity (FLICA) |
| CS0290 |
Caspase 2 FAM-FLICA Assay |
Apoptosis/ Cell Cycle |
Histochemistry |
100 tests |
NA |
mammalian |
Fluorometric or flow cytometry detection of active cellular caspase 2 using Fluorochrome Inhibitor of Caspase 2 activity (FLICA) |
| CASP-3-C |
Caspase 3 Assay Kit, Colorimetric |
Apoptosis/ Cell Cycle |
Enzymatic |
800 tests in 96 plate |
Linear between 1-20 nmole/well |
mammalian |
Quantitative colorimetric assay of Caspase 3 activity detected by the release of p-nitroaniline (pNA) from the Caspase-3 substrate |
| CASP-3-F |
Caspase 3 Assay Kit, Fluorimetric |
Apoptosis/ Cell Cycle |
Enzymatic |
1,000 in microwell plate |
0.1-1.2 nmol in microwell or 0.05-0.5 nmol in 2ml. |
mammalian |
Quantitative fluorimetric assay of Caspase 3 activity detected by the release of 7-amino-4-methylcoumarin (AMC) from the Caspase-3 substrate |
| CASP-6-F |
Caspase 6 Assay Kit, Fluorimetric |
Apoptosis/ Cell Cycle |
Enzymatic |
1,000 in microwell plate |
0.2-2 nmol |
mammalian |
Quantitative fluorimetric assay of Caspase 6 activity detected by the release of 7-amino-4-methylcoumarin (AMC) from the Caspase-6 substrate |
| CASP-8-C |
Caspase 8 Assay Kit, Colorimetric |
Apoptosis/ Cell Cycle |
Enzymatic |
100 tests of 100 ml |
0.003-0.0025 units/test |
mammalian |
Quantitative colorimetric assay of Caspase 8 activity detected by the release of p-nitroaniline (pNA) from the Caspase-8 substrate |
| CASP-8-F |
Caspase 8 Assay Kit, Fluorimetric |
Apoptosis/ Cell Cycle |
Enzymatic |
1,000 in microwell plate |
Linear 0.1-1 nmole standard/well |
mammalian |
Quantitative fluorimetric assay of Caspase 8 activity detected by the release of 7-amino-4-methylcoumarin (AMC) from the Caspase-8 substrate |
| CS0300 |
Caspase 9 FAM-FLICA Assay |
Apoptosis/ Cell Cycle |
Histochemistry |
100 tests |
NA |
mammalian |
Fluorometric or flow cytometry detection of active cellular caspase 9 using Fluorochrome Inhibitor of Caspase 9 activity (FLICA) |
| CAT-100 |
Catalase Assay Kit |
Nitric Oxide/ Cell Stress |
Enzymatic |
100 tests/mL |
0.01-0.08 units/min UV 0.3-1.75 UV units/ 5 min |
mammalian |
Quantitative enzymatic assay of catalase activity in tissue lysates. Can serve as a peroxisomal marker |
| CS0370 |
Cathepsin B Detection Assay |
Apoptosis/ Cell Cycle |
Histochemistry |
100 assays |
NA |
h |
Fluorescence detection of cathepsin B by protease activity in living cells with cresyl violet fluorophore |
| CS0360 |
Cathepsin K Detection Assay |
Apoptosis/ Cell Cycle |
Histochemistry |
100 assays |
NA |
h |
Fluorescence detection of cathepsin K by protease activity in living cells with cresyl violet fluorophore |
| CS0350 |
Cathepsin L Detection Assay |
Apoptosis/ Cell Cycle |
Histochemistry |
100 assays |
NA |
h |
Fluorescence detection of cathepsin L by protease activity in living cells with cresyl violet fluorophore |
| CE0050 |
CelLyticTM MEM Protein Extraction Kit |
Cytoskeleton/ Extracellular Matrix |
Cell Fractionation |
80 tests |
NA |
mammalian |
Isolation of hydrophobic and raft associated proteins from cells using a phase separation method
|
| CG-201 |
cGMP Enzyme Immunoassay Kit |
G Proteins |
EIA |
96 wells |
0.059 pmol/mL |
h |
Quantitative colorimetric immunoassay of cGMP in biological fluids or samples with low cGMP concentration |
| CG-200 |
cGMP Enzyme Immunoassay Kit, Direct |
G Proteins |
EIA |
96 wells |
0.088 pmol/mL |
h |
Quantitative colorimetric immunoassay of total cGMP in tissue and cell cultures |
| CS0720 |
Citrate Synthase Assay Kit |
Apoptosis/ Cell Cycle |
Enzymatic |
100 test/1mL 480 tests/well |
20 ng/ml purified enzyme |
mammalian |
Colorimetric assay of citrate synthase activity, a mitochondrial matrix marker |
| CS0590 |
c-Met [pTyr1230/pTyr1234/pTyr1235] Phosphospecific ELISA |
Protein Phosphorylation |
ELISA Phosphospecific |
96 wells |
<0.25 units/mL |
h |
Quantitative ELISA for measurement of c-Met phosphorylated at [pTyr1230/pTyr1234/pTyr1235] in cell lysates |
| CS0560 |
c-Met ELISA |
Protein Phosphorylation |
ELISA |
96 wells |
<0.4 ng/mL |
h |
Quantitative ELISA for measurement of c-Met in human cell lysates |
| CS0570 |
CREB [pSer133] Phosphospecific ELISA |
Gene Regulation |
ELISA Phosphospecific |
96 wells |
<0.9 units/mL |
h/m |
Quantitative ELISA for measurement of CREB phosphorylated at [pSer133] in cell lysates |
| CS0580 |
CREB ELISA |
Gene Regulation |
ELISA |
96 wells |
<0.025 ng/mL |
h/m |
Quantitative ELISA for measurement of CREB in cell lysates |
| ACE100 |
Cultured Cells Accelularization Kit |
Cytoskeleton/ Extracellular Matrix |
Cell Fractionation |
30 preparations |
NA |
mammalian |
Growth and separation of ECM secreting cells |
| CS0230 |
Cysteinyl Leukotriene EIA |
Lipid Signaling |
EIA |
96 wells |
<26.6 pg/mL |
h/m/r |
Competitive immunoassay (EIA) for the quantitative determination of cysteinyl leukotriene in biological samples and lysates |
| CYTOC-OX1 |
Cytochrome c Oxidase Assay Kit |
Apoptosis/ Cell Cycle |
Enzymatic |
100 tests/mL |
0.4-4 milliunits/min |
mammalian/ yeast |
Colorimetric assay for Cytochrome c oxidase activity. Can serve as a mitochondrial protein marker |
| CY0100 |
Cytochrome c Reductase [NADPH] Assay Kit |
Multi-Drug Resistance |
Enzymatic |
100 assays/mL |
0.8-4.1 milliunits/min |
mammalian/ yeast |
Quantitative enzymatic assay of NADPH-cytochrome c reductase in cell and tissue extracts and in purified ER. Can serve as an ER marker |
| CYTO-1A |
Cytochrome P450 Fluorescent Detection Kit (P4501A1/2) |
Multi-Drug Resistance |
Enzymatic |
7.5 g tissue or 50 assays |
6-250 pmole/min |
mammalian |
Fluorescent assay of Cytochrome P450 in tissue extracts |
| CYTO-2B |
Cytochrome P450 Fluorescent Detection Kit (P4502B4) |
Multi-Drug Resistance |
Enzymatic |
7.5 g tissue or 50 assays |
6-250 pmole/min |
mammalian |
Fluorescent assay of Cytochrome P450 in tissue extracts |
| CS0340 |
Dihydrofolate Reductase Assay Kit |
Nitric Oxide/ Cell Stress |
Enzymatic |
50-100 tests/mL |
500 ng purified enzyme |
mammalian |
Quantitative DHFR activity measurement and inhibitor screening by a spectrophotometric based assay |
| ER0100 |
Endoplasmic Reticulum Isolation Kit |
Intracellular Calcium Signaling |
Cell Fractionation |
~50 g tissue or 20 ml packed cells |
NA |
mammalian |
Isolation of enriched endoplasmic reticulum (ER) fraction from cells and tissues |
| CS0150 |
Epidermal Growth Factor Receptor (EGFR) [pTyr1068] Phosphospecific ELISA, Human |
Cytokines/Growth Factors/Hormones |
ELISA Phosphospecific |
96 wells |
<0.3 units/mL |
h |
Quantitative ELISA for measurement of EGFR phosphorylated on [pTyr1068] in cell lysates |
| CS0140 |
Epidermal Growth Factor Receptor (EGFR) [pTyr1173] Phosphospecific ELISA, Human |
Cytokines/Growth Factors/Hormones |
ELISA Phosphospecific |
96 wells |
<0.3 units/mL |
h |
Quantitative ELISA for measurement of EGFR phosphorylated on [pTyr1173] in cell lysates |
| CS0080 |
Epidermal Growth Factor Receptor (EGFR) ELISA, Human |
Cytokines/Growth Factors/Hormones |
ELISA |
96 wells |
<16 pg/mL |
h |
Quantitative ELISA for measurement of EGFR in cell lysates |
| PE0100 |
ERK 1&2 [pThr185/pTyr187] Phosphospecific ELISA |
Protein Phosphorylation |
ELISA Phosphospecific |
96 wells |
<0.8 units/mL |
h/m/r |
Quantitative ELISA for measurement of ERK 1&2 phosphorylated at [pThr185/pTyr187] in cell lysates |
| EK0100 |
ERK 1&2 ELISA |
Protein Phosphorylation |
ELISA |
96 wells |
<16 pg/mL |
h/m/r |
Quantitative ELISA for measurement of ERK 1&2 in cell lysates |
| CS0260 |
Glutathione Assay Kit |
Nitric Oxide/ Cell Stress |
Enzymatic |
500 tests in microwell |
|
|
Quantative measurment of total glutathione (reduced+disulfide) levels in biological samples |
| CGP-1 |
Glutathione Peroxidase Cellular Activity Assay Kit |
Nitric Oxide/ Cell Stress |
Enzymatic |
100 tests |
0.005-0.1 units/3 min |
mammalian |
Quantitative enzymatic assay of gluthathione peroxidase (GPx) in tissue lysates |
| GR-SA |
Glutathione Reductase Assay Kit |
Nitric Oxide/ Cell Stress |
Enzymatic |
150 tests |
Colorimetric assay: 0.003-0.03 units; UV assay: 0.003-0.012 units |
mammalian/ yeast |
Quantitative enzymatic assay of glutathione reductase in tissue lysates |
| CS0410 |
Glutathione S-Transferase (GST) Assay Kit |
Nitric Oxide/ Cell Stress |
Enzymatic |
500 tests in microwells |
60 ng purified enzyme/test |
mammalian/ bacteria |
Quantitative enzymatic assay of GST activity in bacteria, mammalian cells and tissue lysates |
| CS0630 |
HSP27 [pSer82] Phosphospecific ELISA Human |
Nitric Oxide/ Cell Stress |
ELISA Phosphospecific |
96 wells |
<1.0 units/mL |
h |
Quantitative ELISA for measurement of HSP27 phosphorylated at [pSer82] in human cell lysates |
| CS0640 |
HSP27 ELISA, Human |
Nitric Oxide/ Cell Stress |
ELISA |
96 wells |
<0.1 ng/mL |
h |
Quantitative ELISA for measurement of HSP27 in human cell lysates |
| CS0270 |
Hydrogen Peroxide Colorimetric Assay |
Nitric Oxide/ Cell Stress |
Enzymatic |
96 wells |
51.3 ng/mL |
|
Quantitative H2O2 determination in serum, plasma, urine and cell culture supernatants |
| CS0660 |
IkBa [pSer32] Phosphospecific ELISA |
Gene Regulation |
ELISA Phosphospecific |
96 wells |
<0.5 units/mL |
h |
Quantitative ELISA for measurement of IsymbolBa phosphorylated at [pSer32] in cell lysates |
| CS0620 |
IkBa ELISA |
Gene Regulation |
ELISA |
96 wells |
<30 pg/mL |
h |
Quantitative ELISA for measurement of IkBa in human cell lysates |
| PI0100 |
Insulin Receptor [pTyr1158] Phosphospecific ELISA |
Cytokines/Growth Factors/Hormones |
ELISA Phosphospecific |
96 wells |
<0.8 units/mL |
h/m/r |
Quantitative ELISA for measurement of IR phosphorylated at [pTyr1158] in cell lysates |
| PI0200 |
Insulin Receptor [pTyr1162/pTyr1163] Phosphospecific ELISA |
Cytokines/Growth Factors/Hormones |
ELISA Phosphospecific |
96 wells |
<0.8 units/mL |
h/m/r |
Quantitative ELISA for measurement of IR phosphorylated at [pTyr1162/pTyr1163] in cell lysates |
| CS0090 |
Insulin Receptor b Subunit Receptor ELISA |
Cytokines/Growth Factors/Hormones |
ELISA |
96 wells |
<0.5 ng/mL |
h/m/r |
Quantitative ELISA for measurement of IR b-subunit in cell lysates |
| CS0130 |
JNK 1&2 [pThr183/pTyr185] Phosphospecific ELISA |
Protein Phosphorylation |
ELISA Phosphospecific |
96 wells |
<0.8 units/mL |
h/m |
Quantitative ELISA for measurement of JNK 1&2 phosphorylated at [pThr183/pTyr185] in cell lysates |
| CS0100 |
JNK 1&2 ELISA |
Protein Phosphorylation |
ELISA |
96 wells |
<0.15 ng/mL |
h |
Quantitative ELISA for measurement of JNK 1&2 in cell lysates |
| CS0380 |
JNK Activity Assay Kit |
Protein Phosphorylation |
Immuno-precipitation |
50 reactions |
NA |
mammalian |
Enzymatic activity of JNK immunoprecipitated from cell lysates, assayed by immunoblotting of the phosphorylated JNK substrate ATF2 |
| CS0220 |
Leukotriene B4 EIA |
Lipid Signaling |
EIA |
96 wells |
19.4 pg/mL |
h/m/r |
Competitive immunoassay (EIA) for the quantitative determination of LTB4 in biological samples and lysates |
| LYSIS01 |
Lysosome Isolation Kit |
Lipid Signaling |
Cell Fractionation |
25 g tissue or 20 ml packed cells |
NA |
mammalian |
Isolation of enriched lysosomal fraction from cells and tissues |
| CS0650 |
MEK1 [pSer218/pSer222] Phosphospecific ELISA |
Protein Phosphorylation |
ELISA Phosphospecific |
96 wells |
<1.0 units/mL |
h/m |
Quantitative ELISA for measurement of MEK1 phosphorylated at [pSer218/pSer222] in cell lysates |
| CS0490 |
MEK1 Assay Kit |
Protein Phosphorylation |
Immuno-precipitation |
50 reactions |
NA |
mammalian |
Enzymatic activity of MEK1 immunoprecipitated from cell lysates, assayed by immunoblotting of the phosphorylated MEK1 substrate ERK2 |
| CS0700 |
MEK1 ELISA |
Protein Phosphorylation |
ELISA |
96 wells |
<0.1 ng/mL |
h |
Quantitative ELISA for measurement of MEK1 in cell lysates |
| MD0100 |
MGMT Assay Kit |
Nitric Oxide/ Cell Stress |
Enzymatic |
100-200 tests |
0.002-0.02 mmol/hr |
mammalian |
Radioactive detection of MGMT (DNA repair protein) activity and inhibition in cell lysates |
| MI0500 |
Microtubules Preparation Kit |
Cytoskeleton/ Extracellular Matrix |
Cell Fractionation |
50 g tissue or 30 cell preparations |
NA |
h/r/rb/swine |
Microtubules preparation from cells and tissues by homogenization followed by temperature dependent assembly-dissasembly cycles |
| MITO-ISO1 |
Mitochondria Isolation Kit |
Apoptosis/ Cell Cycle |
Cell Fractionation |
10-20 g tissue, or 50 tests/ 2 ml |
NA |
mammalian |
Isolation of enriched mitochondrial fraction from tissues |
| CS0390 |
Mitochondria Staining Kit |
Apoptosis/ Cell Cycle |
Histochemistry |
200 x 1 ml tests |
NA |
mammalian |
Detection of changes in electrochemical potential of mitochondrial membrane by staining cells with JC-1 dye and fluorometric detection |
| NOS-1 |
Nitric Oxide Synthase (NOS) Detection Kit, Isotopic |
Nitric Oxide/ Cell Stress |
Enzymatic |
50 assays |
|
h/r |
Quantitative enzymatic assay of nitric oxidase synthase (NOS) in cell lysates |
| FCANOS-1 |
Nitric Oxide Synthase Detection System, Fluorometric |
Nitric Oxide/ Cell Stress |
Enzymatic |
96 wells |
|
h/m |
Quantitative enzymatic fluorimetric detection of nitric oxide (NO) in living cells |
| CS0710 |
Ogg1 Assay Kit |
Nitric Oxide/ Cell Stress |
Enzymatic |
180 assays |
10 mg/ml purified enzyme |
h (tested) |
Radioactive detection of Ogg1 enzyme activity (DNA repair) in cell lysates |
| CS0670 |
Osteocalcin (OST) ELISA, Human |
Cytoskeleton/ Extracellular Matrix |
ELISA |
96 wells |
0.4 ng/mL |
h |
Quantitative amplified ELISA for measurement of Osteocalcin in biological fluids |
| PM0100 |
p38 MAPK |
Protein Phosphorylation |
ELISA |
96 wells |
<16 pg/mL |
h/m/r |
Quantitative ELISA for measurement of p38 MAPK in cell lysates |
| CS0020 |
p38 MAPK [pThr180/pTyr182] Phosphospecific ELISA |
Protein Phosphorylation |
ELISA Phosphospecific |
96 wells |
<0.8 units/mL |
h/m/r |
Quantitative ELISA for measurement of p38 MAPK phosphorylated at [pThr180/pTyr182] in cell lysates |
| CS0250 |
p38 MAPK Activity Assay Kit |
Protein Phosphorylation |
Immuno-precipitation |
50 reactions |
NA |
mammalian |
Enzymatic activity of p38 immunoprecipitated from cell lysates, assayed by immunoblotting of the phosphorylated p38 substrate ATF2 |
| CS0040 |
p53 [pSer15] Phosphospecific ELISA, Human |
Apoptosis/ Cell Cycle |
ELISA Phosphospecific |
96 wells |
<0.9 units/mL |
h |
Quantitative ELISA for measurement of p53 phosphorylated at [pSer15] in cell lysates |
| CS0070 |
p53 ELISA, Human |
Apoptosis/ Cell Cycle |
ELISA |
96 wells |
<50 pg/mL |
h |
Quantitative ELISA for measurement of p53 in cell lysates |
| PD-1 |
PeroxiDetect Kit |
Nitric Oxide/ Cell Stress |
Enzymatic |
100 tests for aqueous or organic peroxides |
H2O2 1-7 nmoles/ test |
mammalian |
Quantitative enzymatic assay of hydrogen peroxide in aqueous solutions of cell lysates |
| PEROX-1 |
Peroxisome Isolation Kit |
Gene Regulation |
Cell Fractionation |
50 g tissue or 20 ml packed cells |
NA |
h/r/rb/swine |
Isolation of enriched peroxisomal fraction from cells and tissues |
| CS0200 |
Prostaglandin E1 EIA |
Lipid Signaling |
EIA |
96 wells |
6.06 pg/mL |
h/m/r |
Competitive immunoassay (EIA) for the quantitative determination of PGE1 in biological samples and lysates |
| CS0210 |
Prostaglandin E2 EIA |
Lipid Signaling |
EIA |
96 wells |
15.9 pg/mL |
h/m/r |
Competitive immunoassay (EIA) for the quantitative determination of PGE2 in biological samples and lysates |
| PTK-101 |
Protein Tyrosine Kinase Assay Kit, Non-Radioactive |
Protein Phosphorylation |
EIA |
96 wells |
0.2-0.4 Units |
all species |
Quantitative colorimetric immunoassay of PTK activity in cells and tissue lysates |
| PTP-101 |
Protein Tyrosine Phosphatase Assay Kit, Non-Radioactive |
Protein Phosphorylation |
Enzymatic |
96 wells |
400-500 pmol (PO4--)
|
all species |
The enzymatic reaction and detection of the inorganic phosphate released from PTPase |
| CS0060 |
Retinoblastoma [pSer249/pThr252] Phosphospecific ELISA, Human |
Apoptosis/ Cell Cycle |
ELISA Phosphospecific |
96 wells |
<0.8 units/mL |
h |
Quantitative ELISA for measurement of Rb phosphorylated at [pSer249/pThr252] in cell lysates |
| CS0420 |
Retinoblastoma [pThr821] Phosphospecific ELISA, Human |
Apoptosis/ Cell Cycle |
ELISA Phosphospecific |
96 wells |
<0.5 units/mL |
h |
Quantitative ELISA for measurement of Rb phosphorylated at [pThr821] in cell lysates |
| CS0050 |
Retinoblastoma ELISA, Human |
Apoptosis/ Cell Cycle |
ELISA |
96 wells |
<0.1 ng/mL |
h |
Quantitative ELISA for measurement of Rb in cell lysates |
| CS0030 |
Senescent Cells Staining Kit |
Apoptosis/ Cell Cycle |
Histochemistry |
100 tests |
NA |
mammalian |
Histochemical staining of b-galactosidase activity in senescent cells |
| CS0460 |
Src [pTyr418] Phosphospecific ELISA |
Protein Phosphorylation |
ELISA Phosphospecific |
96 wells |
<0.1 units/mL |
h/m/r |
Quantitative ELISA for measurement of Src phosphorylated at [pTyr418] in mouse cell lysates |
| CS0450 |
Src ELISA, Human |
Protein Phosphorylation |
ELISA |
96 wells |
<1.0 ng/mL |
h/m/r |
Quantitative ELISA for measurement of Src in human cell lysates |
| CS0480 |
STAT1 [pThr701] Phosphospecific ELISA |
Gene Regulation |
ELISA Phosphospecific |
96 wells |
<0.1 units/mL |
h |
Quantitative ELISA for measurement of STAT1 phosphorylated at [pThr701] in cell lysates |
| CS0470 |
STAT1 ELISA |
Gene Regulation |
ELISA |
96 wells |
<0.27 ng/mL |
h |
Quantitative ELISA for measurement of STAT1 in cell lysates |
| CS0680 |
sTNF-RI ELISA |
Cytokines/Growth Factors/Hormones |
ELISA |
96 wells |
0.05 ng/mL |
h |
Quantitative amplified ELISA for measurement of soluble TNFRI in biological fluids |
| CS0690 |
sTNF-RII (75kDa) ELISA |
Cytokines/Growth Factors/Hormones |
ELISA |
96 wells |
0.1 ng/mL |
h |
Quantitative amplified ELISA for measurement of soluble TNFRII in biological fluids |
| CS0180 |
Substance P EIA |
Neurotransmission |
EIA |
96 wells |
8.04 pg/mL |
h/p |
Competitive immunoassay (EIA) for the quantitative determination of Substance P in biological samples and lysates |
| CS0440 |
Tau [pSer199] Phosphospecific ELISA, Mouse |
Cytoskeleton/ Extracellular Matrix |
ELISA Phosphospecific |
96 wells |
<2 pg/mL |
m |
Quantitative ELISA for measurement of Tau phosphorylated at [pSer199] in mouse cell lysates |
| CS0430 |
Tau ELISA, Mouse |
Cytoskeleton/ Extracellular Matrix |
ELISA |
96 wells |
<15 pg/mL |
m |
Quantitative ELISA for measurement of Tau in mouse cell lysates |
| CS0170 |
Thioredoxin Reductase Assay Kit |
Nitric Oxide/ Cell Stress |
Enzymatic |
100 tests |
0.05-0.5 nmoles per well |
mammalian |
Quantitative enzymatic assay of mammalian thioredoxin reductase |
| CS0190 |
Thromboxane B2 EIA |
Lipid Signaling |
EIA |
96 wells |
10.54 pg/mL |
h/m/r |
Competitive immunoassay (EIA) for the quantitative determination of TXB2 in biological samples and lysates |
| CS0510 |
TRAIL ELISA, Human |
Apoptosis/ Cell Cycle |
ELISA |
96 wells |
<20 pg/mL |
h |
Quantitative ELISA for measurement of TRAIL in cell lysates |
| CS0730 |
Tyrosine Kinase Assay Kit |
Protein Phosphorylation |
Dot Blot |
50 reactions |
NA |
mammalian |
Enzymatic activity measurement of tyrosine kinase immunoprecipitated from cell lysates by radioactive or dot blot detection of the phosphorylated poly (Glu, Tyr) substrate |