Sigma-Aldrich
Life Science > Cell Biology > Learning Center > Cell Biology Assays > Kit List - Platform
Cell Signaling & Neuroscience
Kit List - Platform
  Cell Fractionation
Dot Blot
EIA
ELISA
ELISA Phosphospecific
Enzymatic
Flow Cytometry
Fluorescence
Histochemistry
Immunoprecipitation
Product Code NAME Area of Interest Platform Size Sensitivity Species Description
ACE100 Cultured Cells Accelularization Kit Cytoskeleton/Extracellular Matrix Cell Fractionation 30 preparations NA mammalian Growth and separation of ECM secreting cells
CE0050 CelLyticTM MEM Protein Extraction Kit Cytoskeleton/Extracellular Matrix Cell Fractionation 80 tests NA mammalian Isolation of hydrophobic and raft associated proteins from cells using a phase separation method
ER0100 Endoplasmic Reticulum Isolation Kit Intracellular Calcium Signaling Cell Fractionation ~50 g tissue or 20 ml packed cells NA mammalian Isolation of enriched endoplasmic reticulum (ER) fraction from cells and tissues
LYSIS01 Lysosome Isolation Kit Lipid Signaling Cell Fractionation 25 g tissue or 20 ml packed cells NA mammalian Isolation of enriched lysosomal fraction from cells and tissues
MI0500 Microtubules Preparation Kit Cytoskeleton/Extracellular Matrix Cell Fractionation 50 g tissue or 30 cell preparations NA h/r/rb/swine Microtubules preparation from cells and tissues by homogenization followed by temperature dependent assembly-dissasembly cycles
MITO-ISO1 Mitochondria Isolation Kit Apoptosis/Cell Cycle Cell Fractionation 10-20 g tissue, or 50 tests/ 2 ml NA mammalian Isolation of enriched mitochondrial fraction from tissues
PEROX-1 Peroxisome Isolation Kit Gene Regulation Cell Fractionation 50 g tissue or 20 ml packed cells NA h/r/rb/swine Isolation of enriched peroxisomal fraction from cells and tissues
CS0730 Tyrosine Kinase Assay Kit Protein Phosphorylation Dot Blot 50 reactions NA mammalian Enzymatic activity measurement of tyrosine kinase immunoprecipitated from cell lysates by radioactive or dot blot detection of the phosphorylated poly (Glu, Tyr) substrate
CA-201 cAMP Enzyme Immunoassay Kit G Proteins EIA 96 wells 0.037 pmol/mL h Quantitative colorimetric immunoassay of cAMP in biological fluids or samples with low cAMP concentration
CA-200 cAMP Enzyme Immunoassay Kit, Direct G Proteins EIA 96 wells 0.039 pmol/mL h Quantitative colorimetric immunoassay of total cAMP in tissue and cell cultures
CG-201 cGMP Enzyme Immunoassay Kit G Proteins EIA 96 wells 0.059 pmol/mL h Quantitative colorimetric immunoassay of cGMP in biological fluids or samples with low cGMP concentration
CG-200 cGMP Enzyme Immunoassay Kit, Direct G Proteins EIA 96 wells 0.088 pmol/mL h Quantitative colorimetric immunoassay of total cGMP in tissue and cell cultures
CS0230 Cysteinyl Leukotriene EIA Lipid Signaling EIA 96 wells <26.6 pg/mL h/m/r Competitive immunoassay (EIA) for the quantitative determination of cysteinyl leukotriene in biological samples and lysates
CS0220 Leukotriene B4 EIA Lipid Signaling EIA 96 wells 19.4 pg/mL h/m/r Competitive immunoassay (EIA) for the quantitative determination of LTB4 in biological samples and lysates
CS0200 Prostaglandin E1 EIA Lipid Signaling EIA 96 wells 6.06 pg/mL h/m/r Competitive immunoassay (EIA) for the quantitative determination of PGE1 in biological samples and lysates
CS0210 Prostaglandin E2 EIA Lipid Signaling EIA 96 wells 15.9 pg/mL h/m/r Competitive immunoassay (EIA) for the quantitative determination of PGE2 in biological samples and lysates
CS0180 Substance P EIA Neurotransmission EIA 96 wells 8.04 pg/mL h/p Competitive immunoassay (EIA) for the quantitative determination of Substance P in biological samples and lysates
CS0190 Thromboxane B2 EIA Lipid Signaling EIA 96 wells 10.54 pg/mL h/m/r Competitive immunoassay (EIA) for the quantitative determination of TXB2 in biological samples and lysates
PTK-101 Protein Tyrosine Kinase Assay Kit, Non-Radioactive Protein Phosphorylation EIA 96 wells 0.2-0.4 Units all species Quantitative colorimetric immunoassay of PTK activity in cells and tissue lysates
CS0160 Akt/PKB ELISA Protein Phosphorylation ELISA 96 wells <0.4 ng/mL h/m/r Quantitative ELISA for measurement of Akt/PKB in cell lysates
CS0500 Amyloid Precursor Protein (APP) ELISA, Human Apoptosis/Cell Cycle ELISA 96 wells <0.4 ng/mL h Quantitative ELISA for measurement of APP in human cell lysates
BE0100 b-Amyloid 1-40 ELISA, Human Neurobiology ELISA 96 wells <10 pg/mL h Quantitative colorimetric ELISA for measurement of Ab40 in biological fluids
BE0200 b-Amyloid 1-42 ELISA, Human Neurobiology ELISA 96 wells <10 pg/mL h Quantitative colorimetric ELISA for measurement of Ab42 in biological fluids
CS0540 ATF2 ELISA Gene Regulation ELISA 96 wells <1 ng/mL h Quantitative ELISA for measurement of ATF2 in cell lysates
CS0520 Bcl-2 ELISA, Human Apoptosis/Cell Cycle ELISA 96 wells <0.1 ng/mL h Quantitative ELISA for measurement of Bcl-2 in human cell lysates
CS0560 c-Met ELISA Protein Phosphorylation ELISA 96 wells <0.4 ng/mL h Quantitative ELISA for measurement of c-Met in human cell lysates
CS0580 CREB ELISA Gene Regulation ELISA 96 wells <0.025 ng/mL h/m Quantitative ELISA for measurement of CREB in cell lysates
CS0080 Epidermal Growth Factor Receptor (EGFR) ELISA, Human Cytokines/Growth Factors/Hormones ELISA 96 wells <16 pg/mL h Quantitative ELISA for measurement of EGFR in cell lysates
EK0100 ERK 1&2 ELISA Protein Phosphorylation ELISA 96 wells <16 pg/mL h/m/r Quantitative ELISA for measurement of ERK 1&2 in cell lysates
CS0640 HSP27 ELISA, Human Nitric Oxide/Cell Stress ELISA 96 wells <0.1 ng/mL h Quantitative ELISA for measurement of HSP27 in human cell lysates
CS0620 IkBa ELISA Gene Regulation ELISA 96 wells <30 pg/mL h Quantitative ELISA for measurement of IkBain human cell lysates
CS0090 Insulin Receptor b Subunit Receptor ELISA Cytokines/Growth Factors/Hormones ELISA 96 wells <0.5 ng/mL h/m/r Quantitative ELISA for measurement of IR b-subunit in cell lysates
CS0100 JNK 1&2 ELISA Protein Phosphorylation ELISA 96 wells <0.15 ng/mL h Quantitative ELISA for measurement of JNK 1&2 in cell lysates
CS0700 MEK1 ELISA Protein Phosphorylation ELISA 96 wells <0.1 ng/mL h Quantitative ELISA for measurement of MEK1 in cell lysates
CS0670 Osteocalcin (OST) ELISA, Human Cytoskeleton/Extracellular Matrix ELISA 96 wells 0.4 ng/mL h Quantitative amplified ELISA for measurement of Osteocalcin in biological fluids
PM0100 p38 MAPK Protein Phosphorylation ELISA 96 wells <16 pg/mL h/m/r Quantitative ELISA for measurement of p38 MAPK in cell lysates
CS0070 p53 ELISA, Human Apoptosis/Cell Cycle ELISA 96 wells <50 pg/mL h Quantitative ELISA for measurement of p53 in cell lysates
CS0050 Retinoblastoma ELISA, Human Apoptosis/Cell Cycle ELISA 96 wells <0.1 ng/mL h Quantitative ELISA for measurement of Rb in cell lysates
CS0450 Src ELISA, Human Protein Phosphorylation ELISA 96 wells <1.0 ng/mL h/m/r Quantitative ELISA for measurement of Src in human cell lysates
CS0470 STAT1 ELISA Gene Regulation ELISA 96 wells <0.27 ng/mL h Quantitative ELISA for measurement of STAT1 in cell lysates
CS0680 sTNF-RI ELISA Cytokines/Growth Factors/Hormones ELISA 96 wells 0.05 ng/mL h Quantitative amplified ELISA for measurement of soluble TNFRI in biological fluids
CS0690 sTNF-RII (75kDa) ELISA Cytokines/Growth Factors/Hormones ELISA 96 wells 0.1 ng/mL h Quantitative amplified ELISA for measurement of soluble TNFRII in biological fluids
CS0430 Tau ELISA, Mouse Cytoskeleton/Extracellular Matrix ELISA 96 wells <15 pg/mL m Quantitative ELISA for measurement of Tau in mouse cell lysates
CS0510 TRAIL ELISA, Human Apoptosis/Cell Cycle ELISA 96 wells <20 pg/mL h Quantitative ELISA for measurement of TRAIL in cell lysates
CS0120 Akt/PKB [pSer473] Phosphospecific ELISA Protein Phosphorylation ELISA Phosphospecific 96 wells <0.8 units/mL h/m/r Quantitative ELISA for measurement of Akt/PKB phosphorylated at [pSer473] in cell lysates
CS0110 Akt/PKB [pThr308] Phosphospecific ELISA Protein Phosphorylation ELISA Phosphospecific 96 wells <0.8 units/mL h/m/r Quantitative ELISA for measurement of Akt/PKB phosphorylated at [pThr308] in cell lysates
CS0550 ATF2 [pThr69/pThr71] Phosphospecific ELISA Gene Regulation ELISA Phosphospecific 96 wells <0.8 units/mL h/m/r Quantitative ELISA for measurement of ATF2 phosphorylated at [pThr69/pThr71] in cell lysates
CS0530 Bcl-2 [pSer70] Phosphospecific ELISA Apoptosis/Cell Cycle ELISA Phosphospecific 96 wells <0.5 units/mL h Quantitative ELISA for measurement of Bcl-2 phosphorylated at [pSer70] in cell lysates
CS0590 c-Met [pTyr1230/pTyr1234/pTyr1235] Phosphospecific ELISA Protein Phosphorylation ELISA Phosphospecific 96 wells <0.25 units/mL h Quantitative ELISA for measurement of c-Met phosphorylated at [pTyr1230/pTyr1234/pTyr1235] in cell lysates
CS0570 CREB [pSer133] Phosphospecific ELISA Gene Regulation ELISA Phosphospecific 96 wells <0.9 units/mL h/m Quantitative ELISA for measurement of CREB phosphorylated at [pSer133] in cell lysates
CS0150 Epidermal Growth Factor Receptor (EGFR) [pTyr1068] Phosphospecific ELISA, Human Cytokines/Growth Factors/Hormones ELISA Phosphospecific 96 wells <0.3 units/mL h Quantitative ELISA for measurement of EGFR phosphorylated on [pTyr1068] in cell lysates
CS0140 Epidermal Growth Factor Receptor (EGFR) [pTyr1173] Phosphospecific ELISA, Human Cytokines/Growth Factors/Hormones ELISA Phosphospecific 96 wells <0.3 units/mL h Quantitative ELISA for measurement of EGFR phosphorylated on [pTyr1173] in cell lysates
PE0100 ERK 1&2 [pThr185/pTyr187] Phosphospecific ELISA Protein Phosphorylation ELISA Phosphospecific 96 wells <0.8 units/mL h/m/r Quantitative ELISA for measurement of ERK 1&2 phosphorylated at [pThr185/pTyr187] in cell lysates
CS0630 HSP27 [pSer82] Phosphospecific ELISA Human Nitric Oxide/Cell Stress ELISA Phosphospecific 96 wells <1.0 units/mL h Quantitative ELISA for measurement of HSP27 phosphorylated at [pSer82] in human cell lysates
CS0660 IkBa [pSer32] Phosphospecific ELISA Gene Regulation ELISA Phosphospecific 96 wells <0.5 units/mL h Quantitative ELISA for measurement of IsymbolBa phosphorylated at [pSer32] in cell lysates
PI0100 Insulin Receptor [pTyr1158] Phosphospecific ELISA Cytokines/Growth Factors/Hormones ELISA Phosphospecific 96 wells <0.8 units/mL h/m/r Quantitative ELISA for measurement of IR phosphorylated at [pTyr1158] in cell lysates
PI0200 Insulin Receptor [pTyr1162/pTyr1163] Phosphospecific ELISA Cytokines/Growth Factors/Hormones ELISA Phosphospecific 96 wells <0.8 units/mL h/m/r Quantitative ELISA for measurement of IR phosphorylated at [pTyr1162/pTyr1163] in cell lysates
CS0130 JNK 1&2 [pThr183/pTyr185] Phosphospecific ELISA Protein Phosphorylation ELISA Phosphospecific 96 wells <0.8 units/mL h/m Quantitative ELISA for measurement of JNK 1&2 phosphorylated at [pThr183/pTyr185] in cell lysates
CS0650 MEK1 [pSer218/pSer222] Phosphospecific ELISA Protein Phosphorylation ELISA Phosphospecific 96 wells <1.0 units/mL h/m Quantitative ELISA for measurement of MEK1 phosphorylated at [pSer218/pSer222] in cell lysates
CS0020 p38 MAPK [pThr180/pTyr182] Phosphospecific ELISA Protein Phosphorylation ELISA Phosphospecific 96 wells <0.8 units/mL h/m/r Quantitative ELISA for measurement of p38 MAPK phosphorylated at [pThr180/pTyr182] in cell lysates
CS0040 p53 [pSer15] Phosphospecific ELISA, Human Apoptosis/Cell Cycle ELISA Phosphospecific 96 wells <0.9 units/mL h Quantitative ELISA for measurement of p53 phosphorylated at [pSer15] in cell lysates
CS0060 Retinoblastoma [pSer249/pThr252] Phosphospecific ELISA, Human Apoptosis/Cell Cycle ELISA Phosphospecific 96 wells <0.8 units/mL h Quantitative ELISA for measurement of Rb phosphorylated at [pSer249/pThr252] in cell lysates
CS0420 Retinoblastoma [pThr821] Phosphospecific ELISA, Human Apoptosis/Cell Cycle ELISA Phosphospecific 96 wells <0.5 units/mL h Quantitative ELISA for measurement of Rb phosphorylated at [pThr821] in cell lysates
CS0460 Src [pTyr418] Phosphospecific ELISA Protein Phosphorylation ELISA Phosphospecific 96 wells <0.1 units/mL h/m/r Quantitative ELISA for measurement of Src phosphorylated at [pTyr418] in mouse cell lysates
CS0480 STAT1 [pThr701] Phosphospecific ELISA Gene Regulation ELISA Phosphospecific 96 wells <0.1 units/mL h Quantitative ELISA for measurement of STAT1 phosphorylated at [pThr701] in cell lysates
CS0440 Tau [pSer199] Phosphospecific ELISA, Mouse Cytoskeleton/Extracellular Matrix ELISA Phosphospecific 96 wells <2 pg/mL m Quantitative ELISA for measurement of Tau phosphorylated at [pSer199] in mouse cell lysates
CS0740 Acid Phosphatase Assay Kit Nitric Oxide/Cell Stress Enzymatic 1,000 in microwell plate 0.2 mUnit mammalian Quantitative Acid Phosphatase activity measurement in tissues, whole cell extracts, column fractions and purified enzyme. Acid phosphatase can serve as a lysosomal marker
CS0010 BACE1 Activity Assay Kit (FRET) Neurobiology Enzymatic 250 reactions in microwells 10 ng purified enzyme/test h/r (tested) Quantitative b-secretase activity measurement and inhibitor screening by a fluorescence based assay
CS0400 Casein Kinase Assay Kit - General Protein Phosphorylation Enzymatic 100 tests 10 ng CKI and 0.06 units CKII mammalian Enzymatic assay of different casein kinases by an in vitro phosphorylation of a-casein and radioactive detection in cell lysates
CS0600 Casein Kinase I Assay Kit Protein Phosphorylation Enzymatic 50 tests 10 ng purified enzyme mammalian Enzymatic activity of casein kinase I (CK-1) assayed by radioactive detection of the phosphorylated peptide substrate
CS0610 Casein Kinase II Assay Kit Protein Phosphorylation Enzymatic 70 tests 0.02 units mammalian Enzymatic activity of casein kinase II (CK-2) assayed by radioactive detection of the phosphorylated peptide substrate
CASP-3-C Caspase 3 Assay Kit, Colorimetric Apoptosis/Cell Cycle Enzymatic 800 tests in 96 plate Linear between 1-20 nmole/well mammalian Quantitative colorimetric assay of Caspase 3 activity detected by the release of p-nitroaniline (pNA) from the Caspase-3 substrate
CASP-3-F Caspase 3 Assay Kit, Fluorimetric Apoptosis/Cell Cycle Enzymatic 1,000 in microwell plate 0.1-1.2 nmol in microwell or 0.05-0.5 nmol in 2ml. mammalian Quantitative fluorimetric assay of Caspase 3 activity detected by the release of 7-amino-4-methylcoumarin (AMC) from the Caspase-3 substrate
CASP-6-F Caspase 6 Assay Kit, Fluorimetric Apoptosis/Cell Cycle Enzymatic 1,000 in microwell plate 0.2-2 nmol mammalian Quantitative fluorimetric assay of Caspase 6 activity detected by the release of 7-amino-4-methylcoumarin (AMC) from the Caspase-6 substrate
CASP-8-C Caspase 8 Assay Kit, Colorimetric Apoptosis/Cell Cycle Enzymatic 100 tests of 100 ml 0.003-0.0025 units/test mammalian Quantitative colorimetric assay of Caspase 8 activity detected by the release of p-nitroaniline (pNA) from the Caspase-8 substrate
CASP-8-F Caspase 8 Assay Kit, Fluorimetric Apoptosis/Cell Cycle Enzymatic 1,000 in microwell plate Linear 0.1-1 nmole standard/well mammalian Quantitative fluorimetric assay of Caspase 8 activity detected by the release of 7-amino-4-methylcoumarin (AMC) from the Caspase-8 substrate
CAT-100 Catalase Assay Kit Nitric Oxide/Cell Stress Enzymatic 100 tests/mL 0.01-0.08 units/min UV 0.3-1.75 UV units/ 5 min mammalian Quantitative enzymatic assay of catalase activity in tissue lysates. Can serve as a peroxisomal marker
CS0720 Citrate Synthase Assay Kit Apoptosis/Cell Cycle Enzymatic 100 test/1mL 480 tests/well 20 ng/ml purified enzyme mammalian Colorimetric assay of citrate synthase activity, a mitochondrial matrix marker
CYTOC-OX1 Cytochrome c Oxidase Assay Kit Apoptosis/Cell Cycle Enzymatic 100 tests/mL 0.4-4 milliunits/min mammalian/yeast Colorimetric assay for Cytochrome c oxidase activity. Can serve as a mitochondrial protein marker
CY0100 Cytochrome c Reductase [NADPH] Assay Kit Multi-Drug Resistance Enzymatic 100 assays/mL 0.8-4.1 milliunits/min mammalian/yeast Quantitative enzymatic assay of NADPH-cytochrome c reductase in cell and tissue extracts and in purified ER. Can serve as an ER marker
CYTO-1A Cytochrome P450 Fluorescent Detection Kit (P4501A1/2) Multi-Drug Resistance Enzymatic 7.5 g tissue or 50 assays 6-250 pmole/min mammalian Fluorescent assay of Cytochrome P450 in tissue extracts
CYTO-2B Cytochrome P450 Fluorescent Detection Kit (P4502B4) Multi-Drug Resistance Enzymatic 7.5 g tissue or 50 assays 6-250 pmole/min mammalian Fluorescent assay of Cytochrome P450 in tissue extracts
CS0340 Dihydrofolate Reductase Assay Kit Nitric Oxide/Cell Stress Enzymatic 50-100 tests/mL 500 ng purified enzyme mammalian Quantitative DHFR activity measurement and inhibitor screening by a spectrophotometric based assay
CS0260 Glutathione Assay Kit Nitric Oxide/Cell Stress Enzymatic 500 tests in microwell     Quantative measurment of total glutathione (reduced+disulfide) levels in biological samples
CGP-1 Glutathione Peroxidase Cellular Activity Assay Kit Nitric Oxide/Cell Stress Enzymatic 100 tests 0.005-0.1 units/3 min mammalian Quantitative enzymatic assay of gluthathione peroxidase (GPx) in tissue lysates
GR-SA Glutathione Reductase Assay Kit Nitric Oxide/Cell Stress Enzymatic 150 tests Colorimetric assay: 0.003-0.03 units; UV assay: 0.003-0.012 units mammalian/yeast Quantitative enzymatic assay of glutathione reductase in tissue lysates
CS0410 Glutathione S-Transferase (GST) Assay Kit Nitric Oxide/Cell Stress Enzymatic 500 tests in microwells 60 ng purified enzyme/test mammalian/bacteria Quantitative enzymatic assay of GST activity in bacteria, mammalian cells and tissue lysates
CS0270 Hydrogen Peroxide Colorimetric Assay Nitric Oxide/Cell Stress Enzymatic 96 wells 51.3 ng/mL   Quantitative H2O2 determination in serum, plasma, urine and cell culture supernatants
MD0100 MGMT Assay Kit Nitric Oxide/Cell Stress Enzymatic 100-200 tests 0.002-0.02 mmol/hr mammalian Radioactive detection of MGMT (DNA repair protein) activity and inhibition in cell lysates
NOS-1 Nitric Oxide Synthase (NOS) Detection Kit, Isotopic Nitric Oxide/Cell Stress Enzymatic 50 assays   h/r Quantitative enzymatic assay of nitric oxidase synthase (NOS) in cell lysates
FCANOS-1 Nitric Oxide Synthase Detection System, Fluorometric Nitric Oxide/Cell Stress Enzymatic 96 wells   h/m Quantitative enzymatic fluorimetric detection of nitric oxide (NO) in living cells
CS0710 Ogg1 Assay Kit Nitric Oxide/Cell Stress Enzymatic 180 assays 10 mg/ml purified enzyme h (tested) Radioactive detection of Ogg1 enzyme activity (DNA repair) in cell lysates
PD-1 PeroxiDetect Kit Nitric Oxide/Cell Stress Enzymatic 100 tests for aqueous or organic peroxides H2O2 1-7 nmoles/ test mammalian Quantitative enzymatic assay of hydrogen peroxide in aqueous solutions of cell lysates
CS0170 Thioredoxin Reductase Assay Kit Nitric Oxide/Cell Stress Enzymatic 100 tests 0.05-0.5 nmoles per well mammalian Quantitative enzymatic assay of mammalian thioredoxin reductase
PTP-101 Protein Tyrosine Phosphatase Assay Kit, Non-Radioactive Protein Phosphorylation Enzymatic 96 wells 400-500 pmol (PO4--)
all species The enzymatic reaction and detection of the inorganic phosphate released from PTPase
APO-AF Annexin V-FITC Apoptosis Detection Kit Apoptosis/Cell Cycle Flow Cytometry 20 or 50 tests NA mammalian Assay of Annexin V binding as an early apoptosis marker by flow cytometry
APO-BRDU Apoptosis Flow Cytometry Kit Apoptosis/Cell Cycle Flow Cytometry 60 cell susp.   mammalian Immunohistochemical labeling of Br-dUTP in apoptotic cells and flow cytometry detection
APO-AC Annexin V-Cy3 Apoptosis Detection Kit Apoptosis/Cell Cycle Fluorescence 200 tests NA mammalian Fluorescent detection of Annexin V binding as an early apoptosis marker
CS0280 Caspase 1 FAM-FLICA Assay Apoptosis/Cell Cycle Histochemistry 100 tests NA mammalian Fluorometric or flow cytometry detection of active cellular caspase 1 using Fluorochrome Inhibitor of Caspase 1 activity (FLICA)
CS0290 Caspase 2 FAM-FLICA Assay Apoptosis/Cell Cycle Histochemistry 100 tests NA mammalian Fluorometric or flow cytometry detection of active cellular caspase 2 using Fluorochrome Inhibitor of Caspase 2 activity (FLICA)
CS0300 Caspase 9 FAM-FLICA Assay Apoptosis/Cell Cycle Histochemistry 100 tests NA mammalian Fluorometric or flow cytometry detection of active cellular caspase 9 using Fluorochrome Inhibitor of Caspase 9 activity (FLICA)
CS0310 Caspase 10 FAM-FLICA Assay Apoptosis/Cell Cycle Histochemistry 100 tests NA mammalian Fluorometric or flow cytometry detection of active cellular caspase 10 using Fluorochrome Inhibitor of Caspase 10 activity (FLICA)
CS0320 Caspase 13 FAM-FLICA Assay Apoptosis/Cell Cycle Histochemistry 100 tests NA mammalian Fluorometric or flow cytometry detection of active cellular caspase 13 using Fluorochrome Inhibitor of Caspase 13 activity (FLICA)
CS0370 Cathepsin B Detection Assay Apoptosis/Cell Cycle Histochemistry 100 assays NA h Fluorescence detection of cathepsin B by protease activity in living cells with cresyl violet fluorophore
CS0360 Cathepsin K Detection Assay Apoptosis/Cell Cycle Histochemistry 100 assays NA h Fluorescence detection of cathepsin K by protease activity in living cells with cresyl violet fluorophore
CS0350 Cathepsin L Detection Assay Apoptosis/Cell Cycle Histochemistry 100 assays NA h Fluorescence detection of cathepsin L by protease activity in living cells with cresyl violet fluorophore
CS0390 Mitochondria Staining Kit Apoptosis/Cell Cycle Histochemistry 200 x 1 ml tests NA mammalian Detection of changes in electrochemical potential of mitochondrial membrane by staining cells with JC-1 dye and fluorometric detection
CS0030 Senescent Cells Staining Kit Apoptosis/Cell Cycle Histochemistry 100 tests NA mammalian Histochemical staining of b-galactosidase activity in senescent cells
CS0250 p38 MAPK Activity Assay Kit Protein Phosphorylation Immuno-precipitation 50 reactions NA mammalian Enzymatic activity of p38 immunoprecipitated from cell lysates, assayed by immunoblotting of the phosphorylated p38 substrate ATF2
CS0490 MEK1 Assay Kit Protein Phosphorylation Immuno-precipitation 50 reactions NA mammalian Enzymatic activity of MEK1 immunoprecipitated from cell lysates, assayed by immunoblotting of the phosphorylated MEK1 substrate ERK2
CS0380 JNK Activity Assay Kit Protein Phosphorylation Immuno-precipitation 50 reactions NA mammalian Enzymatic activity of JNK immunoprecipitated from cell lysates, assayed by immunoblotting of the phosphorylated JNK substrate ATF2
back to top
back to Cell Signaling