Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Cell Biology > Learning Center > Pathway Slides & Charts  > 6TM Potassium Channel
Cell Signaling & Neuroscience
6TM Potassium Channel
Top Border
Left Border  Right Border
Left Corner Back Download Forward Sigma-Aldrich Right Corner
Figure-3b
6TM Potassium Channel Structure
  1. The α subunit is formed from 6 transmembrane segments and is associated with a regulatory β subunit.
  2. Four α subunits form the pore.
Back to Top