Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Cell Biology > Learning Center > Pathway Slides & Charts  > Activation Inhibition
Cell Signaling & Neuroscience
Activation Inhibition
Top Border
Left Border  Right Border
Left Corner Back Download Forward Sigma-Aldrich Right Corner
Figure-3b
Activation and Inhibition of Apoptosis
Several mechanisms have been identified in mammalian cells for the induction of apoptosis. These mechanisms include factors that lead to perturbation of the mitochondria leading to leakage of cytochrome c or factors that directly activate members of the death receptor family. Fas is a member of the tumor necrosis factor (TNF) receptor superfamily, a family of transmembrane receptors that include neurotrophin receptor (p75NTR), TNF-R1, and a variety of other cell surface receptors. Fas Ligand (Fas L) transmits signals to Fas on a target cell by inducing trimerization of Fas. Activation of Fas causes the recruitment of Fas-associated protein with death domain (FADD) via interactions between the death domain of Fas and FADD and is followed by pro-caspase-8 binding to FADD via interactions between the death effector domains (DED) of FADD and pro-caspase-8 leading to the activation of caspase-8. Activation of caspase-8 leads to the activation of other caspases, in effect beginning a caspase cascade that ultimately leads to apoptosis. Caspase-8 activation can also activate Bid, leading to activation of the apoptotic program. Fas-induced apoptosis can be effectively blocked at several stages by either FLICE-inhibitory protein (FLIP), by Bcl-2, or by the cytokine response modifier A (CrmA). In addition, activation of caspase-3 by caspase-9 can be blocked by inhibitor of apoptosis proteins (IAPs). Moreover, the protein kinase, Akt, can be activated by various growth factors and its activity can be blocked by PTEN. Akt functions to promote cell survival through two distinct pathways. Akt inhibits apoptosis by phosphorylating the Bcl-2 family member Bad, which then interacts with 14-3-3 and dissociates from Bcl-xL allowing for cell survival. Alternatively, Akt activates IKKα that ultimately leads to NFκB activation and cell survival. Proapoptotic Bcl-2 family members, such as Bax and Bak can promote mitochondrial permeability, while Bcl-2 can inhibit their effects. Upon mitochondrial permeability, apoptogenic factors are released from the mitochondrial inter-membrane space and leak into the cytosol. One factor is cytochrome c, which induces the liberation of protease activators (caspases) that ultimately lead to apoptosis through nuclear damage (DNA fragmentation, DNA mutations). In addition, Smac/Diablo is released and can block IAP inhibition of capsase activity. Mitochondrial permeability is also related to the increased generation of reactive oxygen species (ROS), which plays a role in the degradation phase of apoptosis (i.e. plasma membrane alterations).
Back to Top