Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Cell Signaling & Neuroscience
Aquaporin Channel
Top Border
Left Border  Right Border
Left Corner Back Download Forward Sigma-Aldrich Right Corner
Figure-3b
Aquaporin Channel Structure
  1. Aquaporins consist of six membrane-spanning segments arranged in two hemipores, which fold together to form the hourglass-shaped channel
  2. The highly conserved NPA motifs are also shown and may form a size-exclusion pore, giving the channel its high sensitivity
Back to Top