Sigma-Aldrich
Life Science > Cell Biology > Learning Center > Pathway Slides & Charts  > ATM/p53 Signaling Pathway
Cell Signaling & Neuroscience
ATM/p53 Signaling Pathway
Top Border
Left Border  Right Border
Left Corner Back Download Forward Sigma-Aldrich Right Corner
Figure-3b
ATM/p53 Signaling Pathway
The ataxia telangiectasia-mutated gene (ATM) encodes a protein kinase that acts as a tumor suppressor. ATM activation, via IR damage to DNA, stimulates DNA repair and blocks cell cycle progression. One mechanism through which this occurs is ATM dependent phosphorylation of p53. p53 can cause growth arrest of the cell at a checkpoint to allow for DNA damage repair or can cause the cell to undergo apoptosis if the damage cannot be repaired. The critical role of p53 is evident by the fact that it is mutated in over 50% of all human cancers.