Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Cell Biology > Learning Center > Pathway Slides & Charts  > Bone Morphogenic Protein
Cell Signaling & Neuroscience
Bone Morphogenic Protein
Top Border
Left Border  Right Border
Left Corner Back Download Forward Sigma-Aldrich Right Corner
Figure-3b
Bone Morphogenic Protein
Bone morphogenetic protein (BMP) induces ectopic bone formation, and plays an important role in the development of the viscera. Ligand binding to its receptor induces the formation of a complex in which the Type II BMP receptor phosphorylates and activates the Type I BMP receptor. The Type I BMP receptor then propagates the signal by phosphorylating a family of signal transducers, the Smad proteins. Currently, eight Smad proteins have been cloned (Smad 1-7 and Smad 9). Upon phosphorylation by the BMP Type I receptor, Smad1 can interact with either Smad4 or Smad6. The Smad1-Smad6 complex is inactive; however, the Smad1-Smad4 complex triggers the expression of BMP responsive genes. The ratio between Smad4 and Smad6 in the cell can modulate the strength of the signal transduced by BMP.
Back to Top
References:
Fujii, M., et al., Roles of bone morphogenetic protein type I receptors and Smad proteins in osteoblast and chondroblast differentiation. Mol. Biol. Cell., 10, 3801-3813 (1999).
Kawabata, M., et al., Signal transduction by bone morphogenetic proteins. Cytokine Growth Factor Rev., 9, 49-61 (1998).