Sigma-Aldrich
Life Science > Cell Biology > Learning Center > Pathway Slides & Charts  > Cell Signaling in Lipids
Cell Signaling & Neuroscience
Cell Signaling in Lipids
Top Border
Left Border  Right Border
Left Corner Back Download Forward Sigma-Aldrich Right Corner
Figure-3b
Lipids in Cell Signaling
Many of the lipids involved as second messengers in cell signaling pathways arise from the arachidonic acid (AA) pathway. AA is an unsaturated fatty acid that is a normal constituent of membrane phospholipids and is released from the phospholipids by the actions of phospholipase A2 (PLA2). Prostaglandins (PG) arise from a cyclic endoperoxide generated by the enzyme system PG synthetase, a complex of enzymes that includes cyclooxygenase (COX). There is a constitutive (COX-1) and an inducible cyclooxygenase (COX-2). The cyclic endoperoxide intermediate is also a precursor of prostacyclin (PGI2) and thromboxane (TXA3). Other groups of compounds in this class, leukotrienes (LT) and lipoxins (LP), are derived directly from AA without the mediation of a cyclic endoperoxide. Lipoxygenase acts on AA to produce 5-hydroperoxyeicosatetraenoic acid (5-HPETE) that is converted to LTA4. LTA4 is the precursor of LTB4, that induces inflammation by its chemotactic and degranulating actions on polymorphonuclear lymphocytes (PML), and of LTC4, LTD4, and LTE4, the amino acid containing LTs that induce vasoconstriction and bronchoconstriction and are involved in asthma and anaphylaxis.
Back to Top
References:
Heller, A., et al., Lipid mediators in inflammatory disorders. Drugs, 55, 487-496(1998).
Cook, J.A., et al., Prostaglandins, thromboxanes, leukotrienes, and cytochrome P-450 metabolites of arachidonic acid. New Horiz., 1, 60-69(1993).
Dubois, R.N., et al., Cyclooxygenase in biology and disease. FASEB J., 12, 1063-1073(1998).
ORDERING
SERVICE & SUPPORT
CORPORATE
    Follow us on Twitter!  Join us on Facebook!  Join us on LinkedIn!  Watch our videos on YouTube!
Copyrights © 2010 Sigma-Aldrich Co. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.