Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Cell Signaling & Neuroscience
Chloride Channel
Top Border
Left Border  Right Border
Left Corner Back Download Forward Sigma-Aldrich Right Corner
Figure-3b
Schematic Representation of the Chloride (CIC) Channel
Each CIC subunit consists of 18 membrane-associated helices (not shown) and contributes a single pore to a dimeric "double-barreled" channel that contains two independently gated pores. These pores can be closed by a common gate.
Back to Top