Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Cell Biology > Learning Center > Pathway Slides & Charts  > Early Response Events
Cell Signaling & Neuroscience
Early Response Events
Top Border
Left Border  Right Border
Left Corner Back Download Forward Sigma-Aldrich Right Corner
Figure-3b
Early Response Events to Moderate Double Strand Break (DSB) Levels
The MRE11-RAD50-NBS1 (MRN) complex is essential for the initial damage processing of DSBs. Processed DSBs leads to the recruitment and subsequent activation of ATM, which then can phosphorylate its substrates, including MRE11 and NBS1. Phosphorylated NBS1 plays a role in checkpoint activation and can also facilitate the phosphorylation of certain ATM effectors.
Back to Top