|
Cell Signaling & Neuroscience
G1 & S Phases
|
|
|
G 1 and S Phases of the Cell Cycle
In proliferating cells, the cell cycle consists of four phases. Gap 1 (G1) is the interval between mitosis and DNA replication that is characterized by cell growth. The transition that occurs at the restriction point (R) in G1 commits the cell to the proliferative cycle. If the conditions that signal this transition are not present, the cell exits the cell cycle and enters G0, a nonproliferative phase during which growth, differentiation and apoptosis occur. Replication of DNA occurs during the synthesis (S) phase, which is followed by a second gap phase (G2) during which growth and preparation for cell division occurs. Mitosis and the production of two daughter cells occur in M phase.
Passage through the four phases of the cell cycle is regulated by a family of cyclins that act as regulatory subunits for cyclin-dependent kinases (cdks). The activity of the various cyclin/cdk complexes that regulate the progression through G1-S-G2 phases of the cell cycle is controlled by the synthesis of the appropriate cyclins during a specific phase of the cell cycle. The cyclin/cdk complex is then activated by the sequential phosphorylation and dephosphorylation of the key residues of the complex, located principally on the cdk subunits.
The cyclin cdk complex of early G1 is either cdk2, cdk4, or cdk6 bound to a cyclin D isoform. There are several proteins that can inhibit the cell cycle in G1. If DNA damage has occurred, p53 accumulates in the cell and induces the p21-mediated inhibition of cyclin D/cdk. Mdm2, by facilitating the nuclear export/inactivation of p53, becomes part of an inhibitory feedback loop that inactivates p21-mediated G1 arrest. Similarly, activation of TGF-b receptors induces the inhibition of cyclin D/cdk by p15, while cyclic AMP inhibits the cyclin D/cdk complex via p27. If the cyclin D/cdk complex is inhibited, retinoblastoma protein (Rb) is in a state of low phosphorylation and is tightly bound to the transcription factor E2F, inhibiting its activity.
Passage through the restriction point and transition to S phase is triggered by the activation of the cyclin D/cdk complex, which phosphorylates Rb. Phosphorylated Rb dissociates from E2F, which is then free to initiate DNA replication. Cyclin E/cdk2 accumulates during late G phase and triggers the passage into S phase. The entire genome is replicated during S phase. The synthesis and accumulation of cyclin B/cdc2 also begins during S phase, but the complex is phosphorylated at Thr14 -Tyr15 and is inactive. Cyclin A/cdk2 accumulates during S phase and its activation triggers the transition to G2, a phase characterized by the accumulation of cyclin B/cdc2, the inhibition of DNA replication, cell growth and new protein synthesis.
Back to Top
References
Nakayama, K.I., et al., Regulation of the cell cycle at the G1-S transition by proteolysis of cyclin E and p27Kip1. Biochem. Biophys. Res. Commun., 282, 853-860 (2001).
Bartek, J., and Lukas, J., Pathways governing G1/S transition and their response to DNA damage. FEBS Lett., 490, 117-122 (2001).
Harbour, J.W., and Dean, D.C., Rb function in cell-cycle regulation and apoptosis. Nat. Cell Biol., 2, E65-E67 (2000).
Momand, J., et al., MDM2--master regulator of the p53 tumor suppressor protein. Gene, 242, 15-29 (2000).