Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Cell Signaling & Neuroscience
GDNF Family
Top Border
Left Border  Right Border
Left Corner Back Download Forward Sigma-Aldrich Right Corner
Figure-3b
GDNF Family with Receptors
Primary receptor interactions are shown by solid line arrows. Weaker or secondary interactions are shown by dashed line arrows. Each of the four coreceptors can complex with Ret receptor tyrosine kinase (center), believed to be the primary transmembrane subunit for signal transduction. Ligand binding may also generate Ret-independent signaling via a Src kinase associated with lipid rafts in the cell membrane.
Back to Top