Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Cell Signaling & Neuroscience
Hormonal Targeting
Top Border
Left Border  Right Border
Left Corner Back Download Forward Sigma-Aldrich Right Corner
Figure-3b
Hormonal Targeting of Nuclear Complexes to Chromatin
In the absence of ligand, a silencing mediator of retinoid and thyroid hormone receptor (SMRT)-mSin3a-histone deacetylase-1 (HDAC-1) complex associates with the nuclear retinoic acid receptor (RAR)-retinoid X receptor (RXR) heterodimeric complex. The HDAC activity of the complex maintains the tight association between deacetylated histones and chromatin that results in repressed gene transcription. When 9-cis retinoic acid binds to the RAR-RXR heterodimeric complex the corepressor complex dissociates from the receptor complex with simultaneous recruitment of a coactivator complex that includes CREB-binding protein (CBP), CBP-associated factor (p/CAF), and steroid receptor coactivator-1 (SRC-1). The receptor/coactivator complex activates histone acetyltransferase (HAT). Acetylated histone dissociates from the chromatin, allowing the preinitiation complex (PIC) to bind to DNA and the ligand heterodimeric RAR-RXR-coactivator complex to bind to specific DNA response elements (RARE) in the promoter regions of target genes to activate transcription.
Back to Top
References:
Li, H., et al., Characterization of receptor interaction and transcriptional repression by the corepressor SMRT. Mol. Endocrinol., 11, 2025-2037 (1997).
Blanco, J.C., et al., The histone acetylase PCAF is a nuclear receptor coactivator. Genes Dev., 12, 1638-1651 (1998).
Minucci, S., et al., Retinoid receptors in health and disease: co-regulators and the chromatin connection. Semin. Cell Dev. Biol., 10, 215-225.(1999).