Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Cell Biology > Learning Center > Pathway Slides & Charts  > Inductible Nitric Oxide (iNOS)
Cell Signaling & Neuroscience
Inductible Nitric Oxide (iNOS)
Top Border
Left Border  Right Border
Left Corner Back Download Forward Sigma-Aldrich Right Corner
Figure-3b
Inducible Nitric Oxide Synthase (iNOS)
Macrophages are important for early immune responses to invading microorganisms and the production of nitric oxide (NO) is central to this function. NO is generated by inducible nitric oxide synthase (iNOS, macNOS, Type II NOS) following exposure to certain cytokines, such as interferon-g (IFN-g). The IFN-g receptor signals through the Janus kinase (JAK) family and signal transducers and activators of transcription (STAT) proteins. Receptor occupation and dimerization induces the phosphorylation of associated STATs. Activated STATs dimerize and translocate to the nucleus where they increase expression of the transcription factor, IRF-1, that, in turn binds to specific DNA elements in the iNOS gene promoter region to increase iNOS gene expression. iNOS is a soluble enzyme that, unlike eNOS and nNOS, does not require elevated intracellular Ca2+ levels for activation.
Back to Top
References:
Massa, P.T., et al., Increased inducible activation of NF-kappaB and responsive genes in astrocytes deficient in the protein tyrosine phosphatase SHP-1. J. Interferon Cytokine Res., 18, 499-507 (1998).
Fehr, T., et al., Crucial role of interferon consensus sequence binding protein, but neither of interferon regulatory factor 1 nor of nitric oxide synthesis for, protection against murine listeriosis. J. Exp. Med., 185, 921-931 (1997).