Sigma-Aldrich
Life Science > Cell Biology > Learning Center > Pathway Slides & Charts  > Inositol Phosphates
Cell Signaling & Neuroscience
Inositol Phosphates
Top Border
Left Border  Right Border
Left Corner Back Download Forward Sigma-Aldrich Right Corner
Figure-3b
Inositol Phosphates
Inositol phosphates are important intracellular second messengers in eukaryotic cells. Inositol, a cyclic molecule with six hydroxyl groups, forms the hydrophobic head group of membrane inositol phospholipids. The actions of lipid kinases and phosphatases on the inositol ring of inositol phospholipids generate a variety of phosphoinositol phospholipids. As a result of upstream signaling events, specific phospholipases are activated, cleave phosphoinositol phospholipids, and release intracellular inositol phosphate second messenger molecules. The most studied inositol phosphate second messenger is inositol 1,4,5-trisphosphate (IP3). IP3 is released from phosphatidylinositol 4,5-bisphosphate (PIP2) by the action of phospholipase C-b and is responsible for the release of calcium ions from intracellular stores in the endoplasmic reticulum.
Back to Top
References:
Irvine, R.F., and Schell, M.J., Back in the water: the return of inositol phosphates. Nat. Rev. Mol. Cell Biol., 2, 327-338 (2001).
Shears, S.B., Assessing the omnipotence of inositol hexakisphosphate. Cell. Signal., 13, 151-158 (2001).