Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Cell Signaling & Neuroscience
HCN Channel
Top Border
Left Border  Right Border
Left Corner Back Download Forward Sigma-Aldrich Right Corner
Figure-3b
Schematic Representation of the HCN Channel
The HCN channel consists of six transmembrane helices with a positively charged voltage sensor, a pore forming sequence and a cyclic nucleotide binding domain (CNBD) in the C-terminus which mediates the response to cAMP.
Back to Top