Sigma-Aldrich
Life Science > Cell Biology > Learning Center > Pathway Slides & Charts  > Ligand-Gated Ion Channel
Cell Signaling & Neuroscience
Ligand-Gated Ion Channel
Top Border
Left Border  Right Border
Left Corner Back Download Forward Sigma-Aldrich Right Corner
Figure-3b
Ligand-Gated Ion Channel: Structure and Activation States
  1. The intrinsic ion channel in this superfamily of receptors undergoes a conformational change following the binding of an extracellular ligand to its binding site, which allows for the opening of the channel pore.
  2. The pore is formed by a pentameric arrangement of subunits, which are in turn composed of a large extracellular domain (which contains the ligand binding site) and four transmembrane domains.