Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Cell Biology > Learning Center > Pathway Slides & Charts  > Limbic Reward Circuit
Cell Signaling & Neuroscience
Limbic Reward Circuit
Top Border
Left Border  Right Border
Left Corner Back Download Forward Sigma-Aldrich Right Corner
Figure-3b
Limbic Reward Circuit
This diagram represents a sagittal section of the rat brain. Highlighted are the nuclei representing the limbic structures of the basal forebrain including the amygdala, hippocampus, prefrontal cortex (PFC), nucleus accumbens (N. Acc.), ventral pallidum (VP) and ventral tegmental area (VTA). Dopaminergic neurons in the VTA modulate information flow through the limbic circuit via projections to the nucleus accumbens, amygdala, hippocampus, PFC and VP. Increased dopaminergic transmission in limbic nuclei, particularly the nucleus accumbens, underlies the reinforcing effect of virtually every abused drug. Note that psychostimulants increase dopaminergic transmission in areas receiving projections from the VTA via interactions with dopamine transporters.
Back to Top