Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Cell Signaling & Neuroscience
MRP1
Top Border
Left Border  Right Border
Left Corner Back Download Forward Sigma-Aldrich Right Corner
Figure-3b
Domain Organization of Multi-Drug Related Protein (MRP1)
MRP1 is a representative of the second major subfamily, MRP, of multidrug ABC transporters. MRPs have an extra TMD and have the general form of TMD0(ABC-TMD)2. The diagram shows the transporter as an intrinsic membrane protein.
Back to Top
References
Kruh, G.D., et al. MRP subfamily transporters and resistance to anticancer agents. J. Bioenerg. Biomembr., 33, 493-501 (2001).
Borst, P., et al. A family of drug transporters: the multidrug resistance-associated proteins. J. Natl. Cenacer Inst., 92, 1295-1302 (2002).
Rosenberg, M.F., et al. The structure of the multidrug resistance protein 1 (MRP1/ABCC1), crystallization and single-particle analysis. J. Biol. Chem., 276, 16076-16082 (2001).