Sigma-Aldrich
Life Science > Cell Biology > Learning Center > Pathway Slides & Charts  > Neuronal (nNOS)
Cell Signaling & Neuroscience
Neuronal (nNOS)
Top Border
Left Border  Right Border
Left Corner Back Download Forward Sigma-Aldrich Right Corner
Figure-3b
Neuronal Nitric Oxide Synthase (nNOS)
Three isoforms of nitric oxide synthase (NOS) have been identified. All are homodimers with subunits of 130-160 kDa. All have binding sites for NADPH, FAD, and FMN near the carboxyl terminus (the reductase domain), and binding sites for tetrahydrobiopterin (BH4) and heme near the amino terminus (the oxygenase domain). The reductase and oxygenase domains are linked by a calmodulin (CaM) binding site. Occupation of this site facilitates electron transfer from the cofactors in the reductase domain to heme during nitric oxide production. NOS catalyzes the conversion of arginine to citrulline and nitric oxide (NO). Neuronal nitric oxide synthase (nNOS, bNOS, cNOS, Type I) is associated with the post-synaptic density protein (PSD-95) in the neuronal membrane. In response to increased intracellular Ca2+, nNOS interacts with CaM. The Ca2+-CaM complex, in combination with BH4, binds to nNOS and induces its translocation from the plasma membrane to the cytoplasm. The dephosphorylation of nNOS by calcineurin initiates the production NO. NO activates guanylyl cyclase (GC) and activates the various cGMP-regulated signaling pathways. nNOS is in activated by phosphorylation by protein kinase A (PKA) or protein kinase C (PKC).
Back to Top
References:
Dawson, T.M., et al., Regulation of neuronal nitric oxide synthase and identification of novel nitric oxide signaling pathways. Prog. Brain Res., 118, 3-11 (1998).
Wang, Y., et al., Nitric oxide synthases: gene structure and regulation. Adv. Pharmacol., 34, 71-90 (1995).