Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Cell Biology > Learning Center > Pathway Slides & Charts  > Neurotrophins & Receptors
Cell Signaling & Neuroscience
Neurotrophins & Receptors
Top Border
Left Border  Right Border
Left Corner Back Download Forward Sigma-Aldrich Right Corner
Figure-3b
Neurotrophins and Their Receptors
Primary receptor interactions are shown by solid line arrows. Weaker or secondary interactions are shown by dashed line arrows. All four bind to the low affinity receptor (p75NTR), which does not have obvious signaling capability.
Back to Top