Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Cell Biology > Learning Center > Pathway Slides & Charts  > Phosatidic Acid Synthesis
Cell Signaling & Neuroscience
Phosatidic Acid Synthesis
Top Border
Left Border  Right Border
Left Corner Back Download Forward Sigma-Aldrich Right Corner
Figure-3b
Phosphatidic Acid Synthesis
The endoplasmic reticulum (ER) produces most of the lipids required for the generation of new cell membranes. These lipids include cholesterol as well as the phospholipid phosphatidylcholine (PC) and phosphatidic acid (PA). PA is produced by the cleavage of PC to generate PA by the enzyme phospholipase D1 (PLD1). PLD1 activation is induced by agonist binding to G protein-coupled receptors (GPCRs) or receptor tyrosine kinases (RTKs) and is mediated by protein kinase C (PKC) as well as by the small GTP binding proteins Rho (RhoA, Rac1 and Cdc42) and ADP-ribosylation factor1 (ARF1).
Back to Top
References:
Exton, J.H., Phospholipase D. Ann. N.Y. Acad. Sci., 905, 61-68 (2000).
McPhail, L.C., et al., A novel protein kinase target for the lipid second messenger phosphatidic acid. Biochim. Biophys. Acta., 1439, 277-290 (1999).
Daniel, L.W., et al., hospholipase D, tumor promoters, proliferation and prostaglandins. Biochim. Biophys. Acta., 1439, 265-276 (1999).