Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Cell Biology > Learning Center > Pathway Slides & Charts  > Phosphorylation Driven
Cell Signaling & Neuroscience
Phosphorylation Driven
Top Border
Left Border  Right Border
Left Corner Back Download Forward Sigma-Aldrich Right Corner
Figure-3b
Phosphorylation Driven Regulatory Switch from Transcription Initiation to Elongation
The RNA polymerase II (pol II) A form contains an unphosphorylated carboxy-terminal domain (CTD) which initiates transcription producing 20-25 nucleotide 5’-triphosphorylated transcripts. The CTD is then phosphorylated to the RNA pol II O form, changing conformation and binding to RNA capping enzyme (CE). Stimulated by the elongation factor SPT5 and the phosphorylated CTD, CE which has RNA triphosphatase and RNA guanylyltransferase activities modifies the exposed end of the nascent transcript to GpppN. Methyl transferase (MT) binds to CE and to the terminal GpppN. Finally, Importin-a (Impa) stimulates N7 methylation of the cap GpppN by MT, and RNA pol II O form switches from the initiation process to elongation. In addition, the pol II CTD also binds splicing factors and polyadenylation factors. Hence, the pol II CTD is central to the cotranscriptional nature of RNA processing (5’ capping, splicing and 3’polyadenylation).
Back to Top