Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Cell Biology > Learning Center > Pathway Slides & Charts  > Possible Receptor Combos
Cell Signaling & Neuroscience
Possible Receptor Combos
Top Border
Left Border  Right Border
Left Corner Back Download Forward Sigma-Aldrich Right Corner
Figure-3b
Possible Receptor Combinations and Ligand—binding with EGF and HGF Families
Homodimeric and heterodimeric combinations of the four receptors (EGFR, ErbB2, ErbB3 and ErbB4) showing high-affinity binding ligands (IC50 > 100nM) measured by Jones, et al. (1999).
Back to Top