Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Cell Biology > Learning Center > Pathway Slides & Charts  > PPAR Signaling Pathway
Cell Signaling & Neuroscience
PPAR Signaling Pathway
Top Border
Left Border  Right Border
Left Corner Back Download Forward Sigma-Aldrich Right Corner
Figure-3b
PPAR Signaling Pathway
Similar to other nuclear hormone receptors, peroxisome proliferator-activated receptors (PPARs) act as ligand-activated transcription factors. When bound to its fatty acid ligand, PPARa forms a heterodimeric complex with the retinoid X receptor (RXR) to regulate transcription. PPARg is activated by prostaglandins and leukotrienes and regulates the gene expression of proteins involved in the storage of fatty acids. PARb is weakly activated by fatty acids, prostaglandins, and leukotrienes. Its physiological ligand has not been identified.
Back to Top
References:
Fajas, L., et al., Peroxisome proliferator-activated receptor-g: from adipogenesis to carcinogenesis. J. Mol. Endocrinol., 27, 1-9 (2001).
Qi, C., et al., Peroxisome proliferator-activated receptors, coactivators, and downstream targets. Cell. Biochem. Biophys., 32, 187-204 (2000).