Sigma-Aldrich
Life Science > Cell Biology > Learning Center > Pathway Slides & Charts  > PTEN Pathway
Cell Signaling & Neuroscience
PTEN Pathway
Top Border
Left Border  Right Border
Left Corner Back Download Forward Sigma-Aldrich Right Corner
Figure-3b
PTEN Pathway
PTEN is a tumor suppressor gene that is able to dephosphorylate phosphatidylinositol 3,4,5-trisphosphate (PI-3,4,5-P3), the product of phosphatidyl inositol 3-kinase (PIK). Many of the mutations that have arisen in cancer cells have been mapped to the phosphatase catalytic domain of PTEN. Data suggests that the phosphatase activity of PTEN is essential for its function as a tumor suppressor. The activation of Akt/PKB is regulated by the phosphorylation of Akt on Thr308 and Ser473 by phosphoinositide-dependent kinase (PDK) and integrin-linked kinase (ILK), respectively. Inactivation of PTEN allows constitutive and unregulated activation of the Akt/PKB signaling pathway. In addition to regulating the Akt/PKB signaling pathway, PTEN also inhibits growth factor (GF)-induced Shc phosphorylation and suppresses the MAP kinase signaling pathway. PTEN interacts directly with FAK and is able to dephosphorylate activated FAK. PTEN-induced down-regulation of p130CAS through FAK results in inhibition of cell migration and spreading.
Back to Top
References:
Simpson, L., and Parsons, R., PTEN: life as a tumor suppressor. Exp. Cell Res., 264, 29-41 (2001).
Besson, A., et al., PTEN/MMAC1/TEP1 in signal transduction and tumorigenesis. Eur. J. Biochem., 263, 605-611 (1999).